Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14506033-14506534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLBD1
Entrez Gene ID
79887
Gene Name
phospholipase B domain containing 1
Gene Aliases
-
UniGene
Hs.131933
Gene Chromosome Location
Chr.12: 14503663-14567857 on Build GRCh38
Transcripts
NM_024829.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1360405 c.1449C>A p.N483K Chr.12: 14506192-14506192 on Build GRCh38
459599 c.1388C>A p.P463H Chr.12: 14506253-14506253 on Build GRCh38
2002890 c.1394G>A p.S465N Chr.12: 14506247-14506247 on Build GRCh38
4821774 c.1429G>A p.E477K Chr.12: 14506212-14506212 on Build GRCh38
5840640 c.1405C>T p.P469S Chr.12: 14506236-14506236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768632036 Chr.12: 14506243-14506243 on Build GRCh38
rs113022440 Chr.12: 14506299-14506299 on Build GRCh38
rs561023570 Chr.12: 14506116-14506116 on Build GRCh38
rs758804884 Chr.12: 14506279-14506279 on Build GRCh38
rs762570332 Chr.12: 14506181-14506181 on Build GRCh38
rs985313544 Chr.12: 14506066-14506066 on Build GRCh38
rs370331063 Chr.12: 14506310-14506310 on Build GRCh38
rs749213844 Chr.12: 14506242-14506242 on Build GRCh38
rs762004759 Chr.12: 14506162-14506162 on Build GRCh38
rs560503408 Chr.12: 14506107-14506107 on Build GRCh38
rs774106135 Chr.12: 14506246-14506246 on Build GRCh38
rs747528 Chr.12: 14506507-14506507 on Build GRCh38
rs778610835 Chr.12: 14506110-14506110 on Build GRCh38
rs3835045 Chr.12: 14506277-14506278 on Build GRCh38
rs149722587 Chr.12: 14506214-14506214 on Build GRCh38
rs760818746 Chr.12: 14506179-14506179 on Build GRCh38
rs373332103 Chr.12: 14506308-14506308 on Build GRCh38
rs573779703 Chr.12: 14506452-14506452 on Build GRCh38
rs115795697 Chr.12: 14506272-14506272 on Build GRCh38
rs773113855 Chr.12: 14506171-14506171 on Build GRCh38
rs555216090 Chr.12: 14506270-14506271 on Build GRCh38
rs1003699886 Chr.12: 14506461-14506461 on Build GRCh38
rs906256387 Chr.12: 14506409-14506409 on Build GRCh38
rs935047943 Chr.12: 14506098-14506098 on Build GRCh38
rs767824640 Chr.12: 14506165-14506165 on Build GRCh38
rs1040985749 Chr.12: 14506073-14506073 on Build GRCh38
rs756819219 Chr.12: 14506210-14506210 on Build GRCh38
rs767013433 Chr.12: 14506281-14506281 on Build GRCh38
rs746915906 Chr.12: 14506174-14506174 on Build GRCh38
rs755598599 Chr.12: 14506189-14506189 on Build GRCh38
rs754389787 Chr.12: 14506183-14506183 on Build GRCh38
rs1011940179 Chr.12: 14506115-14506115 on Build GRCh38
rs76775307 Chr.12: 14506067-14506067 on Build GRCh38
rs1044793369 Chr.12: 14506394-14506394 on Build GRCh38
rs376855043 Chr.12: 14506305-14506305 on Build GRCh38
rs914906862 Chr.12: 14506382-14506382 on Build GRCh38
rs1002570921 Chr.12: 14506375-14506375 on Build GRCh38
rs1036934150 Chr.12: 14506506-14506506 on Build GRCh38
rs775080311 Chr.12: 14506302-14506302 on Build GRCh38
rs748840920 Chr.12: 14506301-14506301 on Build GRCh38
rs546578493 Chr.12: 14506273-14506273 on Build GRCh38
rs1023310193 Chr.12: 14506130-14506130 on Build GRCh38
rs750448495 Chr.12: 14506114-14506114 on Build GRCh38
rs776899191 Chr.12: 14506269-14506269 on Build GRCh38
rs372438193 Chr.12: 14506247-14506247 on Build GRCh38
rs766726585 Chr.12: 14506180-14506180 on Build GRCh38
rs765672431 Chr.12: 14506191-14506191 on Build GRCh38
rs773484660 Chr.12: 14506249-14506249 on Build GRCh38
rs768439128 Chr.12: 14506141-14506141 on Build GRCh38
rs150751009 Chr.12: 14506419-14506419 on Build GRCh38
rs79900625 Chr.12: 14506384-14506384 on Build GRCh38
rs898969717 Chr.12: 14506462-14506462 on Build GRCh38
rs376671162 Chr.12: 14506248-14506248 on Build GRCh38
rs762670711 Chr.12: 14506128-14506128 on Build GRCh38
rs536838891 Chr.12: 14506395-14506395 on Build GRCh38
rs565623271 Chr.12: 14506334-14506334 on Build GRCh38
rs760870857 Chr.12: 14506265-14506265 on Build GRCh38
rs943829996 Chr.12: 14506072-14506072 on Build GRCh38
rs192458082 Chr.12: 14506338-14506338 on Build GRCh38
rs973153630 Chr.12: 14506076-14506076 on Build GRCh38
rs753152994 Chr.12: 14506193-14506193 on Build GRCh38
rs902395986 Chr.12: 14506089-14506089 on Build GRCh38
rs755656884 Chr.12: 14506215-14506215 on Build GRCh38
rs558568180 Chr.12: 14506307-14506307 on Build GRCh38
rs750087224 Chr.12: 14506286-14506286 on Build GRCh38
rs886397004 Chr.12: 14506412-14506412 on Build GRCh38
rs906726542 Chr.12: 14506131-14506131 on Build GRCh38
rs878920958 Chr.12: 14506293-14506293 on Build GRCh38
rs751593849 Chr.12: 14506315-14506316 on Build GRCh38
rs766790765 Chr.12: 14506173-14506175 on Build GRCh38
rs1004765210 Chr.12: 14506455-14506455 on Build GRCh38
rs747230399 Chr.12: 14506312-14506312 on Build GRCh38
rs571267090 Chr.12: 14506292-14506292 on Build GRCh38
rs202073366 Chr.12: 14506155-14506155 on Build GRCh38
rs779607205 Chr.12: 14506224-14506224 on Build GRCh38
rs549488937 Chr.12: 14506104-14506104 on Build GRCh38

View Full Product Details