Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27780897-27781392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL42
Entrez Gene ID
57542
Gene Name
kelch like family member 42
Gene Aliases
Ctb9, KLHDC5
UniGene
Hs.505104
Gene Chromosome Location
Chr.12: 27779957-27803040 on Build GRCh38
Transcripts
NM_020782.1 XM_017019698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7181251 c.654C>T p.S218S Chr.12: 27780984-27780984 on Build GRCh38
2204357 c.824A>G p.N275S Chr.12: 27781154-27781154 on Build GRCh38
4696399 c.830G>A p.S277N Chr.12: 27781160-27781160 on Build GRCh38
7174136 c.643G>A p.E215K Chr.12: 27780973-27780973 on Build GRCh38
7167140 c.723C>G p.V241V Chr.12: 27781053-27781053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752980706 Chr.12: 27781233-27781233 on Build GRCh38
rs748918755 Chr.12: 27781176-27781176 on Build GRCh38
rs767415804 Chr.12: 27781241-27781241 on Build GRCh38
rs772500023 Chr.12: 27781210-27781210 on Build GRCh38
rs1057131661 Chr.12: 27781023-27781023 on Build GRCh38
rs761329921 Chr.12: 27781212-27781212 on Build GRCh38
rs759707638 Chr.12: 27781229-27781229 on Build GRCh38
rs372576711 Chr.12: 27781189-27781189 on Build GRCh38
rs773773298 Chr.12: 27781114-27781114 on Build GRCh38
rs756094561 Chr.12: 27781250-27781250 on Build GRCh38
rs776041154 Chr.12: 27780939-27780939 on Build GRCh38
rs757794447 Chr.12: 27781230-27781231 on Build GRCh38
rs998851175 Chr.12: 27781236-27781236 on Build GRCh38
rs976840090 Chr.12: 27781099-27781099 on Build GRCh38
rs781208420 Chr.12: 27781062-27781062 on Build GRCh38
rs551232636 Chr.12: 27780937-27780937 on Build GRCh38
rs759874141 Chr.12: 27781135-27781135 on Build GRCh38
rs200244285 Chr.12: 27781184-27781184 on Build GRCh38
rs748393828 Chr.12: 27781208-27781208 on Build GRCh38
rs984222858 Chr.12: 27781331-27781331 on Build GRCh38
rs746975250 Chr.12: 27780925-27780925 on Build GRCh38
rs538490560 Chr.12: 27781107-27781107 on Build GRCh38
rs371309752 Chr.12: 27781058-27781058 on Build GRCh38
rs757187149 Chr.12: 27781059-27781059 on Build GRCh38
rs749010339 Chr.12: 27781090-27781090 on Build GRCh38
rs555315365 Chr.12: 27781317-27781317 on Build GRCh38
rs371189819 Chr.12: 27781169-27781169 on Build GRCh38
rs149062131 Chr.12: 27781137-27781137 on Build GRCh38
rs750436742 Chr.12: 27781165-27781165 on Build GRCh38
rs377651287 Chr.12: 27781140-27781140 on Build GRCh38
rs754312725 Chr.12: 27781202-27781203 on Build GRCh38
rs765512002 Chr.12: 27781230-27781230 on Build GRCh38
rs750421038 Chr.12: 27781029-27781029 on Build GRCh38
rs767613843 Chr.12: 27781020-27781020 on Build GRCh38
rs764208505 Chr.12: 27781134-27781136 on Build GRCh38
rs17852612 Chr.12: 27781164-27781164 on Build GRCh38
rs937353248 Chr.12: 27781009-27781009 on Build GRCh38
rs991883912 Chr.12: 27781011-27781011 on Build GRCh38
rs1006677010 Chr.12: 27781092-27781092 on Build GRCh38
rs763249165 Chr.12: 27780992-27780992 on Build GRCh38
rs773820049 Chr.12: 27781044-27781044 on Build GRCh38
rs201189833 Chr.12: 27781228-27781228 on Build GRCh38
rs927389840 Chr.12: 27781315-27781316 on Build GRCh38
rs754124260 Chr.12: 27781042-27781042 on Build GRCh38
rs539596707 Chr.12: 27781067-27781067 on Build GRCh38
rs984170691 Chr.12: 27780976-27780976 on Build GRCh38
rs201456188 Chr.12: 27781093-27781093 on Build GRCh38
rs757102815 Chr.12: 27781142-27781142 on Build GRCh38
rs375949235 Chr.12: 27781132-27781132 on Build GRCh38
rs764503221 Chr.12: 27780997-27780997 on Build GRCh38
rs891730898 Chr.12: 27781024-27781024 on Build GRCh38
rs200299795 Chr.12: 27781211-27781211 on Build GRCh38
rs761504811 Chr.12: 27781235-27781235 on Build GRCh38
rs1046262740 Chr.12: 27781057-27781057 on Build GRCh38
rs772419743 Chr.12: 27781113-27781113 on Build GRCh38
rs1057183300 Chr.12: 27781321-27781321 on Build GRCh38
rs1042989111 Chr.12: 27781340-27781340 on Build GRCh38
rs774837373 Chr.12: 27781013-27781013 on Build GRCh38
rs534203092 Chr.12: 27781134-27781134 on Build GRCh38
rs756248836 Chr.12: 27781032-27781032 on Build GRCh38
rs777794545 Chr.12: 27780924-27780924 on Build GRCh38
rs762267669 Chr.12: 27781015-27781015 on Build GRCh38
rs573217515 Chr.12: 27781116-27781116 on Build GRCh38
rs904407772 Chr.12: 27781349-27781349 on Build GRCh38
rs750300268 Chr.12: 27781246-27781246 on Build GRCh38
rs143061675 Chr.12: 27781161-27781161 on Build GRCh38
rs557155929 Chr.12: 27781362-27781362 on Build GRCh38
rs779582619 Chr.12: 27781174-27781174 on Build GRCh38
rs771101033 Chr.12: 27781220-27781220 on Build GRCh38
rs774747728 Chr.12: 27781095-27781095 on Build GRCh38
rs1021494561 Chr.12: 27781060-27781060 on Build GRCh38
rs769277069 Chr.12: 27780963-27780963 on Build GRCh38
rs909423083 Chr.12: 27781224-27781224 on Build GRCh38
rs146472087 Chr.12: 27780990-27780990 on Build GRCh38
rs569081413 Chr.12: 27781053-27781053 on Build GRCh38
rs916527623 Chr.12: 27781028-27781028 on Build GRCh38
rs779671062 Chr.12: 27781077-27781077 on Build GRCh38
rs745508584 Chr.12: 27780948-27780948 on Build GRCh38
rs745310781 Chr.12: 27781064-27781064 on Build GRCh38
rs766641437 Chr.12: 27781035-27781035 on Build GRCh38

View Full Product Details