Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2592247-2592748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
UniGene
Hs.591453
Gene Chromosome Location
Chr.1: 2590639-2633042 on Build GRCh38
Transcripts
NM_033467.3 XM_011542122.2 XM_017002310.1 XM_017002311.1 XM_017002312.1 XM_017002313.1 XM_017002314.1 XM_017002315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686896 c.1999G>A p.E667K Chr.1: 2592696-2592696 on Build GRCh38
4781939 c.2056C>A p.Q686K Chr.1: 2592666-2592666 on Build GRCh38
4702648 c.2020C>A p.L674I Chr.1: 2592702-2592702 on Build GRCh38
4702649 c.1993C>A p.L665I Chr.1: 2592702-2592702 on Build GRCh38
4475818 c.1992C>T p.T664T Chr.1: 2592703-2592703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979594442 Chr.1: 2592453-2592453 on Build GRCh38
rs193231151 Chr.1: 2592560-2592560 on Build GRCh38
rs758873133 Chr.1: 2592672-2592672 on Build GRCh38
rs889176469 Chr.1: 2592614-2592614 on Build GRCh38
rs184574713 Chr.1: 2592530-2592530 on Build GRCh38
rs372927351 Chr.1: 2592600-2592600 on Build GRCh38
rs766390484 Chr.1: 2592640-2592640 on Build GRCh38
rs187339910 Chr.1: 2592439-2592439 on Build GRCh38
rs374407417 Chr.1: 2592539-2592540 on Build GRCh38
rs375953913 Chr.1: 2592723-2592723 on Build GRCh38
rs758612411 Chr.1: 2592605-2592622 on Build GRCh38
rs148588192 Chr.1: 2592718-2592718 on Build GRCh38
rs769395265 Chr.1: 2592679-2592679 on Build GRCh38
rs112218652 Chr.1: 2592305-2592305 on Build GRCh38
rs374695686 Chr.1: 2592586-2592608 on Build GRCh38
rs149776280 Chr.1: 2592689-2592689 on Build GRCh38
rs937839569 Chr.1: 2592538-2592538 on Build GRCh38
rs577279334 Chr.1: 2592714-2592714 on Build GRCh38
rs562955707 Chr.1: 2592470-2592470 on Build GRCh38
rs192349427 Chr.1: 2592368-2592368 on Build GRCh38
rs115709124 Chr.1: 2592725-2592725 on Build GRCh38
rs745688096 Chr.1: 2592630-2592643 on Build GRCh38
rs557533195 Chr.1: 2592690-2592690 on Build GRCh38
rs185684435 Chr.1: 2592608-2592608 on Build GRCh38
rs543130237 Chr.1: 2592715-2592715 on Build GRCh38
rs549629773 Chr.1: 2592512-2592512 on Build GRCh38
rs182464181 Chr.1: 2592371-2592371 on Build GRCh38
rs567675473 Chr.1: 2592563-2592563 on Build GRCh38
rs775061503 Chr.1: 2592672-2592672 on Build GRCh38
rs566160325 Chr.1: 2592282-2592282 on Build GRCh38
rs755158932 Chr.1: 2592659-2592659 on Build GRCh38
rs199913369 Chr.1: 2592331-2592331 on Build GRCh38
rs866551031 Chr.1: 2592633-2592633 on Build GRCh38
rs930620860 Chr.1: 2592606-2592606 on Build GRCh38
rs371392756 Chr.1: 2592632-2592632 on Build GRCh38
rs867435 Chr.1: 2592267-2592267 on Build GRCh38
rs939960971 Chr.1: 2592585-2592585 on Build GRCh38
rs572186597 Chr.1: 2592402-2592402 on Build GRCh38
rs566278049 Chr.1: 2592514-2592514 on Build GRCh38
rs1036940368 Chr.1: 2592591-2592591 on Build GRCh38
rs867436 Chr.1: 2592284-2592284 on Build GRCh38
rs776509786 Chr.1: 2592620-2592628 on Build GRCh38
rs777995220 Chr.1: 2592607-2592623 on Build GRCh38
rs764008973 Chr.1: 2592695-2592695 on Build GRCh38
rs747538821 Chr.1: 2592675-2592675 on Build GRCh38
rs182656657 Chr.1: 2592424-2592424 on Build GRCh38
rs770935636 Chr.1: 2592609-2592632 on Build GRCh38
rs774379311 Chr.1: 2592699-2592699 on Build GRCh38
rs146100312 Chr.1: 2592668-2592668 on Build GRCh38
rs369979738 Chr.1: 2592724-2592724 on Build GRCh38
rs765417492 Chr.1: 2592720-2592720 on Build GRCh38
rs140073157 Chr.1: 2592379-2592379 on Build GRCh38
rs754033011 Chr.1: 2592652-2592652 on Build GRCh38
rs776012302 Chr.1: 2592678-2592678 on Build GRCh38
rs9730673 Chr.1: 2592400-2592400 on Build GRCh38
rs770419715 Chr.1: 2592677-2592677 on Build GRCh38
rs187786174 Chr.1: 2592372-2592372 on Build GRCh38
rs571719524 Chr.1: 2592547-2592547 on Build GRCh38
rs868390878 Chr.1: 2592337-2592337 on Build GRCh38
rs377631947 Chr.1: 2592630-2592630 on Build GRCh38
rs187759598 Chr.1: 2592554-2592554 on Build GRCh38
rs897857437 Chr.1: 2592602-2592602 on Build GRCh38
rs757775067 Chr.1: 2592727-2592727 on Build GRCh38
rs966293429 Chr.1: 2592313-2592313 on Build GRCh38
rs201706260 Chr.1: 2592666-2592666 on Build GRCh38
rs867247356 Chr.1: 2592338-2592338 on Build GRCh38
rs1014449977 Chr.1: 2592617-2592617 on Build GRCh38
rs191966582 Chr.1: 2592440-2592440 on Build GRCh38
rs746948980 Chr.1: 2592608-2592628 on Build GRCh38
rs536280138 Chr.1: 2592622-2592622 on Build GRCh38
rs778213405 Chr.1: 2592673-2592673 on Build GRCh38
rs563829676 Chr.1: 2592438-2592438 on Build GRCh38
rs753932423 Chr.1: 2592716-2592716 on Build GRCh38
rs557394064 Chr.1: 2592323-2592323 on Build GRCh38
rs769607498 Chr.1: 2592630-2592631 on Build GRCh38
rs528675131 Chr.1: 2592516-2592516 on Build GRCh38

View Full Product Details