Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2596427-2596925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
UniGene
Hs.591453
Gene Chromosome Location
Chr.1: 2590639-2633042 on Build GRCh38
Transcripts
NM_033467.3 XM_011542122.2 XM_017002310.1 XM_017002311.1 XM_017002312.1 XM_017002313.1 XM_017002314.1 XM_017002315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4424027 c.1293G>A p.V431V Chr.1: 2596669-2596669 on Build GRCh38
1686897 c.1331C>T p.S444F Chr.1: 2596604-2596604 on Build GRCh38
1602108 c.1351T>C p.F451L Chr.1: 2596584-2596584 on Build GRCh38
4873987 c.1303C>T p.R435C Chr.1: 2596659-2596659 on Build GRCh38
4780933 c.1378T>C p.F460L Chr.1: 2596584-2596584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780405370 Chr.1: 2596658-2596658 on Build GRCh38
rs751827091 Chr.1: 2596564-2596564 on Build GRCh38
rs907384460 Chr.1: 2596740-2596740 on Build GRCh38
rs948867170 Chr.1: 2596714-2596714 on Build GRCh38
rs529568137 Chr.1: 2596452-2596452 on Build GRCh38
rs149373691 Chr.1: 2596463-2596463 on Build GRCh38
rs781594128 Chr.1: 2596583-2596583 on Build GRCh38
rs759613852 Chr.1: 2596631-2596631 on Build GRCh38
rs530858345 Chr.1: 2596855-2596856 on Build GRCh38
rs774947812 Chr.1: 2596526-2596526 on Build GRCh38
rs902011512 Chr.1: 2596867-2596867 on Build GRCh38
rs770771638 Chr.1: 2596614-2596614 on Build GRCh38
rs550701899 Chr.1: 2596705-2596705 on Build GRCh38
rs1006284002 Chr.1: 2596823-2596823 on Build GRCh38
rs369019258 Chr.1: 2596550-2596550 on Build GRCh38
rs1041304159 Chr.1: 2596902-2596902 on Build GRCh38
rs770845423 Chr.1: 2596716-2596716 on Build GRCh38
rs760640101 Chr.1: 2596538-2596538 on Build GRCh38
rs577522408 Chr.1: 2596842-2596842 on Build GRCh38
rs762060719 Chr.1: 2596636-2596636 on Build GRCh38
rs759527524 Chr.1: 2596860-2596860 on Build GRCh38
rs761464416 Chr.1: 2596534-2596534 on Build GRCh38
rs549715803 Chr.1: 2596857-2596857 on Build GRCh38
rs766269962 Chr.1: 2596540-2596540 on Build GRCh38
rs144887855 Chr.1: 2596596-2596596 on Build GRCh38
rs760348798 Chr.1: 2596612-2596612 on Build GRCh38
rs992702153 Chr.1: 2596777-2596777 on Build GRCh38
rs911560131 Chr.1: 2596865-2596865 on Build GRCh38
rs936044547 Chr.1: 2596499-2596499 on Build GRCh38
rs749812350 Chr.1: 2596587-2596587 on Build GRCh38
rs779195272 Chr.1: 2596476-2596476 on Build GRCh38
rs551888822 Chr.1: 2596615-2596615 on Build GRCh38
rs767651036 Chr.1: 2596726-2596726 on Build GRCh38
rs768091793 Chr.1: 2596590-2596590 on Build GRCh38
rs984530287 Chr.1: 2596574-2596574 on Build GRCh38
rs182328090 Chr.1: 2596858-2596858 on Build GRCh38
rs891952881 Chr.1: 2596679-2596679 on Build GRCh38
rs956227889 Chr.1: 2596482-2596482 on Build GRCh38
rs564385423 Chr.1: 2596557-2596557 on Build GRCh38
rs2843401 Chr.1: 2596694-2596694 on Build GRCh38
rs778466486 Chr.1: 2596591-2596591 on Build GRCh38
rs771646217 Chr.1: 2596704-2596704 on Build GRCh38
rs146652643 Chr.1: 2596687-2596687 on Build GRCh38
rs139384428 Chr.1: 2596611-2596611 on Build GRCh38
rs759519916 Chr.1: 2596720-2596720 on Build GRCh38
rs961950413 Chr.1: 2596814-2596814 on Build GRCh38
rs781472444 Chr.1: 2596515-2596515 on Build GRCh38
rs1050635653 Chr.1: 2596853-2596853 on Build GRCh38
rs975857433 Chr.1: 2596885-2596885 on Build GRCh38
rs147290642 Chr.1: 2596533-2596533 on Build GRCh38
rs375550510 Chr.1: 2596532-2596532 on Build GRCh38
rs748372222 Chr.1: 2596507-2596507 on Build GRCh38
rs369116437 Chr.1: 2596645-2596645 on Build GRCh38
rs907705191 Chr.1: 2596554-2596554 on Build GRCh38
rs764252107 Chr.1: 2596632-2596632 on Build GRCh38
rs528720649 Chr.1: 2596555-2596555 on Build GRCh38
rs907819714 Chr.1: 2596780-2596780 on Build GRCh38
rs761748162 Chr.1: 2596725-2596725 on Build GRCh38
rs372795128 Chr.1: 2596535-2596535 on Build GRCh38
rs772939933 Chr.1: 2596537-2596537 on Build GRCh38
rs747653777 Chr.1: 2596695-2596695 on Build GRCh38
rs750586927 Chr.1: 2596883-2596883 on Build GRCh38
rs903185319 Chr.1: 2596494-2596494 on Build GRCh38
rs879890516 Chr.1: 2596498-2596498 on Build GRCh38
rs751735070 Chr.1: 2596635-2596635 on Build GRCh38
rs747823892 Chr.1: 2596594-2596594 on Build GRCh38
rs535402049 Chr.1: 2596747-2596747 on Build GRCh38
rs373512211 Chr.1: 2596604-2596604 on Build GRCh38
rs1026539376 Chr.1: 2596757-2596757 on Build GRCh38
rs756379955 Chr.1: 2596653-2596653 on Build GRCh38
rs745803260 Chr.1: 2596627-2596652 on Build GRCh38
rs756530819 Chr.1: 2596586-2596586 on Build GRCh38
rs924575361 Chr.1: 2596663-2596663 on Build GRCh38
rs781039832 Chr.1: 2596532-2596553 on Build GRCh38
rs770249848 Chr.1: 2596524-2596524 on Build GRCh38
rs746677725 Chr.1: 2596715-2596715 on Build GRCh38
rs776625643 Chr.1: 2596718-2596718 on Build GRCh38
rs12133956 Chr.1: 2596901-2596901 on Build GRCh38
rs1013483061 Chr.1: 2596511-2596511 on Build GRCh38
rs868849803 Chr.1: 2596560-2596560 on Build GRCh38
rs569893985 Chr.1: 2596746-2596746 on Build GRCh38
rs888060177 Chr.1: 2596765-2596765 on Build GRCh38
rs757474320 Chr.1: 2596565-2596565 on Build GRCh38
rs377209234 Chr.1: 2596547-2596547 on Build GRCh38
rs150030274 Chr.1: 2596652-2596652 on Build GRCh38
rs1008900576 Chr.1: 2596453-2596454 on Build GRCh38
rs775525581 Chr.1: 2596724-2596724 on Build GRCh38
rs746369278 Chr.1: 2596520-2596520 on Build GRCh38
rs140870456 Chr.1: 2596685-2596685 on Build GRCh38
rs920426085 Chr.1: 2596826-2596826 on Build GRCh38
rs978019025 Chr.1: 2596662-2596662 on Build GRCh38
rs1002025852 Chr.1: 2596735-2596735 on Build GRCh38
rs371381022 Chr.1: 2596585-2596585 on Build GRCh38
rs772146599 Chr.1: 2596509-2596509 on Build GRCh38
rs1054960845 Chr.1: 2596503-2596503 on Build GRCh38
rs771001611 Chr.1: 2596688-2596688 on Build GRCh38
rs1034321173 Chr.1: 2596751-2596751 on Build GRCh38

View Full Product Details