Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6264579-6265073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT7
Entrez Gene ID
11332
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
UniGene
Hs.126137
Gene Chromosome Location
Chr.1: 6264272-6393766 on Build GRCh38
Transcripts
NM_007274.3 NM_181864.2 NM_181865.2 NM_181866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6315826 c.1122C>A p.G374G Chr.1: 6264618-6264618 on Build GRCh38
6325507 c.1023G>A p.K341K Chr.1: 6264627-6264627 on Build GRCh38
6315829 c.969C>A p.G323G Chr.1: 6264618-6264618 on Build GRCh38
1639938 c.1000C>T p.R334W Chr.1: 6264650-6264650 on Build GRCh38
295968 c.1060C>T p.R354W Chr.1: 6264650-6264650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043268841 Chr.1: 6264787-6264787 on Build GRCh38
rs745661134 Chr.1: 6264678-6264678 on Build GRCh38
rs139667005 Chr.1: 6264613-6264613 on Build GRCh38
rs112755158 Chr.1: 6264962-6264962 on Build GRCh38
rs529107914 Chr.1: 6264748-6264748 on Build GRCh38
rs759822992 Chr.1: 6264723-6264723 on Build GRCh38
rs761807487 Chr.1: 6264608-6264608 on Build GRCh38
rs755598589 Chr.1: 6264615-6264615 on Build GRCh38
rs1041409990 Chr.1: 6264811-6264811 on Build GRCh38
rs772998693 Chr.1: 6264613-6264613 on Build GRCh38
rs771262821 Chr.1: 6264712-6264712 on Build GRCh38
rs61739537 Chr.1: 6264687-6264687 on Build GRCh38
rs569124772 Chr.1: 6264807-6264807 on Build GRCh38
rs146498455 Chr.1: 6264693-6264693 on Build GRCh38
rs1005324022 Chr.1: 6264831-6264831 on Build GRCh38
rs1032329132 Chr.1: 6264822-6264822 on Build GRCh38
rs373755282 Chr.1: 6264722-6264722 on Build GRCh38
rs888340391 Chr.1: 6264999-6264999 on Build GRCh38
rs386628058 Chr.1: 6264908-6264909 on Build GRCh38
rs367593933 Chr.1: 6264731-6264731 on Build GRCh38
rs773391670 Chr.1: 6264695-6264695 on Build GRCh38
rs1035061099 Chr.1: 6264869-6264869 on Build GRCh38
rs968894235 Chr.1: 6264807-6264807 on Build GRCh38
rs1003304228 Chr.1: 6264856-6264856 on Build GRCh38
rs1035401771 Chr.1: 6264778-6264778 on Build GRCh38
rs745847219 Chr.1: 6264626-6264626 on Build GRCh38
rs982161357 Chr.1: 6264692-6264692 on Build GRCh38
rs921265055 Chr.1: 6264841-6264841 on Build GRCh38
rs1015285625 Chr.1: 6264686-6264686 on Build GRCh38
rs556518865 Chr.1: 6265044-6265044 on Build GRCh38
rs759452981 Chr.1: 6264947-6264947 on Build GRCh38
rs916189981 Chr.1: 6264777-6264777 on Build GRCh38
rs774559938 Chr.1: 6264607-6264607 on Build GRCh38
rs866773889 Chr.1: 6264859-6264859 on Build GRCh38
rs201437597 Chr.1: 6264605-6264605 on Build GRCh38
rs189098509 Chr.1: 6264875-6264875 on Build GRCh38
rs997293098 Chr.1: 6264819-6264819 on Build GRCh38
rs934743117 Chr.1: 6264797-6264797 on Build GRCh38
rs932655983 Chr.1: 6264992-6264992 on Build GRCh38
rs1016400936 Chr.1: 6264724-6264724 on Build GRCh38
rs1041131485 Chr.1: 6264963-6264963 on Build GRCh38
rs578211150 Chr.1: 6264631-6264631 on Build GRCh38
rs977153216 Chr.1: 6264838-6264838 on Build GRCh38
rs763744988 Chr.1: 6265015-6265015 on Build GRCh38
rs141093267 Chr.1: 6264788-6264788 on Build GRCh38
rs74049517 Chr.1: 6264998-6264998 on Build GRCh38
rs765656734 Chr.1: 6264622-6264622 on Build GRCh38
rs12061848 Chr.1: 6264908-6264908 on Build GRCh38
rs370641390 Chr.1: 6264776-6264800 on Build GRCh38
rs959069001 Chr.1: 6264892-6264892 on Build GRCh38
rs111940368 Chr.1: 6264864-6264865 on Build GRCh38
rs1050175766 Chr.1: 6264736-6264736 on Build GRCh38
rs183552627 Chr.1: 6264705-6264705 on Build GRCh38
rs991759635 Chr.1: 6264749-6264749 on Build GRCh38
rs756883014 Chr.1: 6264649-6264649 on Build GRCh38
rs781365502 Chr.1: 6264780-6264780 on Build GRCh38
rs774046612 Chr.1: 6264832-6264832 on Build GRCh38
rs1028249860 Chr.1: 6265025-6265025 on Build GRCh38
rs896999040 Chr.1: 6265024-6265024 on Build GRCh38
rs978894100 Chr.1: 6264918-6264918 on Build GRCh38
rs376418225 Chr.1: 6264623-6264623 on Build GRCh38
rs968974379 Chr.1: 6264912-6264912 on Build GRCh38
rs192112480 Chr.1: 6264614-6264614 on Build GRCh38
rs748420060 Chr.1: 6264826-6264826 on Build GRCh38
rs932702851 Chr.1: 6264858-6264858 on Build GRCh38
rs12063157 Chr.1: 6265038-6265038 on Build GRCh38
rs559578865 Chr.1: 6264827-6264827 on Build GRCh38
rs555617854 Chr.1: 6264848-6264848 on Build GRCh38
rs977037648 Chr.1: 6265017-6265017 on Build GRCh38
rs12059771 Chr.1: 6264909-6264909 on Build GRCh38
rs765578344 Chr.1: 6264738-6264738 on Build GRCh38
rs371091558 Chr.1: 6264612-6264612 on Build GRCh38
rs556695044 Chr.1: 6264630-6264630 on Build GRCh38
rs776183530 Chr.1: 6264711-6264712 on Build GRCh38
rs367702357 Chr.1: 6264905-6264905 on Build GRCh38
rs561595933 Chr.1: 6264743-6264743 on Build GRCh38
rs376793853 Chr.1: 6264606-6264606 on Build GRCh38
rs761587392 Chr.1: 6265050-6265050 on Build GRCh38
rs74658121 Chr.1: 6265002-6265002 on Build GRCh38
rs760321703 Chr.1: 6264640-6264640 on Build GRCh38
rs776380865 Chr.1: 6264602-6264602 on Build GRCh38
rs61739540 Chr.1: 6264688-6264688 on Build GRCh38
rs145849239 Chr.1: 6264681-6264681 on Build GRCh38
rs901176628 Chr.1: 6264815-6264815 on Build GRCh38
rs373266176 Chr.1: 6264601-6264601 on Build GRCh38
rs74049516 Chr.1: 6264981-6264981 on Build GRCh38
rs566534709 Chr.1: 6264948-6264948 on Build GRCh38
rs909026191 Chr.1: 6264865-6264865 on Build GRCh38
rs1036793129 Chr.1: 6265023-6265023 on Build GRCh38
rs767579371 Chr.1: 6264611-6264611 on Build GRCh38
rs965972789 Chr.1: 6265009-6265009 on Build GRCh38
rs765970009 Chr.1: 6264693-6264693 on Build GRCh38
rs941849337 Chr.1: 6264870-6264870 on Build GRCh38
rs757859106 Chr.1: 6264625-6264625 on Build GRCh38
rs781024703 Chr.1: 6264650-6264650 on Build GRCh38

View Full Product Details