Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6460136-6460681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPN
Entrez Gene ID
83715
Gene Name
espin
Gene Aliases
DFNB36, LP2654
UniGene
Hs.744222
Gene Chromosome Location
Chr.1: 6424776-6461367 on Build GRCh38
Transcripts
NM_031475.2 XM_005263501.2 XM_017002433.1

SNP Detail

SNP ID Chromosome Location
rs756804502 Chr.1: 6460196-6460196 on Build GRCh38
rs931020982 Chr.1: 6460288-6460289 on Build GRCh38
rs1046535803 Chr.1: 6460515-6460515 on Build GRCh38
rs913023966 Chr.1: 6460238-6460238 on Build GRCh38
rs546660823 Chr.1: 6460255-6460255 on Build GRCh38
rs928158520 Chr.1: 6460590-6460590 on Build GRCh38
rs1047249123 Chr.1: 6460476-6460477 on Build GRCh38
rs183581437 Chr.1: 6460551-6460551 on Build GRCh38
rs751279882 Chr.1: 6460194-6460194 on Build GRCh38
rs191330418 Chr.1: 6460421-6460421 on Build GRCh38
rs1027284185 Chr.1: 6460240-6460240 on Build GRCh38
rs574227642 Chr.1: 6460633-6460633 on Build GRCh38
rs912449017 Chr.1: 6460419-6460419 on Build GRCh38
rs760069289 Chr.1: 6460160-6460160 on Build GRCh38
rs527946174 Chr.1: 6460373-6460375 on Build GRCh38
rs990981159 Chr.1: 6460602-6460602 on Build GRCh38
rs961663875 Chr.1: 6460582-6460582 on Build GRCh38
rs41278010 Chr.1: 6460608-6460608 on Build GRCh38
rs919631165 Chr.1: 6460614-6460614 on Build GRCh38
rs3138158 Chr.1: 6460252-6460252 on Build GRCh38
rs995941847 Chr.1: 6460558-6460558 on Build GRCh38
rs566670983 Chr.1: 6460525-6460525 on Build GRCh38
rs949565991 Chr.1: 6460497-6460498 on Build GRCh38
rs898264352 Chr.1: 6460276-6460277 on Build GRCh38
rs557596318 Chr.1: 6460338-6460338 on Build GRCh38
rs1025635753 Chr.1: 6460304-6460304 on Build GRCh38
rs545897938 Chr.1: 6460165-6460165 on Build GRCh38
rs1008758716 Chr.1: 6460163-6460163 on Build GRCh38
rs536497264 Chr.1: 6460237-6460237 on Build GRCh38
rs967610678 Chr.1: 6460402-6460402 on Build GRCh38
rs964510104 Chr.1: 6460188-6460188 on Build GRCh38
rs1014520047 Chr.1: 6460580-6460580 on Build GRCh38
rs982738005 Chr.1: 6460584-6460584 on Build GRCh38
rs959848001 Chr.1: 6460601-6460601 on Build GRCh38
rs982320794 Chr.1: 6460281-6460281 on Build GRCh38
rs1002179475 Chr.1: 6460315-6460315 on Build GRCh38
rs900413241 Chr.1: 6460648-6460648 on Build GRCh38
rs778303174 Chr.1: 6460168-6460168 on Build GRCh38
rs958125432 Chr.1: 6460335-6460336 on Build GRCh38
rs887235435 Chr.1: 6460292-6460292 on Build GRCh38
rs975087105 Chr.1: 6460437-6460438 on Build GRCh38
rs142557911 Chr.1: 6460332-6460332 on Build GRCh38
rs773087256 Chr.1: 6460468-6460468 on Build GRCh38
rs1004528711 Chr.1: 6460577-6460577 on Build GRCh38
rs969791327 Chr.1: 6460189-6460189 on Build GRCh38
rs919557870 Chr.1: 6460453-6460453 on Build GRCh38
rs753014653 Chr.1: 6460166-6460166 on Build GRCh38
rs915359387 Chr.1: 6460605-6460605 on Build GRCh38
rs762597615 Chr.1: 6460182-6460182 on Build GRCh38
rs887316321 Chr.1: 6460488-6460488 on Build GRCh38
rs1040654959 Chr.1: 6460617-6460617 on Build GRCh38
rs539509224 Chr.1: 6460492-6460492 on Build GRCh38
rs905215756 Chr.1: 6460308-6460308 on Build GRCh38
rs1047268093 Chr.1: 6460289-6460289 on Build GRCh38
rs958092805 Chr.1: 6460206-6460206 on Build GRCh38
rs568349878 Chr.1: 6460320-6460320 on Build GRCh38
rs759296967 Chr.1: 6460624-6460624 on Build GRCh38
rs371734244 Chr.1: 6460192-6460192 on Build GRCh38
rs1013877309 Chr.1: 6460299-6460299 on Build GRCh38
rs557833123 Chr.1: 6460501-6460501 on Build GRCh38
rs777716828 Chr.1: 6460571-6460571 on Build GRCh38
rs569050087 Chr.1: 6460467-6460467 on Build GRCh38
rs986761775 Chr.1: 6460403-6460403 on Build GRCh38
rs1027399637 Chr.1: 6460572-6460572 on Build GRCh38
rs547192907 Chr.1: 6460410-6460410 on Build GRCh38
rs951789732 Chr.1: 6460574-6460574 on Build GRCh38
rs988147389 Chr.1: 6460366-6460366 on Build GRCh38
rs923907278 Chr.1: 6460610-6460610 on Build GRCh38
rs748769000 Chr.1: 6460475-6460475 on Build GRCh38
rs544910011 Chr.1: 6460637-6460637 on Build GRCh38
rs961911773 Chr.1: 6460266-6460266 on Build GRCh38
rs1055176550 Chr.1: 6460371-6460371 on Build GRCh38
rs55729241 Chr.1: 6460634-6460634 on Build GRCh38
rs931052077 Chr.1: 6460463-6460463 on Build GRCh38
rs908832163 Chr.1: 6460262-6460262 on Build GRCh38
rs1002377389 Chr.1: 6460533-6460533 on Build GRCh38
rs1042428614 Chr.1: 6460609-6460609 on Build GRCh38
rs576188866 Chr.1: 6460482-6460482 on Build GRCh38
rs944646884 Chr.1: 6460616-6460616 on Build GRCh38
rs1042396874 Chr.1: 6460422-6460422 on Build GRCh38
rs1032276640 Chr.1: 6460318-6460319 on Build GRCh38
rs952349223 Chr.1: 6460657-6460657 on Build GRCh38

View Full Product Details