Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49639417-49639931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1 XM_017019139.1

SNP Detail

SNP ID Chromosome Location
rs374884640 Chr.12: 49639648-49639648 on Build GRCh38
rs961881748 Chr.12: 49639442-49639442 on Build GRCh38
rs200697984 Chr.12: 49639685-49639685 on Build GRCh38
rs181876187 Chr.12: 49639902-49639902 on Build GRCh38
rs913616892 Chr.12: 49639591-49639591 on Build GRCh38
rs186076159 Chr.12: 49639867-49639867 on Build GRCh38
rs780212890 Chr.12: 49639500-49639500 on Build GRCh38
rs952089783 Chr.12: 49639464-49639464 on Build GRCh38
rs779174315 Chr.12: 49639653-49639653 on Build GRCh38
rs1027517663 Chr.12: 49639807-49639807 on Build GRCh38
rs939005333 Chr.12: 49639528-49639528 on Build GRCh38
rs533116604 Chr.12: 49639619-49639619 on Build GRCh38
rs796629062 Chr.12: 49639764-49639764 on Build GRCh38
rs912729599 Chr.12: 49639556-49639556 on Build GRCh38
rs1012258797 Chr.12: 49639470-49639470 on Build GRCh38
rs372973871 Chr.12: 49639782-49639782 on Build GRCh38
rs140284083 Chr.12: 49639690-49639690 on Build GRCh38
rs891135240 Chr.12: 49639704-49639704 on Build GRCh38
rs11837637 Chr.12: 49639890-49639890 on Build GRCh38
rs977522594 Chr.12: 49639539-49639539 on Build GRCh38
rs764323373 Chr.12: 49639823-49639823 on Build GRCh38
rs921288989 Chr.12: 49639846-49639846 on Build GRCh38
rs1027485097 Chr.12: 49639462-49639462 on Build GRCh38
rs770727694 Chr.12: 49639853-49639853 on Build GRCh38
rs897282265 Chr.12: 49639783-49639783 on Build GRCh38
rs541169662 Chr.12: 49639560-49639560 on Build GRCh38
rs980698681 Chr.12: 49639496-49639496 on Build GRCh38
rs932920895 Chr.12: 49639684-49639685 on Build GRCh38
rs189681857 Chr.12: 49639802-49639802 on Build GRCh38
rs189616825 Chr.12: 49639871-49639871 on Build GRCh38
rs749367684 Chr.12: 49639501-49639501 on Build GRCh38
rs748305136 Chr.12: 49639787-49639787 on Build GRCh38
rs529218309 Chr.12: 49639731-49639731 on Build GRCh38
rs750978710 Chr.12: 49639483-49639483 on Build GRCh38
rs1015689207 Chr.12: 49639774-49639775 on Build GRCh38
rs971851105 Chr.12: 49639461-49639461 on Build GRCh38
rs181723547 Chr.12: 49639861-49639861 on Build GRCh38
rs566343142 Chr.12: 49639547-49639547 on Build GRCh38
rs548696733 Chr.12: 49639617-49639617 on Build GRCh38
rs548264817 Chr.12: 49639780-49639780 on Build GRCh38
rs945127434 Chr.12: 49639634-49639634 on Build GRCh38
rs756527613 Chr.12: 49639487-49639487 on Build GRCh38
rs975456871 Chr.12: 49639758-49639758 on Build GRCh38
rs951893386 Chr.12: 49639860-49639860 on Build GRCh38
rs76826244 Chr.12: 49639901-49639901 on Build GRCh38
rs995583243 Chr.12: 49639784-49639784 on Build GRCh38
rs755072227 Chr.12: 49639515-49639515 on Build GRCh38
rs1043909386 Chr.12: 49639641-49639641 on Build GRCh38
rs572169512 Chr.12: 49639445-49639445 on Build GRCh38
rs564183205 Chr.12: 49639587-49639587 on Build GRCh38

View Full Product Details