Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49643539-49644040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL3
Entrez Gene ID
91010
Gene Name
formin like 3
Gene Aliases
FHOD3, FRL2, WBP-3, WBP3
UniGene
Hs.179838
Gene Chromosome Location
Chr.12: 49636499-49707414 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019139.1

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_175736.4 NM_198900.2 XM_005269218.2 XM_011538968.2 XM_011538969.2 XM_011538970.2 XM_011538971.2 XM_011538972.2 XM_011538973.2 XM_011538974.2 NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282948 c.2462G>A p.R821K Chr.12: 49643943-49643943 on Build GRCh38
294725 c.2379G>A p.S793S Chr.12: 49643752-49643752 on Build GRCh38
3812191 c.2330G>A p.R777Q Chr.12: 49643703-49643703 on Build GRCh38
468474 c.2372A>G p.H791R Chr.12: 49643745-49643745 on Build GRCh38
4538052 c.2436G>A p.E812E Chr.12: 49643917-49643917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769996631 Chr.12: 49643836-49643836 on Build GRCh38
rs753178460 Chr.12: 49643644-49643644 on Build GRCh38
rs368213608 Chr.12: 49643695-49643695 on Build GRCh38
rs779725553 Chr.12: 49643779-49643779 on Build GRCh38
rs749060712 Chr.12: 49643732-49643734 on Build GRCh38
rs770900535 Chr.12: 49643772-49643778 on Build GRCh38
rs933555184 Chr.12: 49643588-49643588 on Build GRCh38
rs781138383 Chr.12: 49643953-49643953 on Build GRCh38
rs766956522 Chr.12: 49643872-49643872 on Build GRCh38
rs1021760978 Chr.12: 49643868-49643868 on Build GRCh38
rs746094212 Chr.12: 49643824-49643824 on Build GRCh38
rs767237679 Chr.12: 49643769-49643769 on Build GRCh38
rs964908429 Chr.12: 49643869-49643869 on Build GRCh38
rs891434743 Chr.12: 49643817-49643817 on Build GRCh38
rs954969064 Chr.12: 49643914-49643914 on Build GRCh38
rs545601493 Chr.12: 49643626-49643626 on Build GRCh38
rs375733550 Chr.12: 49643988-49643988 on Build GRCh38
rs759765213 Chr.12: 49643716-49643716 on Build GRCh38
rs890643785 Chr.12: 49643860-49643860 on Build GRCh38
rs552348674 Chr.12: 49643944-49643944 on Build GRCh38
rs546205526 Chr.12: 49643816-49643816 on Build GRCh38
rs760248619 Chr.12: 49643882-49643882 on Build GRCh38
rs766116754 Chr.12: 49643574-49643574 on Build GRCh38
rs775868820 Chr.12: 49643739-49643739 on Build GRCh38
rs554507750 Chr.12: 49643806-49643806 on Build GRCh38
rs1018070489 Chr.12: 49643598-49643598 on Build GRCh38
rs143123725 Chr.12: 49643891-49643891 on Build GRCh38
rs774647232 Chr.12: 49643976-49643976 on Build GRCh38
rs753620551 Chr.12: 49643781-49643781 on Build GRCh38
rs756733866 Chr.12: 49643766-49643766 on Build GRCh38
rs1038166498 Chr.12: 49643673-49643673 on Build GRCh38
rs780805874 Chr.12: 49643670-49643670 on Build GRCh38
rs372342314 Chr.12: 49643725-49643725 on Build GRCh38
rs35275181 Chr.12: 49643962-49643962 on Build GRCh38
rs773106486 Chr.12: 49643563-49643563 on Build GRCh38
rs779050820 Chr.12: 49643712-49643712 on Build GRCh38
rs769196078 Chr.12: 49643844-49643844 on Build GRCh38
rs377741337 Chr.12: 49643663-49643663 on Build GRCh38
rs763460376 Chr.12: 49643741-49643741 on Build GRCh38
rs778972841 Chr.12: 49643789-49643789 on Build GRCh38
rs756966896 Chr.12: 49643669-49643669 on Build GRCh38
rs761119539 Chr.12: 49643997-49643999 on Build GRCh38
rs199498901 Chr.12: 49643671-49643671 on Build GRCh38
rs769038475 Chr.12: 49643974-49643974 on Build GRCh38
rs1007662384 Chr.12: 49643584-49643584 on Build GRCh38
rs375334697 Chr.12: 49643683-49643683 on Build GRCh38
rs760210737 Chr.12: 49644010-49644010 on Build GRCh38
rs948132103 Chr.12: 49643979-49643979 on Build GRCh38
rs770659188 Chr.12: 49643978-49643978 on Build GRCh38
rs763611565 Chr.12: 49643653-49643653 on Build GRCh38
rs754790262 Chr.12: 49643911-49643911 on Build GRCh38
rs775448396 Chr.12: 49643993-49643995 on Build GRCh38
rs752705577 Chr.12: 49643933-49643933 on Build GRCh38
rs3751214 Chr.12: 49643698-49643698 on Build GRCh38
rs762264867 Chr.12: 49643871-49643871 on Build GRCh38
rs144855915 Chr.12: 49643802-49643802 on Build GRCh38
rs764584707 Chr.12: 49643745-49643745 on Build GRCh38
rs566216578 Chr.12: 49643863-49643863 on Build GRCh38
rs148333441 Chr.12: 49643886-49643886 on Build GRCh38
rs759747378 Chr.12: 49643892-49643892 on Build GRCh38
rs983685856 Chr.12: 49643927-49643927 on Build GRCh38
rs758856869 Chr.12: 49643645-49643645 on Build GRCh38
rs554273973 Chr.12: 49643972-49643972 on Build GRCh38
rs201769680 Chr.12: 49644018-49644018 on Build GRCh38
rs776687956 Chr.12: 49643705-49643705 on Build GRCh38
rs772664757 Chr.12: 49643992-49643992 on Build GRCh38
rs749203336 Chr.12: 49643677-49643677 on Build GRCh38
rs532107587 Chr.12: 49643884-49643884 on Build GRCh38
rs1030494775 Chr.12: 49643902-49643902 on Build GRCh38
rs573389120 Chr.12: 49643603-49643603 on Build GRCh38
rs755741125 Chr.12: 49643777-49643777 on Build GRCh38
rs139757720 Chr.12: 49643965-49643965 on Build GRCh38
rs781286595 Chr.12: 49643821-49643821 on Build GRCh38
rs963374748 Chr.12: 49643624-49643624 on Build GRCh38
rs369910502 Chr.12: 49643837-49643837 on Build GRCh38
rs1053421633 Chr.12: 49643753-49643753 on Build GRCh38
rs774806697 Chr.12: 49643852-49643852 on Build GRCh38
rs200990114 Chr.12: 49643770-49643770 on Build GRCh38
rs755024495 Chr.12: 49643786-49643786 on Build GRCh38
rs774623365 Chr.12: 49643828-49643828 on Build GRCh38
rs778813456 Chr.12: 49643912-49643912 on Build GRCh38
rs763287654 Chr.12: 49643838-49643838 on Build GRCh38
rs945655880 Chr.12: 49643815-49643815 on Build GRCh38
rs369007204 Chr.12: 49644016-49644016 on Build GRCh38
rs755831443 Chr.12: 49643672-49643672 on Build GRCh38
rs765141072 Chr.12: 49644019-49644019 on Build GRCh38
rs779858010 Chr.12: 49643676-49643676 on Build GRCh38
rs373535929 Chr.12: 49643973-49643973 on Build GRCh38
rs116569197 Chr.12: 49643746-49643746 on Build GRCh38
rs907853435 Chr.12: 49643649-49643649 on Build GRCh38
rs114536874 Chr.12: 49643702-49643702 on Build GRCh38
rs749888984 Chr.12: 49643875-49643875 on Build GRCh38
rs767569237 Chr.12: 49643918-49643919 on Build GRCh38
rs926327767 Chr.12: 49643703-49643703 on Build GRCh38

View Full Product Details