Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52545323-52545866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT71
Entrez Gene ID
112802
Gene Name
keratin 71
Gene Aliases
HYPT13, K6IRS1, KRT6IRS, KRT6IRS1
UniGene
Hs.660007
Gene Chromosome Location
Chr.12: 52543909-52553147 on Build GRCh38
Transcripts
NM_033448.2 XM_017018749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1606284 c.1357A>C p.I453L Chr.12: 52545568-52545568 on Build GRCh38
1492915 c.1346C>T p.S449F Chr.12: 52545579-52545579 on Build GRCh38
3462329 c.1326G>A p.R442R Chr.12: 52545599-52545599 on Build GRCh38
3462327 c.1347C>T p.S449S Chr.12: 52545578-52545578 on Build GRCh38
1676989 c.1333G>T p.G445* Chr.12: 52545592-52545592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs57480354 Chr.12: 52545348-52545348 on Build GRCh38
rs984964299 Chr.12: 52545510-52545510 on Build GRCh38
rs529512085 Chr.12: 52545501-52545501 on Build GRCh38
rs553079991 Chr.12: 52545407-52545407 on Build GRCh38
rs1010765612 Chr.12: 52545359-52545359 on Build GRCh38
rs542847114 Chr.12: 52545509-52545509 on Build GRCh38
rs766961086 Chr.12: 52545520-52545520 on Build GRCh38
rs752212527 Chr.12: 52545558-52545561 on Build GRCh38
rs755976077 Chr.12: 52545622-52545629 on Build GRCh38
rs767740587 Chr.12: 52545609-52545609 on Build GRCh38
rs765715067 Chr.12: 52545645-52545645 on Build GRCh38
rs369296601 Chr.12: 52545462-52545462 on Build GRCh38
rs550466975 Chr.12: 52545829-52545829 on Build GRCh38
rs766856299 Chr.12: 52545629-52545629 on Build GRCh38
rs1028739453 Chr.12: 52545811-52545811 on Build GRCh38
rs372311616 Chr.12: 52545576-52545576 on Build GRCh38
rs10580903 Chr.12: 52545624-52545626 on Build GRCh38
rs928631340 Chr.12: 52545558-52545558 on Build GRCh38
rs951832311 Chr.12: 52545825-52545825 on Build GRCh38
rs1011185505 Chr.12: 52545454-52545454 on Build GRCh38
rs778746010 Chr.12: 52545549-52545549 on Build GRCh38
rs368599971 Chr.12: 52545626-52545626 on Build GRCh38
rs575283900 Chr.12: 52545461-52545461 on Build GRCh38
rs181727591 Chr.12: 52545655-52545655 on Build GRCh38
rs908013407 Chr.12: 52545827-52545827 on Build GRCh38
rs66941803 Chr.12: 52545627-52545629 on Build GRCh38
rs773208189 Chr.12: 52545524-52545524 on Build GRCh38
rs906177931 Chr.12: 52545414-52545414 on Build GRCh38
rs766646237 Chr.12: 52545716-52545716 on Build GRCh38
rs559836264 Chr.12: 52545582-52545582 on Build GRCh38
rs746514885 Chr.12: 52545740-52545740 on Build GRCh38
rs773173275 Chr.12: 52545620-52545620 on Build GRCh38
rs1022103920 Chr.12: 52545484-52545484 on Build GRCh38
rs371977049 Chr.12: 52545483-52545483 on Build GRCh38
rs774606724 Chr.12: 52545604-52545604 on Build GRCh38
rs141532578 Chr.12: 52545577-52545577 on Build GRCh38
rs369793135 Chr.12: 52545551-52545551 on Build GRCh38
rs908821788 Chr.12: 52545581-52545581 on Build GRCh38
rs906289439 Chr.12: 52545745-52545745 on Build GRCh38
rs374472492 Chr.12: 52545504-52545504 on Build GRCh38
rs1022217231 Chr.12: 52545375-52545375 on Build GRCh38
rs755384353 Chr.12: 52545543-52545543 on Build GRCh38
rs147544464 Chr.12: 52545408-52545408 on Build GRCh38
rs189497260 Chr.12: 52545428-52545428 on Build GRCh38
rs745728380 Chr.12: 52545586-52545586 on Build GRCh38
rs1011976634 Chr.12: 52545368-52545368 on Build GRCh38
rs770484451 Chr.12: 52545762-52545762 on Build GRCh38
rs563931176 Chr.12: 52545499-52545499 on Build GRCh38
rs962204572 Chr.12: 52545832-52545832 on Build GRCh38
rs762045608 Chr.12: 52545608-52545608 on Build GRCh38
rs941602551 Chr.12: 52545584-52545584 on Build GRCh38
rs751776779 Chr.12: 52545801-52545801 on Build GRCh38
rs1043544958 Chr.12: 52545649-52545649 on Build GRCh38
rs777176635 Chr.12: 52545568-52545568 on Build GRCh38
rs956615202 Chr.12: 52545446-52545446 on Build GRCh38
rs186440932 Chr.12: 52545775-52545775 on Build GRCh38
rs200326622 Chr.12: 52545636-52545636 on Build GRCh38
rs769547378 Chr.12: 52545598-52545598 on Build GRCh38
rs984515732 Chr.12: 52545826-52545826 on Build GRCh38
rs528559752 Chr.12: 52545634-52545634 on Build GRCh38
rs762251266 Chr.12: 52545449-52545449 on Build GRCh38
rs199775091 Chr.12: 52545627-52545627 on Build GRCh38
rs758071157 Chr.12: 52545556-52545556 on Build GRCh38
rs981690728 Chr.12: 52545842-52545842 on Build GRCh38
rs201712190 Chr.12: 52545651-52545651 on Build GRCh38
rs74095112 Chr.12: 52545737-52545737 on Build GRCh38
rs544143346 Chr.12: 52545481-52545481 on Build GRCh38
rs766847415 Chr.12: 52545526-52545526 on Build GRCh38
rs372408940 Chr.12: 52545621-52545621 on Build GRCh38
rs60971309 Chr.12: 52545441-52545441 on Build GRCh38
rs940915109 Chr.12: 52545539-52545539 on Build GRCh38
rs999418282 Chr.12: 52545673-52545673 on Build GRCh38
rs754170487 Chr.12: 52545534-52545534 on Build GRCh38

View Full Product Details