Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54030492-54030995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54056030 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC5
Entrez Gene ID
3222
Gene Name
homeobox C5
Gene Aliases
CP11, HOX3, HOX3D
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54035361 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC6
Entrez Gene ID
3223
Gene Name
homeobox C6
Gene Aliases
CP25, HHO.C8, HOX3, HOX3C
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54030823 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

SNP Detail

SNP ID Chromosome Location
rs867218209 Chr.12: 54030743-54030743 on Build GRCh38
rs934835236 Chr.12: 54030821-54030821 on Build GRCh38
rs1039798970 Chr.12: 54030649-54030650 on Build GRCh38
rs1049736407 Chr.12: 54030871-54030871 on Build GRCh38
rs1043470721 Chr.12: 54030791-54030791 on Build GRCh38
rs914978209 Chr.12: 54030855-54030855 on Build GRCh38
rs975200758 Chr.12: 54030937-54030937 on Build GRCh38
rs1024028895 Chr.12: 54030749-54030749 on Build GRCh38
rs990497963 Chr.12: 54030751-54030751 on Build GRCh38
rs530933624 Chr.12: 54030710-54030710 on Build GRCh38
rs961217135 Chr.12: 54030721-54030721 on Build GRCh38
rs577380635 Chr.12: 54030882-54030882 on Build GRCh38
rs926101991 Chr.12: 54030808-54030808 on Build GRCh38
rs933865632 Chr.12: 54030662-54030662 on Build GRCh38
rs946457052 Chr.12: 54030787-54030787 on Build GRCh38
rs796821332 Chr.12: 54030635-54030635 on Build GRCh38
rs973022204 Chr.12: 54030727-54030727 on Build GRCh38
rs924079329 Chr.12: 54030545-54030545 on Build GRCh38
rs935503668 Chr.12: 54030551-54030551 on Build GRCh38
rs942417909 Chr.12: 54030870-54030870 on Build GRCh38
rs886939889 Chr.12: 54030680-54030681 on Build GRCh38
rs899271811 Chr.12: 54030740-54030740 on Build GRCh38
rs190134994 Chr.12: 54030845-54030845 on Build GRCh38
rs1016339601 Chr.12: 54030739-54030739 on Build GRCh38
rs989241660 Chr.12: 54030579-54030579 on Build GRCh38
rs922227246 Chr.12: 54030946-54030946 on Build GRCh38
rs998865861 Chr.12: 54030862-54030862 on Build GRCh38
rs889802561 Chr.12: 54030896-54030896 on Build GRCh38
rs994007880 Chr.12: 54030964-54030964 on Build GRCh38
rs548022112 Chr.12: 54030619-54030619 on Build GRCh38
rs961165736 Chr.12: 54030960-54030960 on Build GRCh38
rs1028779361 Chr.12: 54030614-54030614 on Build GRCh38
rs970930139 Chr.12: 54030521-54030521 on Build GRCh38
rs570765932 Chr.12: 54030828-54030828 on Build GRCh38
rs900853084 Chr.12: 54030650-54030650 on Build GRCh38
rs563121196 Chr.12: 54030670-54030670 on Build GRCh38
rs927910644 Chr.12: 54030731-54030731 on Build GRCh38
rs766300445 Chr.12: 54030786-54030786 on Build GRCh38
rs760238658 Chr.12: 54030664-54030664 on Build GRCh38
rs373542987 Chr.12: 54030544-54030544 on Build GRCh38
rs1047019653 Chr.12: 54030663-54030663 on Build GRCh38
rs990083794 Chr.12: 54030849-54030849 on Build GRCh38
rs552604552 Chr.12: 54030764-54030764 on Build GRCh38
rs982343597 Chr.12: 54030536-54030536 on Build GRCh38
rs4759319 Chr.12: 54030947-54030947 on Build GRCh38
rs901420363 Chr.12: 54030853-54030853 on Build GRCh38
rs1035304709 Chr.12: 54030966-54030966 on Build GRCh38
rs1005615580 Chr.12: 54030936-54030936 on Build GRCh38
rs1005424411 Chr.12: 54030681-54030684 on Build GRCh38

View Full Product Details