Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55938420-55938918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKA
Entrez Gene ID
1606
Gene Name
diacylglycerol kinase alpha
Gene Aliases
DAGK, DAGK1, DGK-alpha
UniGene
Hs.524488
Gene Chromosome Location
Chr.12: 55931076-55954023 on Build GRCh38
Transcripts
NM_001345.4 NM_201444.2 NM_201445.1 NM_201554.1 XM_005268688.2 XM_005268689.2 XM_005268690.2 XM_011537991.2 XM_011537993.2 XM_011537995.2 XM_017018900.1 XM_017018901.1 XM_017018902.1 XM_017018903.1 XM_017018904.1 XM_017018905.1 XM_017018906.1 XM_017018907.1 XM_017018908.1 XM_017018909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6462726 c.400-1G>A p.? Chr.12: 55938914-55938914 on Build GRCh38
6462727 c.400G>A p.E134K Chr.12: 55938915-55938915 on Build GRCh38
4922900 c.393C>T p.D131D Chr.12: 55938554-55938554 on Build GRCh38
4433171 c.400-3C>T p.? Chr.12: 55938912-55938912 on Build GRCh38
4633694 c.402A>G p.E134E Chr.12: 55938917-55938917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776698326 Chr.12: 55938705-55938705 on Build GRCh38
rs374645352 Chr.12: 55938579-55938579 on Build GRCh38
rs765461498 Chr.12: 55938616-55938616 on Build GRCh38
rs144444972 Chr.12: 55938889-55938890 on Build GRCh38
rs896286128 Chr.12: 55938838-55938838 on Build GRCh38
rs752852641 Chr.12: 55938621-55938621 on Build GRCh38
rs765584533 Chr.12: 55938506-55938506 on Build GRCh38
rs368802787 Chr.12: 55938465-55938465 on Build GRCh38
rs755092533 Chr.12: 55938476-55938476 on Build GRCh38
rs764137836 Chr.12: 55938628-55938628 on Build GRCh38
rs771388822 Chr.12: 55938704-55938704 on Build GRCh38
rs908166534 Chr.12: 55938603-55938603 on Build GRCh38
rs1030806319 Chr.12: 55938450-55938450 on Build GRCh38
rs534157100 Chr.12: 55938890-55938890 on Build GRCh38
rs937329359 Chr.12: 55938670-55938670 on Build GRCh38
rs375330439 Chr.12: 55938874-55938874 on Build GRCh38
rs765574999 Chr.12: 55938722-55938722 on Build GRCh38
rs900175320 Chr.12: 55938858-55938858 on Build GRCh38
rs763496527 Chr.12: 55938685-55938685 on Build GRCh38
rs773559454 Chr.12: 55938681-55938681 on Build GRCh38
rs1043108809 Chr.12: 55938735-55938735 on Build GRCh38
rs748818669 Chr.12: 55938661-55938661 on Build GRCh38
rs772598958 Chr.12: 55938578-55938578 on Build GRCh38
rs766447985 Chr.12: 55938593-55938593 on Build GRCh38
rs755714091 Chr.12: 55938894-55938894 on Build GRCh38
rs567139537 Chr.12: 55938887-55938887 on Build GRCh38
rs751570531 Chr.12: 55938881-55938881 on Build GRCh38
rs187690093 Chr.12: 55938615-55938615 on Build GRCh38
rs202099141 Chr.12: 55938597-55938597 on Build GRCh38
rs753045610 Chr.12: 55938636-55938636 on Build GRCh38
rs770429616 Chr.12: 55938526-55938526 on Build GRCh38
rs751330011 Chr.12: 55938635-55938635 on Build GRCh38
rs1053681401 Chr.12: 55938854-55938854 on Build GRCh38
rs111286159 Chr.12: 55938739-55938739 on Build GRCh38
rs773518297 Chr.12: 55938582-55938582 on Build GRCh38
rs947352093 Chr.12: 55938689-55938689 on Build GRCh38
rs368010273 Chr.12: 55938778-55938778 on Build GRCh38
rs758357490 Chr.12: 55938625-55938625 on Build GRCh38
rs753803878 Chr.12: 55938472-55938472 on Build GRCh38
rs755620859 Chr.12: 55938883-55938894 on Build GRCh38
rs144412553 Chr.12: 55938451-55938451 on Build GRCh38
rs780692770 Chr.12: 55938638-55938638 on Build GRCh38
rs779500762 Chr.12: 55938656-55938656 on Build GRCh38
rs371312047 Chr.12: 55938532-55938532 on Build GRCh38
rs748730629 Chr.12: 55938895-55938895 on Build GRCh38
rs745363675 Chr.12: 55938533-55938533 on Build GRCh38
rs745582477 Chr.12: 55938640-55938640 on Build GRCh38
rs752744223 Chr.12: 55938486-55938486 on Build GRCh38
rs762348707 Chr.12: 55938574-55938574 on Build GRCh38
rs760415366 Chr.12: 55938485-55938485 on Build GRCh38
rs192667576 Chr.12: 55938742-55938742 on Build GRCh38
rs558183718 Chr.12: 55938861-55938861 on Build GRCh38
rs764571802 Chr.12: 55938794-55938794 on Build GRCh38
rs766647464 Chr.12: 55938470-55938470 on Build GRCh38
rs772512801 Chr.12: 55938662-55938662 on Build GRCh38
rs1000767449 Chr.12: 55938799-55938799 on Build GRCh38
rs758232701 Chr.12: 55938491-55938491 on Build GRCh38
rs750613194 Chr.12: 55938460-55938460 on Build GRCh38
rs774817032 Chr.12: 55938566-55938566 on Build GRCh38
rs771867258 Chr.12: 55938795-55938795 on Build GRCh38
rs755547693 Chr.12: 55938652-55938652 on Build GRCh38
rs555925579 Chr.12: 55938891-55938891 on Build GRCh38
rs773714429 Chr.12: 55938683-55938683 on Build GRCh38
rs183271731 Chr.12: 55938484-55938484 on Build GRCh38
rs777667745 Chr.12: 55938494-55938494 on Build GRCh38
rs759760849 Chr.12: 55938719-55938719 on Build GRCh38
rs761116222 Chr.12: 55938592-55938592 on Build GRCh38
rs568520742 Chr.12: 55938741-55938741 on Build GRCh38

View Full Product Details