Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56204035-56204578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF41
Entrez Gene ID
10193
Gene Name
ring finger protein 41
Gene Aliases
FLRF, NRDP1, SBBI03
UniGene
Hs.524502
Gene Chromosome Location
Chr.12: 56202504-56221982 on Build GRCh38
Transcripts
NM_001242826.1 NM_005785.3 NM_194358.2 NM_194359.2 NR_040053.1 XM_005268561.4 XM_011537734.2 XM_011537735.2

SNP Detail

SNP ID Chromosome Location
rs111732679 Chr.12: 56204507-56204511 on Build GRCh38
rs904397093 Chr.12: 56204442-56204442 on Build GRCh38
rs575519528 Chr.12: 56204157-56204157 on Build GRCh38
rs556423473 Chr.12: 56204091-56204091 on Build GRCh38
rs564778589 Chr.12: 56204128-56204128 on Build GRCh38
rs1028173103 Chr.12: 56204073-56204073 on Build GRCh38
rs1051043862 Chr.12: 56204319-56204319 on Build GRCh38
rs773654 Chr.12: 56204260-56204260 on Build GRCh38
rs1014565375 Chr.12: 56204556-56204556 on Build GRCh38
rs972666958 Chr.12: 56204098-56204098 on Build GRCh38
rs539460568 Chr.12: 56204189-56204189 on Build GRCh38
rs952757116 Chr.12: 56204486-56204486 on Build GRCh38
rs765959271 Chr.12: 56204523-56204525 on Build GRCh38
rs746686018 Chr.12: 56204308-56204308 on Build GRCh38
rs770548023 Chr.12: 56204406-56204406 on Build GRCh38
rs891728545 Chr.12: 56204522-56204525 on Build GRCh38
rs574186001 Chr.12: 56204365-56204365 on Build GRCh38
rs983942525 Chr.12: 56204493-56204493 on Build GRCh38
rs1052852005 Chr.12: 56204515-56204515 on Build GRCh38
rs889192121 Chr.12: 56204387-56204387 on Build GRCh38
rs969147312 Chr.12: 56204547-56204547 on Build GRCh38
rs965244707 Chr.12: 56204089-56204089 on Build GRCh38
rs1014174286 Chr.12: 56204249-56204260 on Build GRCh38
rs753885391 Chr.12: 56204072-56204072 on Build GRCh38
rs920801897 Chr.12: 56204222-56204222 on Build GRCh38
rs955920165 Chr.12: 56204168-56204168 on Build GRCh38
rs894051061 Chr.12: 56204292-56204292 on Build GRCh38
rs932107235 Chr.12: 56204433-56204433 on Build GRCh38
rs778867163 Chr.12: 56204172-56204172 on Build GRCh38
rs762416233 Chr.12: 56204363-56204363 on Build GRCh38
rs969852851 Chr.12: 56204209-56204209 on Build GRCh38
rs752580227 Chr.12: 56204207-56204207 on Build GRCh38
rs562132454 Chr.12: 56204313-56204313 on Build GRCh38
rs113824028 Chr.12: 56204227-56204227 on Build GRCh38
rs544533254 Chr.12: 56204538-56204538 on Build GRCh38
rs759796789 Chr.12: 56204246-56204246 on Build GRCh38
rs952545035 Chr.12: 56204151-56204151 on Build GRCh38
rs200871261 Chr.12: 56204221-56204221 on Build GRCh38
rs1018896697 Chr.12: 56204415-56204415 on Build GRCh38
rs897866398 Chr.12: 56204436-56204436 on Build GRCh38
rs993468897 Chr.12: 56204445-56204445 on Build GRCh38
rs780893150 Chr.12: 56204452-56204452 on Build GRCh38
rs572979055 Chr.12: 56204234-56204234 on Build GRCh38
rs78669848 Chr.12: 56204217-56204217 on Build GRCh38

View Full Product Details