Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56712830-56713331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACA
Entrez Gene ID
4666
Gene Name
nascent polypeptide-associated complex alpha subunit
Gene Aliases
HSD48, NAC-alpha, NACA1, skNAC
UniGene
Hs.505735
Gene Chromosome Location
Chr.12: 56712427-56726275 on Build GRCh38
Transcripts
NM_001113201.2 NM_001113202.1 NM_001113203.2 NM_001320193.1 NM_001320194.1 NM_005594.5 XM_006719412.1 XM_006719413.2 XM_006719414.1 XM_006719418.1 XM_006719420.3 XM_011538389.1 XM_017019337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336547 c.2526C>T p.S842S Chr.12: 56713176-56713176 on Build GRCh38
5739691 c.582G>T p.K194N Chr.12: 56712837-56712837 on Build GRCh38
6336544 c.2527C>G p.Q843E Chr.12: 56713175-56713175 on Build GRCh38
1180644 c.2674G>C p.E892Q Chr.12: 56712875-56712875 on Build GRCh38
4994030 c.454A>G p.N152D Chr.12: 56713118-56713118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372164873 Chr.12: 56713043-56713043 on Build GRCh38
rs186711868 Chr.12: 56712914-56712914 on Build GRCh38
rs1034010346 Chr.12: 56712990-56712990 on Build GRCh38
rs776757899 Chr.12: 56713206-56713206 on Build GRCh38
rs759903795 Chr.12: 56712946-56712948 on Build GRCh38
rs957512830 Chr.12: 56713283-56713283 on Build GRCh38
rs140448348 Chr.12: 56713086-56713086 on Build GRCh38
rs190289763 Chr.12: 56713209-56713209 on Build GRCh38
rs898609 Chr.12: 56713026-56713026 on Build GRCh38
rs765004308 Chr.12: 56712872-56712872 on Build GRCh38
rs61741738 Chr.12: 56713122-56713122 on Build GRCh38
rs757894002 Chr.12: 56712879-56712879 on Build GRCh38
rs774751792 Chr.12: 56712918-56712919 on Build GRCh38
rs754316696 Chr.12: 56713081-56713081 on Build GRCh38
rs761348765 Chr.12: 56713227-56713227 on Build GRCh38
rs375567238 Chr.12: 56713297-56713297 on Build GRCh38
rs556881553 Chr.12: 56713020-56713020 on Build GRCh38
rs879091640 Chr.12: 56713059-56713059 on Build GRCh38
rs758653483 Chr.12: 56713188-56713188 on Build GRCh38
rs777322987 Chr.12: 56712893-56712893 on Build GRCh38
rs769979002 Chr.12: 56713298-56713298 on Build GRCh38
rs779325302 Chr.12: 56713096-56713096 on Build GRCh38
rs369553170 Chr.12: 56713218-56713218 on Build GRCh38
rs149732816 Chr.12: 56712905-56712905 on Build GRCh38
rs906320955 Chr.12: 56713266-56713266 on Build GRCh38
rs4788 Chr.12: 56712876-56712876 on Build GRCh38
rs878965045 Chr.12: 56713193-56713193 on Build GRCh38
rs955037728 Chr.12: 56713056-56713056 on Build GRCh38
rs556518624 Chr.12: 56712961-56712961 on Build GRCh38
rs936969767 Chr.12: 56713097-56713097 on Build GRCh38
rs200124165 Chr.12: 56713029-56713029 on Build GRCh38
rs986355282 Chr.12: 56713057-56713057 on Build GRCh38
rs771094961 Chr.12: 56713199-56713199 on Build GRCh38
rs749671400 Chr.12: 56712915-56712915 on Build GRCh38
rs745786935 Chr.12: 56713208-56713208 on Build GRCh38
rs773831918 Chr.12: 56713225-56713225 on Build GRCh38
rs527361962 Chr.12: 56713049-56713049 on Build GRCh38
rs146755279 Chr.12: 56713079-56713079 on Build GRCh38
rs534417377 Chr.12: 56712906-56712906 on Build GRCh38
rs1002075686 Chr.12: 56712959-56712959 on Build GRCh38
rs779150027 Chr.12: 56712948-56712948 on Build GRCh38
rs375914904 Chr.12: 56713028-56713028 on Build GRCh38
rs771667951 Chr.12: 56713250-56713251 on Build GRCh38
rs1046120750 Chr.12: 56713264-56713265 on Build GRCh38
rs201182580 Chr.12: 56712957-56712957 on Build GRCh38
rs200948506 Chr.12: 56713176-56713176 on Build GRCh38
rs368635646 Chr.12: 56712951-56712951 on Build GRCh38
rs202101850 Chr.12: 56713214-56713214 on Build GRCh38
rs531385852 Chr.12: 56713292-56713292 on Build GRCh38
rs771007076 Chr.12: 56713025-56713025 on Build GRCh38
rs756746371 Chr.12: 56713241-56713244 on Build GRCh38
rs1009287289 Chr.12: 56712917-56712917 on Build GRCh38
rs544951704 Chr.12: 56712967-56712967 on Build GRCh38
rs542829900 Chr.12: 56713071-56713071 on Build GRCh38
rs763666769 Chr.12: 56712957-56712959 on Build GRCh38
rs766995886 Chr.12: 56713235-56713235 on Build GRCh38
rs753523188 Chr.12: 56713025-56713025 on Build GRCh38
rs894486492 Chr.12: 56713082-56713082 on Build GRCh38
rs868609068 Chr.12: 56713254-56713254 on Build GRCh38
rs748387766 Chr.12: 56713146-56713146 on Build GRCh38
rs939766615 Chr.12: 56712996-56712996 on Build GRCh38
rs202147902 Chr.12: 56713010-56713010 on Build GRCh38
rs756703844 Chr.12: 56713068-56713068 on Build GRCh38
rs775637419 Chr.12: 56713307-56713307 on Build GRCh38
rs755291310 Chr.12: 56712933-56712933 on Build GRCh38
rs759790877 Chr.12: 56713276-56713276 on Build GRCh38
rs762595843 Chr.12: 56713216-56713216 on Build GRCh38

View Full Product Details