Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69568750-69569298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS2
Entrez Gene ID
10818
Gene Name
fibroblast growth factor receptor substrate 2
Gene Aliases
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1
UniGene
Hs.593446
Gene Chromosome Location
Chr.12: 69470349-69579793 on Build GRCh38
Transcripts
NM_001042555.2 NM_001278351.1 NM_001278353.1 NM_001278354.1 NM_001278355.1 NM_001278356.1 NM_001278357.1 NM_006654.4 XM_017018717.1 XM_017018718.1 XM_017018719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695296 c.43G>T p.D15Y Chr.12: 69569073-69569073 on Build GRCh38
6137808 c.27T>A p.D9E Chr.12: 69569057-69569057 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009261786 Chr.12: 69568996-69568996 on Build GRCh38
rs945822208 Chr.12: 69568828-69568828 on Build GRCh38
rs901135238 Chr.12: 69569004-69569004 on Build GRCh38
rs753441533 Chr.12: 69569207-69569210 on Build GRCh38
rs911727044 Chr.12: 69568823-69568823 on Build GRCh38
rs781370399 Chr.12: 69569017-69569017 on Build GRCh38
rs550635349 Chr.12: 69568794-69568794 on Build GRCh38
rs764065492 Chr.12: 69569079-69569079 on Build GRCh38
rs935644542 Chr.12: 69568874-69568874 on Build GRCh38
rs533839846 Chr.12: 69569201-69569201 on Build GRCh38
rs371752169 Chr.12: 69569157-69569157 on Build GRCh38
rs962720432 Chr.12: 69569159-69569159 on Build GRCh38
rs368147839 Chr.12: 69569140-69569140 on Build GRCh38
rs536498953 Chr.12: 69569006-69569006 on Build GRCh38
rs1028912957 Chr.12: 69569218-69569218 on Build GRCh38
rs369674581 Chr.12: 69569078-69569078 on Build GRCh38
rs1046359325 Chr.12: 69569008-69569008 on Build GRCh38
rs530751446 Chr.12: 69569220-69569220 on Build GRCh38
rs750603755 Chr.12: 69569119-69569119 on Build GRCh38
rs770627638 Chr.12: 69569044-69569044 on Build GRCh38
rs752138417 Chr.12: 69569132-69569132 on Build GRCh38
rs902423444 Chr.12: 69568918-69568919 on Build GRCh38
rs373106662 Chr.12: 69569090-69569090 on Build GRCh38
rs780230690 Chr.12: 69569125-69569125 on Build GRCh38
rs894682630 Chr.12: 69568961-69568961 on Build GRCh38
rs977942234 Chr.12: 69569234-69569234 on Build GRCh38
rs759053200 Chr.12: 69569071-69569071 on Build GRCh38
rs758545723 Chr.12: 69568998-69568998 on Build GRCh38
rs748513447 Chr.12: 69569031-69569031 on Build GRCh38
rs754636600 Chr.12: 69569138-69569138 on Build GRCh38
rs987672064 Chr.12: 69568782-69568782 on Build GRCh38
rs765619140 Chr.12: 69569111-69569111 on Build GRCh38
rs553808386 Chr.12: 69569143-69569145 on Build GRCh38
rs750585279 Chr.12: 69568994-69568994 on Build GRCh38
rs944555482 Chr.12: 69569270-69569271 on Build GRCh38
rs987399237 Chr.12: 69569258-69569258 on Build GRCh38
rs1055984876 Chr.12: 69568889-69568889 on Build GRCh38
rs879182723 Chr.12: 69568902-69568902 on Build GRCh38
rs554759899 Chr.12: 69569060-69569060 on Build GRCh38
rs758564188 Chr.12: 69569120-69569120 on Build GRCh38
rs953459899 Chr.12: 69569250-69569250 on Build GRCh38
rs755477608 Chr.12: 69569136-69569136 on Build GRCh38
rs1041435344 Chr.12: 69568849-69568849 on Build GRCh38
rs1004325712 Chr.12: 69569115-69569115 on Build GRCh38
rs539345874 Chr.12: 69568982-69568982 on Build GRCh38
rs374495404 Chr.12: 69569165-69569165 on Build GRCh38
rs996867754 Chr.12: 69569011-69569011 on Build GRCh38
rs74572384 Chr.12: 69569110-69569110 on Build GRCh38
rs946540687 Chr.12: 69568804-69568804 on Build GRCh38
rs781715320 Chr.12: 69569145-69569145 on Build GRCh38
rs780708337 Chr.12: 69569005-69569005 on Build GRCh38
rs377647712 Chr.12: 69569072-69569072 on Build GRCh38
rs567483540 Chr.12: 69568855-69568855 on Build GRCh38
rs923733599 Chr.12: 69569257-69569257 on Build GRCh38
rs990476878 Chr.12: 69569194-69569194 on Build GRCh38
rs1005717600 Chr.12: 69569082-69569082 on Build GRCh38
rs371094152 Chr.12: 69569142-69569142 on Build GRCh38
rs772875085 Chr.12: 69569179-69569189 on Build GRCh38
rs561617652 Chr.12: 69569103-69569103 on Build GRCh38
rs904220394 Chr.12: 69568852-69568852 on Build GRCh38
rs996816753 Chr.12: 69569207-69569207 on Build GRCh38
rs1014425603 Chr.12: 69568989-69568989 on Build GRCh38

View Full Product Details