Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69585299-69585824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
UniGene
Hs.189772
Gene Chromosome Location
Chr.12: 69585428-69601577 on Build GRCh38
Transcripts
NM_001198842.1 NM_006431.2

SNP Detail

SNP ID Chromosome Location
rs980229496 Chr.12: 69585712-69585712 on Build GRCh38
rs564805236 Chr.12: 69585708-69585708 on Build GRCh38
rs535665325 Chr.12: 69585365-69585365 on Build GRCh38
rs750288666 Chr.12: 69585531-69585531 on Build GRCh38
rs958092466 Chr.12: 69585610-69585610 on Build GRCh38
rs1011441615 Chr.12: 69585691-69585691 on Build GRCh38
rs775223273 Chr.12: 69585496-69585496 on Build GRCh38
rs76947730 Chr.12: 69585439-69585439 on Build GRCh38
rs768563666 Chr.12: 69585482-69585482 on Build GRCh38
rs531195601 Chr.12: 69585687-69585687 on Build GRCh38
rs1021034368 Chr.12: 69585625-69585625 on Build GRCh38
rs564442984 Chr.12: 69585425-69585425 on Build GRCh38
rs773761894 Chr.12: 69585491-69585491 on Build GRCh38
rs575014725 Chr.12: 69585359-69585359 on Build GRCh38
rs989977356 Chr.12: 69585527-69585527 on Build GRCh38
rs1017560665 Chr.12: 69585384-69585384 on Build GRCh38
rs772528559 Chr.12: 69585477-69585477 on Build GRCh38
rs140997100 Chr.12: 69585745-69585745 on Build GRCh38
rs1026545543 Chr.12: 69585427-69585427 on Build GRCh38
rs1031631632 Chr.12: 69585680-69585680 on Build GRCh38
rs949783535 Chr.12: 69585787-69585787 on Build GRCh38
rs770481913 Chr.12: 69585560-69585560 on Build GRCh38
rs931555069 Chr.12: 69585743-69585743 on Build GRCh38
rs1007221277 Chr.12: 69585377-69585377 on Build GRCh38
rs1031515524 Chr.12: 69585501-69585501 on Build GRCh38
rs748237416 Chr.12: 69585558-69585558 on Build GRCh38
rs572403472 Chr.12: 69585400-69585400 on Build GRCh38
rs760253565 Chr.12: 69585504-69585504 on Build GRCh38
rs866544432 Chr.12: 69585448-69585448 on Build GRCh38
rs1044442682 Chr.12: 69585455-69585455 on Build GRCh38
rs201916315 Chr.12: 69585552-69585552 on Build GRCh38
rs901146926 Chr.12: 69585651-69585651 on Build GRCh38
rs781734175 Chr.12: 69585488-69585488 on Build GRCh38
rs889865605 Chr.12: 69585480-69585480 on Build GRCh38
rs934418096 Chr.12: 69585432-69585432 on Build GRCh38
rs74749076 Chr.12: 69585721-69585721 on Build GRCh38
rs1024150563 Chr.12: 69585578-69585578 on Build GRCh38
rs995524957 Chr.12: 69585323-69585323 on Build GRCh38
rs562850253 Chr.12: 69585438-69585438 on Build GRCh38
rs758219946 Chr.12: 69585700-69585700 on Build GRCh38
rs558264488 Chr.12: 69585747-69585747 on Build GRCh38
rs997435319 Chr.12: 69585676-69585676 on Build GRCh38
rs375855066 Chr.12: 69585481-69585481 on Build GRCh38
rs554290842 Chr.12: 69585394-69585394 on Build GRCh38
rs1005592661 Chr.12: 69585331-69585331 on Build GRCh38
rs886268013 Chr.12: 69585565-69585565 on Build GRCh38
rs560204523 Chr.12: 69585534-69585534 on Build GRCh38
rs1049031617 Chr.12: 69585623-69585623 on Build GRCh38
rs1007369629 Chr.12: 69585641-69585641 on Build GRCh38
rs1037212600 Chr.12: 69585582-69585582 on Build GRCh38
rs1041071226 Chr.12: 69585648-69585648 on Build GRCh38
rs774632172 Chr.12: 69585468-69585468 on Build GRCh38
rs938949062 Chr.12: 69585562-69585562 on Build GRCh38
rs546233367 Chr.12: 69585423-69585423 on Build GRCh38
rs368802453 Chr.12: 69585701-69585701 on Build GRCh38
rs199514078 Chr.12: 69585540-69585540 on Build GRCh38
rs747301038 Chr.12: 69585576-69585578 on Build GRCh38
rs1024266328 Chr.12: 69585454-69585454 on Build GRCh38
rs759951747 Chr.12: 69585476-69585476 on Build GRCh38
rs1034068530 Chr.12: 69585596-69585596 on Build GRCh38
rs914044586 Chr.12: 69585483-69585483 on Build GRCh38
rs751685855 Chr.12: 69585538-69585538 on Build GRCh38
rs373387339 Chr.12: 69585536-69585536 on Build GRCh38
rs966843971 Chr.12: 69585675-69585675 on Build GRCh38
rs780270578 Chr.12: 69585537-69585537 on Build GRCh38
rs78079911 Chr.12: 69585583-69585583 on Build GRCh38
rs905707679 Chr.12: 69585795-69585795 on Build GRCh38
rs763404110 Chr.12: 69585492-69585492 on Build GRCh38
rs994600580 Chr.12: 69585417-69585417 on Build GRCh38
rs111702592 Chr.12: 69585740-69585740 on Build GRCh38
rs765141979 Chr.12: 69585517-69585517 on Build GRCh38
rs889909101 Chr.12: 69585640-69585640 on Build GRCh38
rs372102946 Chr.12: 69585479-69585479 on Build GRCh38
rs374499372 Chr.12: 69585778-69585778 on Build GRCh38
rs556425259 Chr.12: 69585789-69585789 on Build GRCh38
rs975037823 Chr.12: 69585731-69585731 on Build GRCh38
rs544552585 Chr.12: 69585461-69585461 on Build GRCh38
rs763985725 Chr.12: 69585490-69585490 on Build GRCh38
rs975944696 Chr.12: 69585397-69585397 on Build GRCh38
rs959661046 Chr.12: 69585735-69585735 on Build GRCh38
rs761999593 Chr.12: 69585567-69585567 on Build GRCh38
rs547602714 Chr.12: 69585783-69585783 on Build GRCh38
rs375160132 Chr.12: 69585471-69585471 on Build GRCh38
rs2601007 Chr.12: 69585335-69585335 on Build GRCh38
rs924423168 Chr.12: 69585399-69585399 on Build GRCh38
rs1014129755 Chr.12: 69585570-69585570 on Build GRCh38
rs17106752 Chr.12: 69585347-69585347 on Build GRCh38
rs1045565306 Chr.12: 69585792-69585792 on Build GRCh38
rs966014205 Chr.12: 69585489-69585489 on Build GRCh38
rs763468533 Chr.12: 69585506-69585506 on Build GRCh38
rs541611936 Chr.12: 69585495-69585495 on Build GRCh38
rs368910864 Chr.12: 69585515-69585515 on Build GRCh38
rs955930696 Chr.12: 69585685-69585685 on Build GRCh38
rs2601006 Chr.12: 69585737-69585737 on Build GRCh38
rs948451229 Chr.12: 69585451-69585451 on Build GRCh38
rs111365352 Chr.12: 69585526-69585526 on Build GRCh38
rs1034349513 Chr.12: 69585334-69585334 on Build GRCh38
rs576546053 Chr.12: 69585764-69585764 on Build GRCh38
rs1038776175 Chr.12: 69585390-69585390 on Build GRCh38
rs931520962 Chr.12: 69585609-69585609 on Build GRCh38
rs764087041 Chr.12: 69585546-69585546 on Build GRCh38
rs369960707 Chr.12: 69585473-69585473 on Build GRCh38
rs562630091 Chr.12: 69585472-69585472 on Build GRCh38
rs755176707 Chr.12: 69585793-69585793 on Build GRCh38
rs765476053 Chr.12: 69585573-69585573 on Build GRCh38
rs950546368 Chr.12: 69585433-69585433 on Build GRCh38
rs377616113 Chr.12: 69585612-69585612 on Build GRCh38
rs369443707 Chr.12: 69585759-69585759 on Build GRCh38
rs897301575 Chr.12: 69585474-69585474 on Build GRCh38
rs901253424 Chr.12: 69585791-69585791 on Build GRCh38
rs550104627 Chr.12: 69585698-69585698 on Build GRCh38
rs369196394 Chr.12: 69585574-69585574 on Build GRCh38
rs911358212 Chr.12: 69585761-69585761 on Build GRCh38

View Full Product Details