Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85036229-85036720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
-
UniGene
Hs.402200
Gene Chromosome Location
Chr.12: 85036321-85273004 on Build GRCh38
Transcripts
NM_001079910.1 XM_011538817.2 XM_011538818.2 XM_011538819.2 XM_011538823.2 XM_011538824.2 XM_011538825.2 XM_017020019.1 NM_001100917.1

Gene Symbol
TSPAN19
Entrez Gene ID
144448
Gene Name
tetraspanin 19
Gene Aliases
-
UniGene
Hs.156962
Gene Chromosome Location
Chr.12: 85014311-85036277 on Build GRCh38
Transcripts
NM_001079910.1 XM_011538817.2 XM_011538818.2 XM_011538819.2 XM_011538823.2 XM_011538824.2 XM_011538825.2 XM_017020019.1 NM_001100917.1

SNP Detail

SNP ID Chromosome Location
rs75487867 Chr.12: 85036405-85036405 on Build GRCh38
rs979797849 Chr.12: 85036414-85036415 on Build GRCh38
rs778842338 Chr.12: 85036645-85036645 on Build GRCh38
rs574703469 Chr.12: 85036529-85036529 on Build GRCh38
rs756386559 Chr.12: 85036604-85036605 on Build GRCh38
rs749934021 Chr.12: 85036306-85036306 on Build GRCh38
rs926915188 Chr.12: 85036416-85036416 on Build GRCh38
rs575169961 Chr.12: 85036524-85036524 on Build GRCh38
rs1031687369 Chr.12: 85036281-85036281 on Build GRCh38
rs948915026 Chr.12: 85036401-85036401 on Build GRCh38
rs141615305 Chr.12: 85036346-85036346 on Build GRCh38
rs922504793 Chr.12: 85036357-85036357 on Build GRCh38
rs35453500 Chr.12: 85036409-85036409 on Build GRCh38
rs111331871 Chr.12: 85036587-85036587 on Build GRCh38
rs949796679 Chr.12: 85036408-85036408 on Build GRCh38
rs773085161 Chr.12: 85036615-85036615 on Build GRCh38
rs933917134 Chr.12: 85036358-85036358 on Build GRCh38
rs1011617301 Chr.12: 85036478-85036478 on Build GRCh38
rs1032826190 Chr.12: 85036387-85036387 on Build GRCh38
rs147897065 Chr.12: 85036305-85036305 on Build GRCh38
rs994270686 Chr.12: 85036288-85036288 on Build GRCh38
rs1038072781 Chr.12: 85036376-85036376 on Build GRCh38
rs145567614 Chr.12: 85036379-85036379 on Build GRCh38
rs866376794 Chr.12: 85036674-85036674 on Build GRCh38
rs868265148 Chr.12: 85036342-85036342 on Build GRCh38
rs955855510 Chr.12: 85036399-85036399 on Build GRCh38
rs558146259 Chr.12: 85036446-85036446 on Build GRCh38
rs541325395 Chr.12: 85036435-85036435 on Build GRCh38
rs948485105 Chr.12: 85036654-85036654 on Build GRCh38
rs563262107 Chr.12: 85036604-85036604 on Build GRCh38
rs982384456 Chr.12: 85036367-85036367 on Build GRCh38
rs891148395 Chr.12: 85036308-85036308 on Build GRCh38
rs558510967 Chr.12: 85036413-85036413 on Build GRCh38
rs554779240 Chr.12: 85036359-85036359 on Build GRCh38
rs1007351648 Chr.12: 85036562-85036562 on Build GRCh38
rs763033648 Chr.12: 85036623-85036623 on Build GRCh38
rs893127620 Chr.12: 85036469-85036469 on Build GRCh38
rs1051214397 Chr.12: 85036482-85036483 on Build GRCh38
rs568263631 Chr.12: 85036317-85036317 on Build GRCh38
rs141124483 Chr.12: 85036666-85036666 on Build GRCh38
rs112776838 Chr.12: 85036680-85036680 on Build GRCh38
rs372046075 Chr.12: 85036339-85036339 on Build GRCh38
rs557911832 Chr.12: 85036522-85036522 on Build GRCh38
rs144754342 Chr.12: 85036298-85036298 on Build GRCh38
rs530606617 Chr.12: 85036639-85036639 on Build GRCh38
rs955268180 Chr.12: 85036585-85036585 on Build GRCh38
rs1000093558 Chr.12: 85036595-85036595 on Build GRCh38
rs980229332 Chr.12: 85036682-85036682 on Build GRCh38
rs370602703 Chr.12: 85036667-85036667 on Build GRCh38
rs769769991 Chr.12: 85036575-85036575 on Build GRCh38
rs1029923642 Chr.12: 85036578-85036578 on Build GRCh38
rs926996310 Chr.12: 85036696-85036696 on Build GRCh38
rs941054730 Chr.12: 85036375-85036375 on Build GRCh38
rs151078141 Chr.12: 85036425-85036425 on Build GRCh38
rs74873900 Chr.12: 85036566-85036566 on Build GRCh38
rs999124054 Chr.12: 85036437-85036437 on Build GRCh38
rs975690045 Chr.12: 85036349-85036349 on Build GRCh38
rs918781569 Chr.12: 85036455-85036455 on Build GRCh38
rs941023760 Chr.12: 85036610-85036610 on Build GRCh38
rs1045874116 Chr.12: 85036668-85036668 on Build GRCh38
rs908236300 Chr.12: 85036372-85036372 on Build GRCh38
rs957415912 Chr.12: 85036290-85036290 on Build GRCh38
rs371463061 Chr.12: 85036305-85036306 on Build GRCh38

View Full Product Details