Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85045873-85046372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRIQ1
Entrez Gene ID
84125
Gene Name
leucine rich repeats and IQ motif containing 1
Gene Aliases
-
UniGene
Hs.402200
Gene Chromosome Location
Chr.12: 85036321-85273004 on Build GRCh38
Transcripts
NM_001079910.1 XM_011538817.2 XM_011538818.2 XM_011538819.2 XM_011538823.2 XM_011538824.2 XM_011538825.2 XM_017020019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
944021 c.340T>G p.L114V Chr.12: 85046023-85046023 on Build GRCh38
3738575 c.357A>G p.K119K Chr.12: 85046040-85046040 on Build GRCh38
695604 c.454+9A>C p.? Chr.12: 85046146-85046146 on Build GRCh38
1727023 c.444T>A p.H148Q Chr.12: 85046127-85046127 on Build GRCh38
5795745 c.382G>C p.D128H Chr.12: 85046065-85046065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770286393 Chr.12: 85046022-85046022 on Build GRCh38
rs769342717 Chr.12: 85046134-85046134 on Build GRCh38
rs751719623 Chr.12: 85046151-85046151 on Build GRCh38
rs774053980 Chr.12: 85046032-85046032 on Build GRCh38
rs748850421 Chr.12: 85046012-85046012 on Build GRCh38
rs766607387 Chr.12: 85046148-85046148 on Build GRCh38
rs994880790 Chr.12: 85045912-85045912 on Build GRCh38
rs373571099 Chr.12: 85046188-85046188 on Build GRCh38
rs755814867 Chr.12: 85046071-85046071 on Build GRCh38
rs747105725 Chr.12: 85046123-85046123 on Build GRCh38
rs759258021 Chr.12: 85046034-85046034 on Build GRCh38
rs770540563 Chr.12: 85046088-85046088 on Build GRCh38
rs965065594 Chr.12: 85045961-85045961 on Build GRCh38
rs953837600 Chr.12: 85046338-85046338 on Build GRCh38
rs750557537 Chr.12: 85046071-85046072 on Build GRCh38
rs878957502 Chr.12: 85046270-85046270 on Build GRCh38
rs775245624 Chr.12: 85046105-85046105 on Build GRCh38
rs753870637 Chr.12: 85046158-85046158 on Build GRCh38
rs769699904 Chr.12: 85046003-85046003 on Build GRCh38
rs149503974 Chr.12: 85046086-85046086 on Build GRCh38
rs376473643 Chr.12: 85046133-85046133 on Build GRCh38
rs113738480 Chr.12: 85046219-85046219 on Build GRCh38
rs982591731 Chr.12: 85045899-85045899 on Build GRCh38
rs757934158 Chr.12: 85046182-85046182 on Build GRCh38
rs976452941 Chr.12: 85046330-85046330 on Build GRCh38
rs1034572144 Chr.12: 85046014-85046014 on Build GRCh38
rs777200475 Chr.12: 85046127-85046127 on Build GRCh38
rs148230281 Chr.12: 85046042-85046042 on Build GRCh38
rs368550518 Chr.12: 85046053-85046053 on Build GRCh38
rs779218936 Chr.12: 85045976-85045976 on Build GRCh38
rs747402781 Chr.12: 85046124-85046124 on Build GRCh38
rs202034616 Chr.12: 85046028-85046028 on Build GRCh38
rs750553326 Chr.12: 85045985-85045985 on Build GRCh38
rs754777002 Chr.12: 85045988-85045988 on Build GRCh38
rs372230744 Chr.12: 85046125-85046125 on Build GRCh38
rs561923140 Chr.12: 85045930-85045930 on Build GRCh38
rs140252116 Chr.12: 85046087-85046087 on Build GRCh38
rs930619816 Chr.12: 85046314-85046314 on Build GRCh38
rs778273113 Chr.12: 85046094-85046094 on Build GRCh38
rs905211755 Chr.12: 85046335-85046335 on Build GRCh38
rs750714670 Chr.12: 85046063-85046063 on Build GRCh38
rs758609487 Chr.12: 85046064-85046064 on Build GRCh38
rs1049115860 Chr.12: 85046331-85046331 on Build GRCh38
rs183091794 Chr.12: 85046172-85046172 on Build GRCh38
rs1004460383 Chr.12: 85046214-85046214 on Build GRCh38
rs912013990 Chr.12: 85046244-85046244 on Build GRCh38
rs370031935 Chr.12: 85046068-85046068 on Build GRCh38
rs777529870 Chr.12: 85046004-85046004 on Build GRCh38
rs1054435832 Chr.12: 85045997-85045997 on Build GRCh38
rs754252243 Chr.12: 85046044-85046044 on Build GRCh38
rs369716749 Chr.12: 85046143-85046143 on Build GRCh38
rs773256074 Chr.12: 85046138-85046138 on Build GRCh38
rs1037021299 Chr.12: 85046284-85046284 on Build GRCh38
rs925690697 Chr.12: 85046194-85046194 on Build GRCh38
rs780034416 Chr.12: 85046076-85046077 on Build GRCh38
rs779223027 Chr.12: 85046039-85046041 on Build GRCh38
rs758621180 Chr.12: 85046073-85046074 on Build GRCh38
rs931710587 Chr.12: 85046206-85046206 on Build GRCh38
rs529035839 Chr.12: 85045960-85045960 on Build GRCh38
rs765420248 Chr.12: 85046137-85046137 on Build GRCh38
rs781016761 Chr.12: 85046065-85046065 on Build GRCh38
rs373306195 Chr.12: 85046155-85046155 on Build GRCh38
rs745401350 Chr.12: 85046178-85046178 on Build GRCh38
rs756983066 Chr.12: 85045971-85045972 on Build GRCh38
rs553346589 Chr.12: 85046232-85046232 on Build GRCh38
rs200257302 Chr.12: 85046096-85046096 on Build GRCh38
rs187690645 Chr.12: 85046301-85046301 on Build GRCh38
rs771818423 Chr.12: 85046101-85046101 on Build GRCh38
rs761966699 Chr.12: 85046047-85046047 on Build GRCh38
rs993117644 Chr.12: 85046052-85046052 on Build GRCh38
rs781741529 Chr.12: 85046182-85046182 on Build GRCh38
rs761303056 Chr.12: 85046041-85046041 on Build GRCh38
rs202151623 Chr.12: 85046036-85046036 on Build GRCh38
rs375328847 Chr.12: 85046160-85046160 on Build GRCh38
rs1050146776 Chr.12: 85046208-85046208 on Build GRCh38
rs944895537 Chr.12: 85046260-85046260 on Build GRCh38
rs143115844 Chr.12: 85046223-85046223 on Build GRCh38
rs560842349 Chr.12: 85046198-85046198 on Build GRCh38
rs535640844 Chr.12: 85046337-85046337 on Build GRCh38
rs780965285 Chr.12: 85045993-85045993 on Build GRCh38
rs763788334 Chr.12: 85046156-85046156 on Build GRCh38
rs2305102 Chr.12: 85045996-85045996 on Build GRCh38

View Full Product Details