Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7850727-7851230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTS2
Entrez Gene ID
10911
Gene Name
urotensin 2
Gene Aliases
PRO1068, U-II, UCN2, UII
UniGene
Hs.715862
Gene Chromosome Location
Chr.1: 7846345-7913615 on Build GRCh38
Transcripts
NM_006786.3 NM_021995.2 XM_011540537.2 XM_011540538.1 XM_017000116.1 XM_017000117.1 XM_017000118.1 XM_017000119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6819345 c.166C>T p.L56L Chr.1: 7850860-7850860 on Build GRCh38
2242849 c.118G>A p.A40T Chr.1: 7850908-7850908 on Build GRCh38
3977913 c.205C>A p.Q69K Chr.1: 7850866-7850866 on Build GRCh38
1320891 c.187G>C p.E63Q Chr.1: 7850839-7850839 on Build GRCh38
3977914 c.160C>A p.Q54K Chr.1: 7850866-7850866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188767719 Chr.1: 7851099-7851099 on Build GRCh38
rs1017840609 Chr.1: 7851178-7851178 on Build GRCh38
rs757516672 Chr.1: 7850853-7850853 on Build GRCh38
rs228650 Chr.1: 7851169-7851169 on Build GRCh38
rs746687012 Chr.1: 7850968-7850968 on Build GRCh38
rs228651 Chr.1: 7851039-7851039 on Build GRCh38
rs1037788875 Chr.1: 7850808-7850808 on Build GRCh38
rs995242184 Chr.1: 7850887-7850887 on Build GRCh38
rs754978437 Chr.1: 7850925-7850935 on Build GRCh38
rs183976354 Chr.1: 7850780-7850780 on Build GRCh38
rs771653404 Chr.1: 7850895-7850895 on Build GRCh38
rs779208983 Chr.1: 7850888-7850888 on Build GRCh38
rs367793101 Chr.1: 7850894-7850894 on Build GRCh38
rs141801702 Chr.1: 7850964-7850964 on Build GRCh38
rs753948446 Chr.1: 7850940-7850940 on Build GRCh38
rs920282734 Chr.1: 7850830-7850830 on Build GRCh38
rs964796317 Chr.1: 7851022-7851022 on Build GRCh38
rs749652622 Chr.1: 7850767-7850767 on Build GRCh38
rs750461235 Chr.1: 7850857-7850857 on Build GRCh38
rs776604807 Chr.1: 7850905-7850905 on Build GRCh38
rs780441020 Chr.1: 7850766-7850766 on Build GRCh38
rs770384435 Chr.1: 7850904-7850904 on Build GRCh38
rs753851958 Chr.1: 7850908-7850910 on Build GRCh38
rs954491500 Chr.1: 7850996-7850996 on Build GRCh38
rs773963465 Chr.1: 7850768-7850768 on Build GRCh38
rs772795371 Chr.1: 7850900-7850900 on Build GRCh38
rs192375639 Chr.1: 7850973-7850973 on Build GRCh38
rs778812136 Chr.1: 7850932-7850932 on Build GRCh38
rs990251669 Chr.1: 7851007-7851007 on Build GRCh38
rs188959804 Chr.1: 7850966-7850966 on Build GRCh38
rs767859403 Chr.1: 7850856-7850856 on Build GRCh38
rs771743335 Chr.1: 7850772-7850772 on Build GRCh38
rs1046804689 Chr.1: 7851114-7851114 on Build GRCh38
rs750642306 Chr.1: 7850792-7850792 on Build GRCh38
rs780607056 Chr.1: 7850875-7850875 on Build GRCh38
rs745380769 Chr.1: 7850974-7850974 on Build GRCh38
rs895984654 Chr.1: 7850891-7850891 on Build GRCh38
rs779579181 Chr.1: 7851016-7851016 on Build GRCh38
rs566711319 Chr.1: 7851127-7851127 on Build GRCh38
rs765176159 Chr.1: 7850804-7850804 on Build GRCh38
rs1001808914 Chr.1: 7851149-7851149 on Build GRCh38
rs747886196 Chr.1: 7850997-7850997 on Build GRCh38
rs556637044 Chr.1: 7851032-7851032 on Build GRCh38
rs752566569 Chr.1: 7850833-7850833 on Build GRCh38
rs1024509069 Chr.1: 7851147-7851147 on Build GRCh38
rs1034378952 Chr.1: 7850972-7850973 on Build GRCh38
rs767822642 Chr.1: 7850771-7850771 on Build GRCh38
rs942663406 Chr.1: 7851072-7851072 on Build GRCh38
rs780014134 Chr.1: 7850879-7850882 on Build GRCh38
rs201000488 Chr.1: 7850907-7850907 on Build GRCh38
rs540804193 Chr.1: 7851125-7851125 on Build GRCh38
rs759289088 Chr.1: 7851171-7851171 on Build GRCh38
rs891801480 Chr.1: 7851133-7851133 on Build GRCh38
rs762857373 Chr.1: 7850914-7850914 on Build GRCh38
rs907350523 Chr.1: 7851148-7851148 on Build GRCh38
rs912377261 Chr.1: 7851066-7851066 on Build GRCh38
rs184753681 Chr.1: 7851077-7851077 on Build GRCh38
rs140152981 Chr.1: 7850903-7850903 on Build GRCh38
rs767513022 Chr.1: 7850830-7850830 on Build GRCh38
rs369414632 Chr.1: 7851084-7851084 on Build GRCh38
rs906055704 Chr.1: 7851116-7851116 on Build GRCh38
rs942611286 Chr.1: 7850859-7850859 on Build GRCh38
rs967523536 Chr.1: 7851181-7851181 on Build GRCh38
rs930953611 Chr.1: 7851103-7851103 on Build GRCh38
rs533261976 Chr.1: 7850757-7850757 on Build GRCh38
rs917505746 Chr.1: 7851079-7851079 on Build GRCh38
rs971711208 Chr.1: 7850945-7850945 on Build GRCh38
rs759501467 Chr.1: 7850906-7850906 on Build GRCh38
rs228649 Chr.1: 7851182-7851182 on Build GRCh38
rs973204322 Chr.1: 7851024-7851024 on Build GRCh38
rs913724452 Chr.1: 7850756-7850756 on Build GRCh38
rs760777830 Chr.1: 7850929-7850929 on Build GRCh38
rs1044977690 Chr.1: 7851102-7851102 on Build GRCh38
rs13306065 Chr.1: 7851132-7851132 on Build GRCh38
rs943017838 Chr.1: 7850788-7850788 on Build GRCh38
rs141389663 Chr.1: 7850874-7850874 on Build GRCh38
rs374753674 Chr.1: 7850909-7850909 on Build GRCh38
rs552387361 Chr.1: 7850771-7850771 on Build GRCh38
rs766324228 Chr.1: 7850786-7850786 on Build GRCh38
rs910297503 Chr.1: 7850783-7850783 on Build GRCh38
rs1014839792 Chr.1: 7850899-7850899 on Build GRCh38
rs753046309 Chr.1: 7850958-7850958 on Build GRCh38
rs755497614 Chr.1: 7850884-7850884 on Build GRCh38
rs528396117 Chr.1: 7850932-7850932 on Build GRCh38
rs201977733 Chr.1: 7850908-7850908 on Build GRCh38
rs1493615 Chr.1: 7851168-7851168 on Build GRCh38
rs79479445 Chr.1: 7851161-7851161 on Build GRCh38
rs749800934 Chr.1: 7850879-7850879 on Build GRCh38
rs112754200 Chr.1: 7851186-7851186 on Build GRCh38
rs759370503 Chr.1: 7850797-7850797 on Build GRCh38
rs147306007 Chr.1: 7850915-7850915 on Build GRCh38
rs891005798 Chr.1: 7850839-7850839 on Build GRCh38
rs775521806 Chr.1: 7851118-7851118 on Build GRCh38
rs779405487 Chr.1: 7850955-7850955 on Build GRCh38
rs776497171 Chr.1: 7850787-7850787 on Build GRCh38
rs1009479997 Chr.1: 7850951-7850951 on Build GRCh38
rs755409662 Chr.1: 7850941-7850941 on Build GRCh38
rs1010246238 Chr.1: 7851150-7851150 on Build GRCh38
rs772710601 Chr.1: 7850773-7850773 on Build GRCh38
rs145544704 Chr.1: 7851049-7851049 on Build GRCh38

View Full Product Details