Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8974255-8974761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA6
Entrez Gene ID
765
Gene Name
carbonic anhydrase 6
Gene Aliases
CA-VI, GUSTIN
UniGene
Hs.100322
Gene Chromosome Location
Chr.1: 8945786-8975092 on Build GRCh38
Transcripts
NM_001215.3 NM_001270500.1 NM_001270501.1 NM_001270502.1 XM_011542083.2 XM_011542084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493222 c.866T>C p.F289S Chr.1: 8974643-8974643 on Build GRCh38
4648179 c.908T>C p.L303S Chr.1: 8974685-8974685 on Build GRCh38
3493223 c.870G>A p.Q290Q Chr.1: 8974647-8974647 on Build GRCh38
6127906 c.880C>T p.H294Y Chr.1: 8974657-8974657 on Build GRCh38
4668244 c.853C>A p.L285I Chr.1: 8974630-8974630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532265641 Chr.1: 8974610-8974610 on Build GRCh38
rs187434042 Chr.1: 8974378-8974378 on Build GRCh38
rs1010418588 Chr.1: 8974542-8974542 on Build GRCh38
rs376589511 Chr.1: 8974330-8974330 on Build GRCh38
rs61749464 Chr.1: 8974658-8974658 on Build GRCh38
rs762564967 Chr.1: 8974367-8974367 on Build GRCh38
rs766232011 Chr.1: 8974605-8974605 on Build GRCh38
rs754130068 Chr.1: 8974300-8974300 on Build GRCh38
rs200236268 Chr.1: 8974646-8974646 on Build GRCh38
rs115942471 Chr.1: 8974551-8974551 on Build GRCh38
rs781660920 Chr.1: 8974487-8974487 on Build GRCh38
rs767617801 Chr.1: 8974600-8974600 on Build GRCh38
rs540873466 Chr.1: 8974689-8974689 on Build GRCh38
rs36181061 Chr.1: 8974674-8974674 on Build GRCh38
rs1047574540 Chr.1: 8974654-8974654 on Build GRCh38
rs769496436 Chr.1: 8974360-8974360 on Build GRCh38
rs776181637 Chr.1: 8974433-8974447 on Build GRCh38
rs530555064 Chr.1: 8974287-8974287 on Build GRCh38
rs377187876 Chr.1: 8974409-8974409 on Build GRCh38
rs751655020 Chr.1: 8974635-8974635 on Build GRCh38
rs748293927 Chr.1: 8974326-8974326 on Build GRCh38
rs952606546 Chr.1: 8974533-8974533 on Build GRCh38
rs768263093 Chr.1: 8974734-8974734 on Build GRCh38
rs533773944 Chr.1: 8974513-8974513 on Build GRCh38
rs773672221 Chr.1: 8974594-8974594 on Build GRCh38
rs761384115 Chr.1: 8974596-8974596 on Build GRCh38
rs771348039 Chr.1: 8974354-8974354 on Build GRCh38
rs759124833 Chr.1: 8974713-8974718 on Build GRCh38
rs113261721 Chr.1: 8974739-8974739 on Build GRCh38
rs147408666 Chr.1: 8974644-8974644 on Build GRCh38
rs949046648 Chr.1: 8974352-8974352 on Build GRCh38
rs966461851 Chr.1: 8974552-8974552 on Build GRCh38
rs1001817496 Chr.1: 8974532-8974532 on Build GRCh38
rs982717414 Chr.1: 8974321-8974321 on Build GRCh38
rs3765964 Chr.1: 8974362-8974362 on Build GRCh38
rs772019791 Chr.1: 8974335-8974335 on Build GRCh38
rs3765963 Chr.1: 8974539-8974539 on Build GRCh38
rs77420168 Chr.1: 8974462-8974462 on Build GRCh38
rs758532550 Chr.1: 8974473-8974473 on Build GRCh38
rs758491150 Chr.1: 8974623-8974623 on Build GRCh38
rs34125121 Chr.1: 8974339-8974339 on Build GRCh38
rs926343680 Chr.1: 8974587-8974587 on Build GRCh38
rs184177635 Chr.1: 8974311-8974311 on Build GRCh38
rs79381171 Chr.1: 8974494-8974494 on Build GRCh38
rs759191599 Chr.1: 8974358-8974358 on Build GRCh38
rs369632657 Chr.1: 8974529-8974529 on Build GRCh38
rs184099715 Chr.1: 8974556-8974556 on Build GRCh38
rs148213961 Chr.1: 8974301-8974301 on Build GRCh38
rs764238187 Chr.1: 8974627-8974627 on Build GRCh38
rs923187787 Chr.1: 8974616-8974616 on Build GRCh38
rs771273995 Chr.1: 8974482-8974482 on Build GRCh38
rs753401957 Chr.1: 8974358-8974358 on Build GRCh38
rs79453719 Chr.1: 8974450-8974450 on Build GRCh38
rs780343840 Chr.1: 8974684-8974684 on Build GRCh38
rs944324311 Chr.1: 8974710-8974710 on Build GRCh38
rs111329595 Chr.1: 8974481-8974481 on Build GRCh38
rs774042044 Chr.1: 8974737-8974737 on Build GRCh38
rs908197068 Chr.1: 8974347-8974347 on Build GRCh38
rs191566022 Chr.1: 8974387-8974387 on Build GRCh38
rs745918744 Chr.1: 8974690-8974690 on Build GRCh38
rs747433301 Chr.1: 8974350-8974350 on Build GRCh38
rs755957225 Chr.1: 8974449-8974449 on Build GRCh38
rs937491601 Chr.1: 8974398-8974398 on Build GRCh38
rs746335041 Chr.1: 8974666-8974666 on Build GRCh38
rs776532966 Chr.1: 8974303-8974303 on Build GRCh38
rs74050967 Chr.1: 8974424-8974424 on Build GRCh38
rs1027213548 Chr.1: 8974530-8974530 on Build GRCh38
rs770714367 Chr.1: 8974312-8974312 on Build GRCh38
rs779271357 Chr.1: 8974312-8974312 on Build GRCh38
rs769991497 Chr.1: 8974368-8974368 on Build GRCh38
rs762467917 Chr.1: 8974578-8974578 on Build GRCh38
rs192896447 Chr.1: 8974288-8974288 on Build GRCh38
rs763472285 Chr.1: 8974369-8974369 on Build GRCh38
rs142823347 Chr.1: 8974555-8974555 on Build GRCh38
rs752932558 Chr.1: 8974465-8974465 on Build GRCh38
rs996978730 Chr.1: 8974501-8974501 on Build GRCh38
rs776922659 Chr.1: 8974357-8974357 on Build GRCh38
rs778379935 Chr.1: 8974342-8974342 on Build GRCh38
rs942150419 Chr.1: 8974673-8974673 on Build GRCh38
rs369091466 Chr.1: 8974675-8974675 on Build GRCh38
rs372650598 Chr.1: 8974601-8974601 on Build GRCh38
rs116135939 Chr.1: 8974488-8974488 on Build GRCh38
rs766044610 Chr.1: 8974477-8974477 on Build GRCh38
rs371826757 Chr.1: 8974534-8974534 on Build GRCh38

View Full Product Details