Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9014551-9015048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A7
Entrez Gene ID
155184
Gene Name
solute carrier family 2 member 7
Gene Aliases
GLUT7
UniGene
Hs.531239
Gene Chromosome Location
Chr.1: 8992534-9026384 on Build GRCh38
Transcripts
NM_207420.2 XM_011540824.2 XM_011540825.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5480402 c.716-5C>T p.? Chr.1: 9014873-9014873 on Build GRCh38
5992819 c.789G>C p.E263D Chr.1: 9014795-9014795 on Build GRCh38
5481454 c.832C>T p.R278W Chr.1: 9014752-9014752 on Build GRCh38
3419557 c.902C>T p.A301V Chr.1: 9014682-9014682 on Build GRCh38
1344792 c.795C>T p.A265A Chr.1: 9014789-9014789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34287135 Chr.1: 9014755-9014755 on Build GRCh38
rs748230912 Chr.1: 9014789-9014789 on Build GRCh38
rs556830241 Chr.1: 9014720-9014720 on Build GRCh38
rs980211835 Chr.1: 9014842-9014842 on Build GRCh38
rs747372560 Chr.1: 9014851-9014851 on Build GRCh38
rs370934872 Chr.1: 9014890-9014890 on Build GRCh38
rs751809784 Chr.1: 9014716-9014716 on Build GRCh38
rs756277044 Chr.1: 9014791-9014791 on Build GRCh38
rs1015364309 Chr.1: 9015023-9015023 on Build GRCh38
rs770481613 Chr.1: 9014662-9014662 on Build GRCh38
rs535655666 Chr.1: 9014989-9014989 on Build GRCh38
rs144688962 Chr.1: 9014844-9014844 on Build GRCh38
rs373819947 Chr.1: 9014859-9014859 on Build GRCh38
rs750624529 Chr.1: 9014789-9014789 on Build GRCh38
rs759561594 Chr.1: 9014910-9014910 on Build GRCh38
rs748644098 Chr.1: 9014841-9014841 on Build GRCh38
rs543972558 Chr.1: 9014983-9014983 on Build GRCh38
rs761307002 Chr.1: 9014693-9014693 on Build GRCh38
rs749385393 Chr.1: 9014793-9014793 on Build GRCh38
rs761675328 Chr.1: 9014893-9014893 on Build GRCh38
rs1039463778 Chr.1: 9014990-9014990 on Build GRCh38
rs749997562 Chr.1: 9014698-9014698 on Build GRCh38
rs983511639 Chr.1: 9014639-9014639 on Build GRCh38
rs910683807 Chr.1: 9014930-9014930 on Build GRCh38
rs771765656 Chr.1: 9014745-9014745 on Build GRCh38
rs766339197 Chr.1: 9014706-9014706 on Build GRCh38
rs553834240 Chr.1: 9014743-9014743 on Build GRCh38
rs527732606 Chr.1: 9014995-9014995 on Build GRCh38
rs544616432 Chr.1: 9014901-9014901 on Build GRCh38
rs779315736 Chr.1: 9014840-9014840 on Build GRCh38
rs767883719 Chr.1: 9014781-9014781 on Build GRCh38
rs748506077 Chr.1: 9014797-9014797 on Build GRCh38
rs1050711231 Chr.1: 9014957-9014957 on Build GRCh38
rs776073629 Chr.1: 9014804-9014804 on Build GRCh38
rs772823908 Chr.1: 9014751-9014751 on Build GRCh38
rs745410612 Chr.1: 9014872-9014872 on Build GRCh38
rs202111875 Chr.1: 9014806-9014806 on Build GRCh38
rs752491553 Chr.1: 9014779-9014779 on Build GRCh38
rs745938213 Chr.1: 9014641-9014641 on Build GRCh38
rs780217546 Chr.1: 9014792-9014792 on Build GRCh38
rs749701167 Chr.1: 9014739-9014739 on Build GRCh38
rs758779294 Chr.1: 9014843-9014843 on Build GRCh38
rs971466797 Chr.1: 9014837-9014837 on Build GRCh38
rs534062397 Chr.1: 9014636-9014636 on Build GRCh38
rs952581511 Chr.1: 9014685-9014685 on Build GRCh38
rs763923070 Chr.1: 9014812-9014812 on Build GRCh38
rs1040835291 Chr.1: 9014956-9014956 on Build GRCh38
rs944869286 Chr.1: 9014933-9014933 on Build GRCh38
rs1025513705 Chr.1: 9014682-9014682 on Build GRCh38
rs774648387 Chr.1: 9014742-9014742 on Build GRCh38
rs754821597 Chr.1: 9014708-9014708 on Build GRCh38
rs917319840 Chr.1: 9014642-9014642 on Build GRCh38
rs369016105 Chr.1: 9014818-9014818 on Build GRCh38
rs912269711 Chr.1: 9014959-9014959 on Build GRCh38
rs776109119 Chr.1: 9014664-9014664 on Build GRCh38
rs147603199 Chr.1: 9014735-9014735 on Build GRCh38
rs760159285 Chr.1: 9014699-9014699 on Build GRCh38
rs762915853 Chr.1: 9014885-9014885 on Build GRCh38
rs200306616 Chr.1: 9014727-9014727 on Build GRCh38
rs765214485 Chr.1: 9014760-9014760 on Build GRCh38
rs780876954 Chr.1: 9014861-9014861 on Build GRCh38
rs777142618 Chr.1: 9014800-9014800 on Build GRCh38
rs775302926 Chr.1: 9014877-9014877 on Build GRCh38
rs758190479 Chr.1: 9014780-9014780 on Build GRCh38
rs1751676 Chr.1: 9014669-9014669 on Build GRCh38
rs1556757 Chr.1: 9014873-9014873 on Build GRCh38
rs148489455 Chr.1: 9014788-9014788 on Build GRCh38
rs1003668572 Chr.1: 9014864-9014864 on Build GRCh38
rs115204359 Chr.1: 9014803-9014803 on Build GRCh38
rs942406202 Chr.1: 9014989-9014989 on Build GRCh38
rs79414710 Chr.1: 9014998-9014998 on Build GRCh38
rs780075652 Chr.1: 9014811-9014811 on Build GRCh38
rs774068416 Chr.1: 9014680-9014680 on Build GRCh38
rs1049723506 Chr.1: 9015001-9015001 on Build GRCh38
rs753910234 Chr.1: 9014830-9014830 on Build GRCh38
rs769071889 Chr.1: 9014741-9014741 on Build GRCh38
rs768708903 Chr.1: 9014889-9014889 on Build GRCh38
rs1040687411 Chr.1: 9014605-9014605 on Build GRCh38
rs566567307 Chr.1: 9014985-9014985 on Build GRCh38
rs201194130 Chr.1: 9014670-9014670 on Build GRCh38
rs12402973 Chr.1: 9014802-9014802 on Build GRCh38
rs755498344 Chr.1: 9014794-9014794 on Build GRCh38
rs772382963 Chr.1: 9014798-9014798 on Build GRCh38
rs141021827 Chr.1: 9014831-9014831 on Build GRCh38
rs779456894 Chr.1: 9014796-9014796 on Build GRCh38
rs927186023 Chr.1: 9014702-9014702 on Build GRCh38
rs865854251 Chr.1: 9014709-9014709 on Build GRCh38
rs78914166 Chr.1: 9014810-9014810 on Build GRCh38
rs754047524 Chr.1: 9014903-9014903 on Build GRCh38
rs941246924 Chr.1: 9015013-9015013 on Build GRCh38
rs765822995 Chr.1: 9014752-9014752 on Build GRCh38
rs766692718 Chr.1: 9014815-9014815 on Build GRCh38
rs946893880 Chr.1: 9014786-9014786 on Build GRCh38
rs149495842 Chr.1: 9014707-9014707 on Build GRCh38
rs950708358 Chr.1: 9014592-9014592 on Build GRCh38
rs144633163 Chr.1: 9014719-9014719 on Build GRCh38
rs780730733 Chr.1: 9014737-9014737 on Build GRCh38
rs144995721 Chr.1: 9014586-9014586 on Build GRCh38
rs149017120 Chr.1: 9014814-9014814 on Build GRCh38
rs867783283 Chr.1: 9014898-9014898 on Build GRCh38
rs986713854 Chr.1: 9014911-9014911 on Build GRCh38
rs759322593 Chr.1: 9014758-9014758 on Build GRCh38

View Full Product Details