Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11043258-11043768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MASP2
Entrez Gene ID
10747
Gene Name
mannan binding lectin serine peptidase 2
Gene Aliases
MAP19, MASP-2, MASP1P1, sMAP
UniGene
Hs.655645
Gene Chromosome Location
Chr.1: 11026523-11047239 on Build GRCh38
Transcripts
NM_006610.3 XM_017000097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471065 c.626C>T p.S209F Chr.1: 11043454-11043454 on Build GRCh38
5491019 c.584A>G p.E195G Chr.1: 11043496-11043496 on Build GRCh38
4556013 c.675G>T p.L225L Chr.1: 11043405-11043405 on Build GRCh38
5818898 c.717C>T p.T239T Chr.1: 11043363-11043363 on Build GRCh38
296707 c.729C>T p.Y243Y Chr.1: 11043351-11043351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026839706 Chr.1: 11043741-11043741 on Build GRCh38
rs763440352 Chr.1: 11043562-11043562 on Build GRCh38
rs758613711 Chr.1: 11043297-11043297 on Build GRCh38
rs771680406 Chr.1: 11043464-11043464 on Build GRCh38
rs577897621 Chr.1: 11043588-11043588 on Build GRCh38
rs780948027 Chr.1: 11043327-11043327 on Build GRCh38
rs914217375 Chr.1: 11043717-11043717 on Build GRCh38
rs746782434 Chr.1: 11043324-11043324 on Build GRCh38
rs137985602 Chr.1: 11043605-11043605 on Build GRCh38
rs7536030 Chr.1: 11043351-11043351 on Build GRCh38
rs760909511 Chr.1: 11043361-11043361 on Build GRCh38
rs556379403 Chr.1: 11043584-11043584 on Build GRCh38
rs924334847 Chr.1: 11043676-11043676 on Build GRCh38
rs747850339 Chr.1: 11043460-11043460 on Build GRCh38
rs3737612 Chr.1: 11043295-11043295 on Build GRCh38
rs949069867 Chr.1: 11043511-11043511 on Build GRCh38
rs759872641 Chr.1: 11043386-11043386 on Build GRCh38
rs375366541 Chr.1: 11043477-11043477 on Build GRCh38
rs771723749 Chr.1: 11043524-11043524 on Build GRCh38
rs747676906 Chr.1: 11043521-11043521 on Build GRCh38
rs775086060 Chr.1: 11043296-11043296 on Build GRCh38
rs756947952 Chr.1: 11043419-11043419 on Build GRCh38
rs749243026 Chr.1: 11043509-11043509 on Build GRCh38
rs753979683 Chr.1: 11043454-11043455 on Build GRCh38
rs768313255 Chr.1: 11043457-11043457 on Build GRCh38
rs867601234 Chr.1: 11043395-11043395 on Build GRCh38
rs761850554 Chr.1: 11043568-11043568 on Build GRCh38
rs981173952 Chr.1: 11043675-11043675 on Build GRCh38
rs571956401 Chr.1: 11043692-11043692 on Build GRCh38
rs750213898 Chr.1: 11043489-11043489 on Build GRCh38
rs200464626 Chr.1: 11043503-11043503 on Build GRCh38
rs755421910 Chr.1: 11043739-11043739 on Build GRCh38
rs769743224 Chr.1: 11043425-11043425 on Build GRCh38
rs778471941 Chr.1: 11043518-11043518 on Build GRCh38
rs374783350 Chr.1: 11043394-11043394 on Build GRCh38
rs774082092 Chr.1: 11043458-11043458 on Build GRCh38
rs1008723809 Chr.1: 11043368-11043368 on Build GRCh38
rs934397711 Chr.1: 11043683-11043683 on Build GRCh38
rs755908031 Chr.1: 11043587-11043587 on Build GRCh38
rs774460559 Chr.1: 11043564-11043564 on Build GRCh38
rs746742964 Chr.1: 11043398-11043398 on Build GRCh38
rs748760302 Chr.1: 11043454-11043454 on Build GRCh38
rs991813670 Chr.1: 11043649-11043649 on Build GRCh38
rs768403060 Chr.1: 11043354-11043354 on Build GRCh38
rs780191217 Chr.1: 11043434-11043434 on Build GRCh38
rs754479594 Chr.1: 11043512-11043512 on Build GRCh38
rs566765351 Chr.1: 11043469-11043469 on Build GRCh38
rs746193180 Chr.1: 11043526-11043526 on Build GRCh38
rs780086747 Chr.1: 11043372-11043373 on Build GRCh38
rs781032168 Chr.1: 11043420-11043420 on Build GRCh38
rs531290078 Chr.1: 11043728-11043728 on Build GRCh38
rs770777702 Chr.1: 11043326-11043326 on Build GRCh38
rs372145572 Chr.1: 11043381-11043381 on Build GRCh38
rs1045425659 Chr.1: 11043280-11043280 on Build GRCh38
rs778008254 Chr.1: 11043301-11043301 on Build GRCh38
rs902854168 Chr.1: 11043286-11043286 on Build GRCh38
rs971598668 Chr.1: 11043660-11043660 on Build GRCh38
rs72550853 Chr.1: 11043468-11043468 on Build GRCh38
rs775905080 Chr.1: 11043539-11043539 on Build GRCh38
rs28604502 Chr.1: 11043627-11043627 on Build GRCh38
rs187907824 Chr.1: 11043563-11043563 on Build GRCh38
rs898466058 Chr.1: 11043629-11043629 on Build GRCh38
rs147658846 Chr.1: 11043350-11043350 on Build GRCh38
rs949826923 Chr.1: 11043278-11043278 on Build GRCh38
rs78775425 Chr.1: 11043525-11043525 on Build GRCh38
rs932876565 Chr.1: 11043520-11043520 on Build GRCh38
rs550788156 Chr.1: 11043730-11043730 on Build GRCh38
rs140547549 Chr.1: 11043453-11043453 on Build GRCh38
rs7536396 Chr.1: 11043727-11043727 on Build GRCh38
rs758518990 Chr.1: 11043402-11043402 on Build GRCh38
rs182267163 Chr.1: 11043634-11043634 on Build GRCh38
rs769682108 Chr.1: 11043340-11043340 on Build GRCh38
rs1008322435 Chr.1: 11043550-11043550 on Build GRCh38
rs989796662 Chr.1: 11043686-11043686 on Build GRCh38
rs191530740 Chr.1: 11043569-11043569 on Build GRCh38
rs754240264 Chr.1: 11043384-11043384 on Build GRCh38
rs745718846 Chr.1: 11043421-11043421 on Build GRCh38
rs368040764 Chr.1: 11043338-11043338 on Build GRCh38
rs373318594 Chr.1: 11043473-11043473 on Build GRCh38
rs750682072 Chr.1: 11043318-11043319 on Build GRCh38
rs1022831902 Chr.1: 11043658-11043658 on Build GRCh38
rs891278287 Chr.1: 11043534-11043534 on Build GRCh38
rs369013383 Chr.1: 11043450-11043450 on Build GRCh38
rs370819108 Chr.1: 11043726-11043726 on Build GRCh38
rs1054646787 Chr.1: 11043495-11043495 on Build GRCh38
rs774170040 Chr.1: 11043355-11043355 on Build GRCh38
rs971980318 Chr.1: 11043413-11043413 on Build GRCh38
rs537478265 Chr.1: 11043685-11043685 on Build GRCh38
rs1008226989 Chr.1: 11043721-11043721 on Build GRCh38
rs777918417 Chr.1: 11043403-11043403 on Build GRCh38
rs897180827 Chr.1: 11043580-11043580 on Build GRCh38
rs751834466 Chr.1: 11043409-11043409 on Build GRCh38
rs777047280 Chr.1: 11043467-11043467 on Build GRCh38
rs201189421 Chr.1: 11043387-11043387 on Build GRCh38
rs755997737 Chr.1: 11043499-11043499 on Build GRCh38
rs756305173 Chr.1: 11043342-11043342 on Build GRCh38
rs770171499 Chr.1: 11043531-11043531 on Build GRCh38
rs751652769 Chr.1: 11043472-11043472 on Build GRCh38
rs72550852 Chr.1: 11043291-11043291 on Build GRCh38

View Full Product Details