Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98733429-98733943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
XM_017019653.1 NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
XM_017019653.1 NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1

SNP Detail

SNP ID Chromosome Location
rs922994856 Chr.12: 98733673-98733673 on Build GRCh38
rs188259081 Chr.12: 98733738-98733738 on Build GRCh38
rs553163136 Chr.12: 98733806-98733806 on Build GRCh38
rs140030216 Chr.12: 98733474-98733474 on Build GRCh38
rs532822025 Chr.12: 98733660-98733660 on Build GRCh38
rs543790149 Chr.12: 98733509-98733509 on Build GRCh38
rs894963528 Chr.12: 98733711-98733711 on Build GRCh38
rs973165353 Chr.12: 98733831-98733831 on Build GRCh38
rs11296996 Chr.12: 98733918-98733918 on Build GRCh38
rs952025064 Chr.12: 98733615-98733615 on Build GRCh38
rs143208155 Chr.12: 98733579-98733579 on Build GRCh38
rs539913464 Chr.12: 98733875-98733875 on Build GRCh38
rs1055297552 Chr.12: 98733772-98733772 on Build GRCh38
rs776940896 Chr.12: 98733591-98733591 on Build GRCh38
rs1055181303 Chr.12: 98733479-98733479 on Build GRCh38
rs188204280 Chr.12: 98733469-98733469 on Build GRCh38
rs895316251 Chr.12: 98733499-98733499 on Build GRCh38
rs1013659413 Chr.12: 98733514-98733514 on Build GRCh38
rs895348769 Chr.12: 98733795-98733795 on Build GRCh38
rs1131019 Chr.12: 98733572-98733572 on Build GRCh38
rs964836290 Chr.12: 98733475-98733475 on Build GRCh38
rs567542438 Chr.12: 98733797-98733797 on Build GRCh38
rs950435537 Chr.12: 98733866-98733866 on Build GRCh38
rs955143409 Chr.12: 98733796-98733796 on Build GRCh38
rs180779783 Chr.12: 98733751-98733751 on Build GRCh38
rs925565865 Chr.12: 98733743-98733743 on Build GRCh38
rs920346642 Chr.12: 98733842-98733842 on Build GRCh38
rs1026245399 Chr.12: 98733580-98733580 on Build GRCh38
rs865866330 Chr.12: 98733695-98733695 on Build GRCh38
rs747092806 Chr.12: 98733484-98733484 on Build GRCh38
rs192992501 Chr.12: 98733547-98733547 on Build GRCh38
rs549041592 Chr.12: 98733774-98733774 on Build GRCh38
rs961426364 Chr.12: 98733576-98733576 on Build GRCh38
rs983090674 Chr.12: 98733916-98733916 on Build GRCh38
rs375602833 Chr.12: 98733452-98733452 on Build GRCh38
rs531488001 Chr.12: 98733805-98733805 on Build GRCh38
rs984622581 Chr.12: 98733617-98733617 on Build GRCh38
rs1043157215 Chr.12: 98733742-98733742 on Build GRCh38
rs1031644756 Chr.12: 98733771-98733771 on Build GRCh38
rs184748337 Chr.12: 98733685-98733685 on Build GRCh38
rs964898658 Chr.12: 98733812-98733812 on Build GRCh38
rs758831215 Chr.12: 98733859-98733859 on Build GRCh38
rs12829605 Chr.12: 98733653-98733653 on Build GRCh38
rs1047344827 Chr.12: 98733633-98733633 on Build GRCh38
rs975856549 Chr.12: 98733669-98733669 on Build GRCh38
rs562393330 Chr.12: 98733539-98733539 on Build GRCh38
rs572588413 Chr.12: 98733815-98733815 on Build GRCh38

View Full Product Details