Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100473981-100474477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1H4
Entrez Gene ID
9971
Gene Name
nuclear receptor subfamily 1 group H member 4
Gene Aliases
BAR, FXR, HRR-1, HRR1, RIP14
UniGene
Hs.282735
Gene Chromosome Location
Chr.12: 100473773-100563867 on Build GRCh38
Transcripts
NM_001206977.1 NM_001206978.1 NM_001206979.1 NM_005123.3 NR_135146.1

SNP Detail

SNP ID Chromosome Location
rs570372270 Chr.12: 100474232-100474232 on Build GRCh38
rs555723748 Chr.12: 100474374-100474374 on Build GRCh38
rs965328012 Chr.12: 100474307-100474307 on Build GRCh38
rs113640779 Chr.12: 100474212-100474212 on Build GRCh38
rs536543777 Chr.12: 100474382-100474382 on Build GRCh38
rs1032667527 Chr.12: 100474301-100474301 on Build GRCh38
rs967357846 Chr.12: 100474269-100474269 on Build GRCh38
rs78243656 Chr.12: 100474375-100474375 on Build GRCh38
rs949630115 Chr.12: 100474431-100474431 on Build GRCh38
rs916867064 Chr.12: 100474429-100474429 on Build GRCh38
rs1024602608 Chr.12: 100474259-100474259 on Build GRCh38
rs532510468 Chr.12: 100474044-100474044 on Build GRCh38
rs749871132 Chr.12: 100474067-100474067 on Build GRCh38
rs991172227 Chr.12: 100474424-100474424 on Build GRCh38
rs117796786 Chr.12: 100474404-100474404 on Build GRCh38
rs1027095921 Chr.12: 100474255-100474255 on Build GRCh38
rs760641423 Chr.12: 100474346-100474346 on Build GRCh38
rs1006928965 Chr.12: 100474290-100474290 on Build GRCh38
rs76613494 Chr.12: 100474151-100474151 on Build GRCh38
rs913030951 Chr.12: 100474031-100474031 on Build GRCh38
rs945811750 Chr.12: 100474053-100474053 on Build GRCh38
rs1018690625 Chr.12: 100474302-100474302 on Build GRCh38
rs978549469 Chr.12: 100474388-100474388 on Build GRCh38
rs540393415 Chr.12: 100474434-100474434 on Build GRCh38
rs1046609866 Chr.12: 100474192-100474192 on Build GRCh38
rs998916785 Chr.12: 100474276-100474276 on Build GRCh38
rs767470035 Chr.12: 100474083-100474083 on Build GRCh38
rs976281352 Chr.12: 100474421-100474421 on Build GRCh38
rs562805327 Chr.12: 100474125-100474125 on Build GRCh38
rs148564242 Chr.12: 100474392-100474392 on Build GRCh38
rs976595232 Chr.12: 100474012-100474012 on Build GRCh38
rs910743395 Chr.12: 100474051-100474051 on Build GRCh38
rs560741656 Chr.12: 100474191-100474191 on Build GRCh38
rs994272379 Chr.12: 100474245-100474245 on Build GRCh38
rs533661508 Chr.12: 100474190-100474190 on Build GRCh38
rs565407004 Chr.12: 100474441-100474441 on Build GRCh38
rs537872593 Chr.12: 100474233-100474233 on Build GRCh38
rs1002320724 Chr.12: 100474175-100474175 on Build GRCh38
rs531075573 Chr.12: 100474045-100474045 on Build GRCh38
rs929609529 Chr.12: 100474443-100474443 on Build GRCh38
rs887855402 Chr.12: 100474163-100474163 on Build GRCh38
rs985818613 Chr.12: 100474364-100474364 on Build GRCh38
rs552841361 Chr.12: 100474046-100474046 on Build GRCh38
rs888517511 Chr.12: 100474272-100474272 on Build GRCh38
rs548454703 Chr.12: 100474160-100474160 on Build GRCh38
rs925759271 Chr.12: 100474403-100474403 on Build GRCh38
rs566617463 Chr.12: 100474162-100474162 on Build GRCh38
rs753753732 Chr.12: 100474433-100474433 on Build GRCh38
rs957100173 Chr.12: 100474363-100474363 on Build GRCh38
rs75762522 Chr.12: 100474117-100474117 on Build GRCh38
rs755912891 Chr.12: 100474341-100474341 on Build GRCh38

View Full Product Details