Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110381631-110382130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
UniGene
Hs.719935
Gene Chromosome Location
Chr.12: 110372900-110403730 on Build GRCh38
Transcripts
NM_001137664.1 NM_016238.2 XM_005253903.3 XM_011538454.2 XM_017019425.1 XM_017019426.1 XM_017019427.1 XM_017019428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687948 c.1189C>T p.R397W Chr.12: 110381797-110381797 on Build GRCh38
4038643 c.967C>T p.R323W Chr.12: 110381917-110381917 on Build GRCh38
5882183 c.1008T>A p.S336R Chr.12: 110381876-110381876 on Build GRCh38
6838392 c.1007G>C p.S336T Chr.12: 110381877-110381877 on Build GRCh38
4929609 c.1079A>G p.Y360C Chr.12: 110381907-110381907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781367378 Chr.12: 110381780-110381780 on Build GRCh38
rs1021529656 Chr.12: 110382086-110382086 on Build GRCh38
rs760262629 Chr.12: 110381968-110381968 on Build GRCh38
rs775145723 Chr.12: 110381716-110381716 on Build GRCh38
rs745876646 Chr.12: 110381973-110381974 on Build GRCh38
rs200556644 Chr.12: 110381954-110381954 on Build GRCh38
rs778283085 Chr.12: 110381972-110381973 on Build GRCh38
rs1022002717 Chr.12: 110381727-110381727 on Build GRCh38
rs140284932 Chr.12: 110382024-110382024 on Build GRCh38
rs753319774 Chr.12: 110381879-110381879 on Build GRCh38
rs759361572 Chr.12: 110381951-110381952 on Build GRCh38
rs756538413 Chr.12: 110381676-110381676 on Build GRCh38
rs930175620 Chr.12: 110381907-110381907 on Build GRCh38
rs755299232 Chr.12: 110381969-110381970 on Build GRCh38
rs757263211 Chr.12: 110382082-110382082 on Build GRCh38
rs779542207 Chr.12: 110381962-110381962 on Build GRCh38
rs756834403 Chr.12: 110381971-110381972 on Build GRCh38
rs553248526 Chr.12: 110381954-110381954 on Build GRCh38
rs911292528 Chr.12: 110381796-110381796 on Build GRCh38
rs796595026 Chr.12: 110381955-110381956 on Build GRCh38
rs778681300 Chr.12: 110381769-110381769 on Build GRCh38
rs1002637210 Chr.12: 110381667-110381667 on Build GRCh38
rs954719143 Chr.12: 110382094-110382094 on Build GRCh38
rs914482085 Chr.12: 110381969-110381969 on Build GRCh38
rs78217750 Chr.12: 110381956-110381956 on Build GRCh38
rs765197462 Chr.12: 110381955-110381958 on Build GRCh38
rs570065421 Chr.12: 110382071-110382071 on Build GRCh38
rs145251143 Chr.12: 110382076-110382076 on Build GRCh38
rs764136297 Chr.12: 110381744-110381744 on Build GRCh38
rs368110252 Chr.12: 110382090-110382090 on Build GRCh38
rs775879078 Chr.12: 110381966-110381966 on Build GRCh38
rs1035834999 Chr.12: 110381675-110381675 on Build GRCh38
rs938459940 Chr.12: 110382054-110382054 on Build GRCh38
rs752310239 Chr.12: 110381737-110381737 on Build GRCh38
rs1057330236 Chr.12: 110382055-110382055 on Build GRCh38
rs938444673 Chr.12: 110382032-110382032 on Build GRCh38
rs61942585 Chr.12: 110381955-110381955 on Build GRCh38
rs975364525 Chr.12: 110381967-110381967 on Build GRCh38
rs111406162 Chr.12: 110381954-110381955 on Build GRCh38
rs777720148 Chr.12: 110381959-110381959 on Build GRCh38
rs201433415 Chr.12: 110382079-110382079 on Build GRCh38
rs761999222 Chr.12: 110381955-110381957 on Build GRCh38
rs200431504 Chr.12: 110381735-110381736 on Build GRCh38
rs771626202 Chr.12: 110381961-110381961 on Build GRCh38
rs745656677 Chr.12: 110381776-110381776 on Build GRCh38
rs748298890 Chr.12: 110381782-110381782 on Build GRCh38
rs772059942 Chr.12: 110381952-110381953 on Build GRCh38
rs757172491 Chr.12: 110381757-110381757 on Build GRCh38
rs760339465 Chr.12: 110381874-110381874 on Build GRCh38
rs200704477 Chr.12: 110381728-110381728 on Build GRCh38
rs749383338 Chr.12: 110381960-110381960 on Build GRCh38
rs138556976 Chr.12: 110381708-110381708 on Build GRCh38
rs755560278 Chr.12: 110381742-110381742 on Build GRCh38
rs529845970 Chr.12: 110381946-110381946 on Build GRCh38
rs965473042 Chr.12: 110381830-110381830 on Build GRCh38
rs767331395 Chr.12: 110381864-110381864 on Build GRCh38
rs749774380 Chr.12: 110381829-110381829 on Build GRCh38
rs35699984 Chr.12: 110381955-110381955 on Build GRCh38
rs377365901 Chr.12: 110381953-110381954 on Build GRCh38
rs774922917 Chr.12: 110381855-110381855 on Build GRCh38
rs752884167 Chr.12: 110381957-110381957 on Build GRCh38
rs77132224 Chr.12: 110381958-110381958 on Build GRCh38
rs753753946 Chr.12: 110381746-110381746 on Build GRCh38
rs546965741 Chr.12: 110382087-110382087 on Build GRCh38
rs751929988 Chr.12: 110381963-110381964 on Build GRCh38
rs972481587 Chr.12: 110381756-110381756 on Build GRCh38
rs796776917 Chr.12: 110381952-110381953 on Build GRCh38
rs954976927 Chr.12: 110381858-110381858 on Build GRCh38
rs748871984 Chr.12: 110381970-110381971 on Build GRCh38
rs375151847 Chr.12: 110381872-110381872 on Build GRCh38
rs148111109 Chr.12: 110381870-110381870 on Build GRCh38
rs1041795593 Chr.12: 110381997-110381997 on Build GRCh38
rs79387430 Chr.12: 110381969-110381973 on Build GRCh38
rs373516411 Chr.12: 110381712-110381712 on Build GRCh38
rs768346328 Chr.12: 110381970-110381970 on Build GRCh38
rs143938358 Chr.12: 110381924-110381924 on Build GRCh38
rs755248162 Chr.12: 110381779-110381779 on Build GRCh38
rs368825963 Chr.12: 110381726-110381726 on Build GRCh38
rs74361481 Chr.12: 110381972-110381972 on Build GRCh38
rs773293057 Chr.12: 110381823-110381823 on Build GRCh38
rs759966133 Chr.12: 110381857-110381857 on Build GRCh38
rs746278576 Chr.12: 110381964-110381964 on Build GRCh38
rs369373794 Chr.12: 110381939-110381939 on Build GRCh38
rs750264243 Chr.12: 110381953-110381953 on Build GRCh38
rs771499699 Chr.12: 110381832-110381832 on Build GRCh38
rs763538022 Chr.12: 110381875-110381875 on Build GRCh38
rs776059081 Chr.12: 110381971-110381971 on Build GRCh38
rs143660720 Chr.12: 110381785-110381785 on Build GRCh38
rs764442142 Chr.12: 110381955-110381956 on Build GRCh38

View Full Product Details