Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113374567-113375075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLBD2
Entrez Gene ID
196463
Gene Name
phospholipase B domain containing 2
Gene Aliases
P76
UniGene
Hs.115896
Gene Chromosome Location
Chr.12: 113358566-113391582 on Build GRCh38
Transcripts
NM_001159727.1 NM_173542.3 XM_011538023.2 XM_017018977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6135744 c.757G>T p.G253C Chr.12: 113374905-113374905 on Build GRCh38
4503558 c.642C>T p.F214F Chr.12: 113374572-113374572 on Build GRCh38
4506048 c.709C>T p.P237S Chr.12: 113374857-113374857 on Build GRCh38
935429 c.825G>T p.K275N Chr.12: 113374973-113374973 on Build GRCh38
4038995 c.764G>A p.S255N Chr.12: 113374912-113374912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976484841 Chr.12: 113374989-113374989 on Build GRCh38
rs987287869 Chr.12: 113375047-113375047 on Build GRCh38
rs779016756 Chr.12: 113374778-113374778 on Build GRCh38
rs772778854 Chr.12: 113374593-113374594 on Build GRCh38
rs535571554 Chr.12: 113374688-113374688 on Build GRCh38
rs200947644 Chr.12: 113374614-113374614 on Build GRCh38
rs767953567 Chr.12: 113374747-113374747 on Build GRCh38
rs762419536 Chr.12: 113375033-113375033 on Build GRCh38
rs769014843 Chr.12: 113375024-113375024 on Build GRCh38
rs765819114 Chr.12: 113374991-113374991 on Build GRCh38
rs758215940 Chr.12: 113374782-113374782 on Build GRCh38
rs963239353 Chr.12: 113374631-113374631 on Build GRCh38
rs896807236 Chr.12: 113374593-113374593 on Build GRCh38
rs777315928 Chr.12: 113374768-113374768 on Build GRCh38
rs754312977 Chr.12: 113374886-113374886 on Build GRCh38
rs769587953 Chr.12: 113374992-113374992 on Build GRCh38
rs762735757 Chr.12: 113374848-113374848 on Build GRCh38
rs540131201 Chr.12: 113374873-113374873 on Build GRCh38
rs781017550 Chr.12: 113374962-113374962 on Build GRCh38
rs748883660 Chr.12: 113374797-113374797 on Build GRCh38
rs770686211 Chr.12: 113374861-113374861 on Build GRCh38
rs141234639 Chr.12: 113374963-113374963 on Build GRCh38
rs929659252 Chr.12: 113374613-113374613 on Build GRCh38
rs745884386 Chr.12: 113374911-113374911 on Build GRCh38
rs921745458 Chr.12: 113374662-113374662 on Build GRCh38
rs769302794 Chr.12: 113374843-113374843 on Build GRCh38
rs748795079 Chr.12: 113375007-113375007 on Build GRCh38
rs774078309 Chr.12: 113375021-113375021 on Build GRCh38
rs528859360 Chr.12: 113375032-113375032 on Build GRCh38
rs575100483 Chr.12: 113374752-113374752 on Build GRCh38
rs746780787 Chr.12: 113374803-113374803 on Build GRCh38
rs145564746 Chr.12: 113374841-113374841 on Build GRCh38
rs761114976 Chr.12: 113374878-113374878 on Build GRCh38
rs760002561 Chr.12: 113374743-113374743 on Build GRCh38
rs762456896 Chr.12: 113374858-113374860 on Build GRCh38
rs1044665386 Chr.12: 113375038-113375038 on Build GRCh38
rs139554738 Chr.12: 113374909-113374909 on Build GRCh38
rs758665072 Chr.12: 113374917-113374917 on Build GRCh38
rs375319838 Chr.12: 113375041-113375041 on Build GRCh38
rs149301819 Chr.12: 113375046-113375046 on Build GRCh38
rs372296081 Chr.12: 113374922-113374922 on Build GRCh38
rs761817689 Chr.12: 113374895-113374895 on Build GRCh38
rs868596404 Chr.12: 113374857-113374857 on Build GRCh38
rs765316447 Chr.12: 113374897-113374897 on Build GRCh38
rs1014511961 Chr.12: 113375022-113375023 on Build GRCh38
rs556997160 Chr.12: 113374706-113374706 on Build GRCh38
rs1035857446 Chr.12: 113374879-113374879 on Build GRCh38
rs761384034 Chr.12: 113374587-113374587 on Build GRCh38
rs754748249 Chr.12: 113374949-113374949 on Build GRCh38
rs973709019 Chr.12: 113375034-113375034 on Build GRCh38
rs564114452 Chr.12: 113374807-113374807 on Build GRCh38
rs1047946588 Chr.12: 113374632-113374632 on Build GRCh38
rs777031868 Chr.12: 113375029-113375029 on Build GRCh38
rs373691519 Chr.12: 113374759-113374759 on Build GRCh38
rs1010673096 Chr.12: 113374862-113374862 on Build GRCh38
rs368747510 Chr.12: 113374772-113374772 on Build GRCh38
rs969049751 Chr.12: 113374885-113374885 on Build GRCh38
rs751363091 Chr.12: 113374936-113374936 on Build GRCh38
rs200426485 Chr.12: 113374760-113374760 on Build GRCh38
rs760960784 Chr.12: 113375022-113375022 on Build GRCh38
rs750557727 Chr.12: 113375043-113375043 on Build GRCh38
rs935375182 Chr.12: 113375050-113375050 on Build GRCh38
rs746038065 Chr.12: 113374779-113374779 on Build GRCh38
rs770991605 Chr.12: 113374869-113374869 on Build GRCh38
rs777476005 Chr.12: 113374993-113374993 on Build GRCh38
rs764512805 Chr.12: 113374880-113374880 on Build GRCh38
rs772866260 Chr.12: 113374742-113374742 on Build GRCh38
rs777982412 Chr.12: 113374670-113374670 on Build GRCh38
rs747629350 Chr.12: 113374842-113374842 on Build GRCh38

View Full Product Details