Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114355829-114356279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
72931 c.1123C>T p.R375W Chr.12: 114355966-114355966 on Build GRCh38
3983825 c.1129G>A p.A377T Chr.12: 114355960-114355960 on Build GRCh38
3456500 c.1250C>T p.P417L Chr.12: 114355839-114355839 on Build GRCh38
7281648 c.1178T>C p.L393P Chr.12: 114355911-114355911 on Build GRCh38
6071340 c.1006C>T p.H336Y Chr.12: 114356083-114356083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751588696 Chr.12: 114355889-114355889 on Build GRCh38
rs775667911 Chr.12: 114355964-114355964 on Build GRCh38
rs140945366 Chr.12: 114356007-114356007 on Build GRCh38
rs762204624 Chr.12: 114355865-114355865 on Build GRCh38
rs368472780 Chr.12: 114356114-114356114 on Build GRCh38
rs944423586 Chr.12: 114356079-114356079 on Build GRCh38
rs148977955 Chr.12: 114356069-114356069 on Build GRCh38
rs201105444 Chr.12: 114356118-114356118 on Build GRCh38
rs1010525892 Chr.12: 114356225-114356225 on Build GRCh38
rs763390789 Chr.12: 114355947-114355947 on Build GRCh38
rs753688559 Chr.12: 114355928-114355928 on Build GRCh38
rs201816980 Chr.12: 114356113-114356113 on Build GRCh38
rs372827955 Chr.12: 114356026-114356026 on Build GRCh38
rs778011764 Chr.12: 114356050-114356050 on Build GRCh38
rs745319218 Chr.12: 114355931-114355931 on Build GRCh38
rs867170084 Chr.12: 114356248-114356248 on Build GRCh38
rs188839350 Chr.12: 114355856-114355856 on Build GRCh38
rs567939875 Chr.12: 114356226-114356226 on Build GRCh38
rs750350579 Chr.12: 114356105-114356105 on Build GRCh38
rs374446377 Chr.12: 114356094-114356094 on Build GRCh38
rs764322332 Chr.12: 114356076-114356076 on Build GRCh38
rs755849744 Chr.12: 114355918-114355918 on Build GRCh38
rs267603320 Chr.12: 114355857-114355857 on Build GRCh38
rs765204502 Chr.12: 114356005-114356005 on Build GRCh38
rs143068551 Chr.12: 114355974-114355974 on Build GRCh38
rs773582799 Chr.12: 114355946-114355946 on Build GRCh38
rs546488473 Chr.12: 114356204-114356204 on Build GRCh38
rs863223778 Chr.12: 114355963-114355963 on Build GRCh38
rs745997257 Chr.12: 114356061-114356061 on Build GRCh38
rs2113433 Chr.12: 114356252-114356252 on Build GRCh38
rs774419025 Chr.12: 114355952-114355952 on Build GRCh38
rs966823240 Chr.12: 114356220-114356220 on Build GRCh38
rs774825317 Chr.12: 114356060-114356060 on Build GRCh38
rs766007057 Chr.12: 114356116-114356116 on Build GRCh38
rs181923192 Chr.12: 114356136-114356136 on Build GRCh38
rs771409350 Chr.12: 114355949-114355949 on Build GRCh38
rs757596478 Chr.12: 114356145-114356145 on Build GRCh38
rs918196518 Chr.12: 114356251-114356252 on Build GRCh38
rs907617752 Chr.12: 114356068-114356068 on Build GRCh38
rs752553883 Chr.12: 114355910-114355910 on Build GRCh38
rs530882236 Chr.12: 114355938-114355938 on Build GRCh38
rs149792175 Chr.12: 114356013-114356013 on Build GRCh38
rs760830193 Chr.12: 114356074-114356074 on Build GRCh38
rs566876542 Chr.12: 114356236-114356236 on Build GRCh38
rs146269931 Chr.12: 114355905-114355905 on Build GRCh38
rs377532269 Chr.12: 114355966-114355966 on Build GRCh38
rs763129045 Chr.12: 114355883-114355883 on Build GRCh38
rs765327234 Chr.12: 114356098-114356098 on Build GRCh38
rs759766836 Chr.12: 114355890-114355890 on Build GRCh38
rs759607221 Chr.12: 114355954-114355954 on Build GRCh38
rs200073406 Chr.12: 114355937-114355937 on Build GRCh38
rs770387292 Chr.12: 114355874-114355874 on Build GRCh38
rs763029336 Chr.12: 114355988-114355988 on Build GRCh38
rs779437406 Chr.12: 114356149-114356149 on Build GRCh38
rs907294334 Chr.12: 114356250-114356250 on Build GRCh38
rs145784562 Chr.12: 114356025-114356025 on Build GRCh38
rs763827720 Chr.12: 114355973-114355973 on Build GRCh38
rs114124210 Chr.12: 114355855-114355855 on Build GRCh38
rs771857554 Chr.12: 114355932-114355932 on Build GRCh38
rs139371720 Chr.12: 114355927-114355927 on Build GRCh38
rs757158327 Chr.12: 114355976-114355976 on Build GRCh38
rs375002260 Chr.12: 114356121-114356121 on Build GRCh38
rs776297033 Chr.12: 114356085-114356085 on Build GRCh38
rs79040021 Chr.12: 114355975-114355975 on Build GRCh38
rs377649723 Chr.12: 114355886-114355886 on Build GRCh38
rs893044550 Chr.12: 114356221-114356221 on Build GRCh38
rs749598173 Chr.12: 114356055-114356055 on Build GRCh38
rs771408470 Chr.12: 114356057-114356057 on Build GRCh38
rs772109169 Chr.12: 114356067-114356067 on Build GRCh38
rs370133092 Chr.12: 114355847-114355847 on Build GRCh38
rs371873510 Chr.12: 114356119-114356119 on Build GRCh38
rs142807139 Chr.12: 114356104-114356104 on Build GRCh38
rs931630578 Chr.12: 114356127-114356127 on Build GRCh38
rs773568096 Chr.12: 114355881-114355881 on Build GRCh38
rs1020820745 Chr.12: 114355936-114355936 on Build GRCh38

View Full Product Details