Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114399434-114399944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3456511 c.338G>A p.R113K Chr.12: 114399537-114399537 on Build GRCh38
1359047 c.310A>G p.M104V Chr.12: 114399565-114399565 on Build GRCh38
3456509 c.348C>T p.F116F Chr.12: 114399527-114399527 on Build GRCh38
3456510 c.338G>A p.R113K Chr.12: 114399537-114399537 on Build GRCh38
3456508 c.348C>T p.F116F Chr.12: 114399527-114399527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941817547 Chr.12: 114399629-114399629 on Build GRCh38
rs754267671 Chr.12: 114399525-114399525 on Build GRCh38
rs28730763 Chr.12: 114399566-114399566 on Build GRCh38
rs935134038 Chr.12: 114399850-114399850 on Build GRCh38
rs556671717 Chr.12: 114399519-114399519 on Build GRCh38
rs947417657 Chr.12: 114399855-114399855 on Build GRCh38
rs748009824 Chr.12: 114399568-114399568 on Build GRCh38
rs371344278 Chr.12: 114399659-114399659 on Build GRCh38
rs750683290 Chr.12: 114399652-114399652 on Build GRCh38
rs765162022 Chr.12: 114399658-114399659 on Build GRCh38
rs775593443 Chr.12: 114399603-114399603 on Build GRCh38
rs904718816 Chr.12: 114399843-114399843 on Build GRCh38
rs530906055 Chr.12: 114399877-114399877 on Build GRCh38
rs762717587 Chr.12: 114399881-114399881 on Build GRCh38
rs765618600 Chr.12: 114399651-114399651 on Build GRCh38
rs151249768 Chr.12: 114399521-114399521 on Build GRCh38
rs907509181 Chr.12: 114399496-114399496 on Build GRCh38
rs778885169 Chr.12: 114399610-114399610 on Build GRCh38
rs1043412780 Chr.12: 114399861-114399862 on Build GRCh38
rs537209826 Chr.12: 114399836-114399836 on Build GRCh38
rs892208251 Chr.12: 114399868-114399868 on Build GRCh38
rs893707932 Chr.12: 114399854-114399854 on Build GRCh38
rs768690315 Chr.12: 114399469-114399469 on Build GRCh38
rs371958704 Chr.12: 114399476-114399476 on Build GRCh38
rs77357563 Chr.12: 114399544-114399544 on Build GRCh38
rs747851699 Chr.12: 114399751-114399751 on Build GRCh38
rs758964536 Chr.12: 114399611-114399611 on Build GRCh38
rs1052402821 Chr.12: 114399851-114399851 on Build GRCh38
rs765947194 Chr.12: 114399505-114399505 on Build GRCh38
rs1001662354 Chr.12: 114399718-114399718 on Build GRCh38
rs1015183019 Chr.12: 114399909-114399909 on Build GRCh38
rs980700901 Chr.12: 114399901-114399901 on Build GRCh38
rs750969565 Chr.12: 114399516-114399516 on Build GRCh38
rs749667596 Chr.12: 114399548-114399548 on Build GRCh38
rs749255660 Chr.12: 114399586-114399586 on Build GRCh38
rs767197919 Chr.12: 114399614-114399614 on Build GRCh38
rs79492814 Chr.12: 114399916-114399916 on Build GRCh38
rs773263145 Chr.12: 114399500-114399500 on Build GRCh38
rs367994558 Chr.12: 114399465-114399465 on Build GRCh38
rs772844823 Chr.12: 114399582-114399582 on Build GRCh38
rs541205665 Chr.12: 114399677-114399677 on Build GRCh38
rs770717773 Chr.12: 114399598-114399598 on Build GRCh38
rs757248939 Chr.12: 114399658-114399658 on Build GRCh38
rs376621016 Chr.12: 114399553-114399553 on Build GRCh38
rs763556418 Chr.12: 114399473-114399473 on Build GRCh38
rs756340269 Chr.12: 114399665-114399665 on Build GRCh38
rs199606641 Chr.12: 114399662-114399662 on Build GRCh38
rs890394778 Chr.12: 114399597-114399597 on Build GRCh38
rs535284360 Chr.12: 114399735-114399735 on Build GRCh38
rs552203759 Chr.12: 114399882-114399882 on Build GRCh38
rs905909732 Chr.12: 114399699-114399699 on Build GRCh38
rs530126948 Chr.12: 114399768-114399768 on Build GRCh38
rs769736865 Chr.12: 114399581-114399581 on Build GRCh38
rs367579977 Chr.12: 114399664-114399664 on Build GRCh38
rs776576558 Chr.12: 114399486-114399486 on Build GRCh38
rs147710408 Chr.12: 114399559-114399559 on Build GRCh38
rs775101163 Chr.12: 114399638-114399638 on Build GRCh38
rs891819123 Chr.12: 114399657-114399657 on Build GRCh38
rs515726234 Chr.12: 114399574-114399574 on Build GRCh38
rs141457646 Chr.12: 114399557-114399557 on Build GRCh38
rs919311929 Chr.12: 114399681-114399681 on Build GRCh38
rs773055417 Chr.12: 114399865-114399865 on Build GRCh38
rs548237118 Chr.12: 114399839-114399839 on Build GRCh38
rs943612834 Chr.12: 114399825-114399825 on Build GRCh38
rs760271246 Chr.12: 114399649-114399649 on Build GRCh38
rs1015587405 Chr.12: 114399770-114399770 on Build GRCh38
rs769477336 Chr.12: 114399800-114399800 on Build GRCh38
rs751705347 Chr.12: 114399660-114399660 on Build GRCh38
rs769896137 Chr.12: 114399487-114399487 on Build GRCh38
rs483353129 Chr.12: 114399537-114399537 on Build GRCh38
rs886039131 Chr.12: 114399615-114399615 on Build GRCh38
rs371966470 Chr.12: 114399503-114399503 on Build GRCh38
rs963109490 Chr.12: 114399788-114399788 on Build GRCh38
rs563535187 Chr.12: 114399857-114399857 on Build GRCh38
rs753329048 Chr.12: 114399636-114399637 on Build GRCh38
rs1000282063 Chr.12: 114399844-114399844 on Build GRCh38
rs1032515451 Chr.12: 114399846-114399846 on Build GRCh38
rs773971213 Chr.12: 114399599-114399599 on Build GRCh38
rs555214695 Chr.12: 114399833-114399833 on Build GRCh38
rs747283420 Chr.12: 114399466-114399466 on Build GRCh38
rs1051770459 Chr.12: 114399756-114399756 on Build GRCh38
rs757586102 Chr.12: 114399535-114399535 on Build GRCh38
rs890728210 Chr.12: 114399823-114399823 on Build GRCh38
rs745985270 Chr.12: 114399560-114399560 on Build GRCh38

View Full Product Details