Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132771596-132772089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
UniGene
Hs.507333
Gene Chromosome Location
Chr.12: 132768909-132829131 on Build GRCh38
Transcripts
NM_005895.3 XM_005266164.4 XM_005266167.4 XM_006719736.3 XM_006719737.3 XM_011534793.2 XM_011534794.2

SNP Detail

SNP ID Chromosome Location
rs115225745 Chr.12: 132771762-132771762 on Build GRCh38
rs577032884 Chr.12: 132771892-132771892 on Build GRCh38
rs974729371 Chr.12: 132771707-132771707 on Build GRCh38
rs1008353596 Chr.12: 132771859-132771859 on Build GRCh38
rs1045227038 Chr.12: 132771968-132771968 on Build GRCh38
rs979264882 Chr.12: 132771950-132771950 on Build GRCh38
rs143472641 Chr.12: 132771777-132771777 on Build GRCh38
rs1028056457 Chr.12: 132771637-132771637 on Build GRCh38
rs941701541 Chr.12: 132771988-132771988 on Build GRCh38
rs1044756721 Chr.12: 132771848-132771850 on Build GRCh38
rs758125448 Chr.12: 132771849-132771854 on Build GRCh38
rs980098047 Chr.12: 132771724-132771724 on Build GRCh38
rs559750306 Chr.12: 132771867-132771867 on Build GRCh38
rs932522643 Chr.12: 132771977-132771977 on Build GRCh38
rs928651556 Chr.12: 132771754-132771754 on Build GRCh38
rs1027176314 Chr.12: 132771884-132771884 on Build GRCh38
rs878906937 Chr.12: 132771786-132771786 on Build GRCh38
rs749769840 Chr.12: 132771992-132771992 on Build GRCh38
rs942956454 Chr.12: 132771713-132771713 on Build GRCh38
rs970020011 Chr.12: 132771718-132771718 on Build GRCh38
rs1044022179 Chr.12: 132771814-132771814 on Build GRCh38
rs781769153 Chr.12: 132771927-132771927 on Build GRCh38
rs564989424 Chr.12: 132771858-132771863 on Build GRCh38
rs141023897 Chr.12: 132771886-132771886 on Build GRCh38
rs920214188 Chr.12: 132771717-132771717 on Build GRCh38
rs577792736 Chr.12: 132771691-132771691 on Build GRCh38
rs993724035 Chr.12: 132772045-132772045 on Build GRCh38
rs12369059 Chr.12: 132771913-132771913 on Build GRCh38
rs572476880 Chr.12: 132772019-132772019 on Build GRCh38
rs34716325 Chr.12: 132771909-132771910 on Build GRCh38
rs945184255 Chr.12: 132771939-132771939 on Build GRCh38
rs996784161 Chr.12: 132771824-132771824 on Build GRCh38
rs564362477 Chr.12: 132771667-132771667 on Build GRCh38
rs567250520 Chr.12: 132771919-132771919 on Build GRCh38
rs73487155 Chr.12: 132771650-132771650 on Build GRCh38
rs184670346 Chr.12: 132772042-132772042 on Build GRCh38
rs1049396127 Chr.12: 132771826-132771826 on Build GRCh38
rs756739673 Chr.12: 132771622-132771622 on Build GRCh38
rs1053208760 Chr.12: 132771763-132771763 on Build GRCh38
rs955553049 Chr.12: 132771685-132771685 on Build GRCh38
rs781774572 Chr.12: 132771851-132771853 on Build GRCh38
rs536301952 Chr.12: 132771926-132771926 on Build GRCh38
rs558599203 Chr.12: 132771887-132771887 on Build GRCh38
rs949138642 Chr.12: 132771947-132771947 on Build GRCh38
rs549118514 Chr.12: 132771846-132771846 on Build GRCh38
rs545350864 Chr.12: 132771839-132771839 on Build GRCh38
rs961864309 Chr.12: 132771615-132771615 on Build GRCh38
rs1037577946 Chr.12: 132772008-132772008 on Build GRCh38
rs995721086 Chr.12: 132771882-132771882 on Build GRCh38
rs63011561 Chr.12: 132771848-132771853 on Build GRCh38
rs528419917 Chr.12: 132771885-132771885 on Build GRCh38
rs537236161 Chr.12: 132771912-132771912 on Build GRCh38
rs897623013 Chr.12: 132771881-132771881 on Build GRCh38
rs986853718 Chr.12: 132771702-132771702 on Build GRCh38
rs956857723 Chr.12: 132771925-132771925 on Build GRCh38
rs1007991669 Chr.12: 132771668-132771668 on Build GRCh38
rs1016143993 Chr.12: 132771675-132771675 on Build GRCh38
rs925121387 Chr.12: 132771957-132771957 on Build GRCh38
rs148009650 Chr.12: 132771791-132771791 on Build GRCh38
rs879071883 Chr.12: 132771866-132771866 on Build GRCh38
rs917637741 Chr.12: 132771944-132771944 on Build GRCh38
rs201875283 Chr.12: 132771881-132771881 on Build GRCh38
rs943754515 Chr.12: 132772034-132772034 on Build GRCh38
rs572412377 Chr.12: 132771621-132771621 on Build GRCh38
rs11147071 Chr.12: 132771987-132771987 on Build GRCh38
rs903794782 Chr.12: 132771823-132771823 on Build GRCh38
rs780542796 Chr.12: 132771851-132771856 on Build GRCh38
rs961570892 Chr.12: 132771695-132771695 on Build GRCh38
rs925414087 Chr.12: 132771720-132771720 on Build GRCh38
rs374841983 Chr.12: 132771723-132771724 on Build GRCh38
rs1006063634 Chr.12: 132771880-132771881 on Build GRCh38
rs747908907 Chr.12: 132771778-132771778 on Build GRCh38

View Full Product Details