Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132926835-132927311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF605
Entrez Gene ID
100289635
Gene Name
zinc finger protein 605
Gene Aliases
-
UniGene
Hs.29698
Gene Chromosome Location
Chr.12: 132921433-132956306 on Build GRCh38
Transcripts
NM_001164715.1 NM_183238.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
165882 c.364G>C p.E122Q Chr.12: 132926935-132926935 on Build GRCh38
328509 c.244A>T p.S82C Chr.12: 132927055-132927055 on Build GRCh38
5632299 c.445A>G p.R149G Chr.12: 132926854-132926854 on Build GRCh38
73462 c.230A>G p.K77R Chr.12: 132927069-132927069 on Build GRCh38
6418385 c.149A>T p.D50V Chr.12: 132927150-132927150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74797472 Chr.12: 132927173-132927173 on Build GRCh38
rs976696030 Chr.12: 132927218-132927218 on Build GRCh38
rs1030045627 Chr.12: 132927071-132927071 on Build GRCh38
rs376743657 Chr.12: 132927180-132927180 on Build GRCh38
rs1002624137 Chr.12: 132926970-132926970 on Build GRCh38
rs1021690825 Chr.12: 132927027-132927027 on Build GRCh38
rs907038191 Chr.12: 132926875-132926875 on Build GRCh38
rs973659429 Chr.12: 132927160-132927160 on Build GRCh38
rs948045456 Chr.12: 132927188-132927188 on Build GRCh38
rs989831110 Chr.12: 132927248-132927248 on Build GRCh38
rs112402378 Chr.12: 132927039-132927039 on Build GRCh38
rs200907965 Chr.12: 132927028-132927028 on Build GRCh38
rs983615151 Chr.12: 132927105-132927105 on Build GRCh38
rs891705591 Chr.12: 132927011-132927011 on Build GRCh38
rs922085272 Chr.12: 132927219-132927219 on Build GRCh38
rs914258961 Chr.12: 132927249-132927249 on Build GRCh38
rs942242536 Chr.12: 132927139-132927139 on Build GRCh38
rs1015213170 Chr.12: 132927079-132927079 on Build GRCh38
rs952000769 Chr.12: 132927013-132927013 on Build GRCh38
rs916582430 Chr.12: 132927178-132927178 on Build GRCh38
rs1046921304 Chr.12: 132927240-132927240 on Build GRCh38
rs7968834 Chr.12: 132927241-132927241 on Build GRCh38
rs983431567 Chr.12: 132927041-132927041 on Build GRCh38
rs1031590018 Chr.12: 132926999-132926999 on Build GRCh38
rs1011224068 Chr.12: 132927024-132927024 on Build GRCh38
rs116985064 Chr.12: 132927084-132927084 on Build GRCh38
rs749278517 Chr.12: 132927030-132927030 on Build GRCh38
rs1046448497 Chr.12: 132926869-132926869 on Build GRCh38
rs908155529 Chr.12: 132927130-132927130 on Build GRCh38
rs966312272 Chr.12: 132927083-132927083 on Build GRCh38

View Full Product Details