Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21177191-21177689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL57
Entrez Gene ID
78988
Gene Name
mitochondrial ribosomal protein L57
Gene Aliases
MRP63, bMRP63
UniGene
Hs.458367
Gene Chromosome Location
Chr.13: 21176233-21179084 on Build GRCh38
Transcripts
NM_024026.4 XM_017020740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
945963 c.297G>T p.K99N Chr.13: 21177213-21177213 on Build GRCh38
223623 c.305C>A p.S102Y Chr.13: 21177221-21177221 on Build GRCh38
6342287 c.289G>T p.V97F Chr.13: 21177205-21177205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763467293 Chr.13: 21177227-21177227 on Build GRCh38
rs907304031 Chr.13: 21177319-21177319 on Build GRCh38
rs758258811 Chr.13: 21177272-21177272 on Build GRCh38
rs556134045 Chr.13: 21177396-21177396 on Build GRCh38
rs778414567 Chr.13: 21177246-21177247 on Build GRCh38
rs1052693591 Chr.13: 21177269-21177269 on Build GRCh38
rs1055801993 Chr.13: 21177586-21177586 on Build GRCh38
rs869301112 Chr.13: 21177395-21177396 on Build GRCh38
rs757093257 Chr.13: 21177277-21177277 on Build GRCh38
rs879069039 Chr.13: 21177384-21177384 on Build GRCh38
rs1001108435 Chr.13: 21177333-21177333 on Build GRCh38
rs1008826789 Chr.13: 21177419-21177419 on Build GRCh38
rs1035072363 Chr.13: 21177305-21177305 on Build GRCh38
rs397709559 Chr.13: 21177394-21177395 on Build GRCh38
rs954183634 Chr.13: 21177373-21177373 on Build GRCh38
rs200909331 Chr.13: 21177395-21177395 on Build GRCh38
rs146280426 Chr.13: 21177221-21177221 on Build GRCh38
rs767654661 Chr.13: 21177240-21177240 on Build GRCh38
rs1001517484 Chr.13: 21177331-21177331 on Build GRCh38
rs748929326 Chr.13: 21177516-21177516 on Build GRCh38
rs373340879 Chr.13: 21177281-21177281 on Build GRCh38
rs775185609 Chr.13: 21177222-21177222 on Build GRCh38
rs774502708 Chr.13: 21177366-21177366 on Build GRCh38
rs138373378 Chr.13: 21177644-21177644 on Build GRCh38
rs372686263 Chr.13: 21177239-21177239 on Build GRCh38
rs537800312 Chr.13: 21177356-21177356 on Build GRCh38
rs11553933 Chr.13: 21177418-21177418 on Build GRCh38
rs1020948748 Chr.13: 21177320-21177320 on Build GRCh38
rs903831000 Chr.13: 21177645-21177645 on Build GRCh38
rs113463350 Chr.13: 21177389-21177389 on Build GRCh38
rs754048983 Chr.13: 21177259-21177259 on Build GRCh38
rs896578271 Chr.13: 21177307-21177307 on Build GRCh38
rs975038476 Chr.13: 21177470-21177470 on Build GRCh38
rs543261467 Chr.13: 21177593-21177593 on Build GRCh38
rs182874238 Chr.13: 21177546-21177546 on Build GRCh38
rs188787111 Chr.13: 21177606-21177606 on Build GRCh38
rs11436868 Chr.13: 21177389-21177390 on Build GRCh38
rs926538488 Chr.13: 21177545-21177545 on Build GRCh38
rs963789990 Chr.13: 21177440-21177440 on Build GRCh38
rs968484587 Chr.13: 21177328-21177328 on Build GRCh38
rs879093229 Chr.13: 21177311-21177312 on Build GRCh38
rs937443394 Chr.13: 21177570-21177570 on Build GRCh38
rs760781755 Chr.13: 21177254-21177254 on Build GRCh38
rs965765240 Chr.13: 21177429-21177429 on Build GRCh38
rs1009941681 Chr.13: 21177311-21177311 on Build GRCh38
rs949222537 Chr.13: 21177542-21177542 on Build GRCh38
rs972258126 Chr.13: 21177436-21177436 on Build GRCh38
rs930327844 Chr.13: 21177534-21177534 on Build GRCh38
rs924987300 Chr.13: 21177493-21177493 on Build GRCh38
rs928690654 Chr.13: 21177548-21177548 on Build GRCh38
rs1008334333 Chr.13: 21177627-21177627 on Build GRCh38
rs898457953 Chr.13: 21177590-21177590 on Build GRCh38
rs766563042 Chr.13: 21177256-21177256 on Build GRCh38
rs1047283042 Chr.13: 21177604-21177604 on Build GRCh38
rs898735433 Chr.13: 21177383-21177383 on Build GRCh38
rs937791999 Chr.13: 21177564-21177564 on Build GRCh38
rs763811528 Chr.13: 21177275-21177275 on Build GRCh38
rs575939527 Chr.13: 21177280-21177280 on Build GRCh38
rs545465535 Chr.13: 21177504-21177504 on Build GRCh38
rs867142107 Chr.13: 21177639-21177639 on Build GRCh38
rs560407456 Chr.13: 21177544-21177544 on Build GRCh38
rs1026320070 Chr.13: 21177408-21177409 on Build GRCh38
rs750426913 Chr.13: 21177251-21177251 on Build GRCh38
rs373458505 Chr.13: 21177224-21177224 on Build GRCh38
rs769136461 Chr.13: 21177235-21177235 on Build GRCh38
rs532048590 Chr.13: 21177617-21177617 on Build GRCh38

View Full Product Details