Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15065768-15066296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
UniGene
Hs.368823
Gene Chromosome Location
Chr.1: 13893387-15118048 on Build GRCh38
Transcripts
NM_001017999.2 NM_001018000.3 NM_001018001.2 NM_015209.2 NM_201628.2 XM_005245795.4 XM_011541074.2 XM_011541075.2 XM_011541076.2 XM_011541077.2 XM_011541080.2 XM_011541081.2 XM_017000770.1 XM_017000771.1 XM_017000772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7300614 c.1238C>T p.A413V Chr.1: 15065787-15065787 on Build GRCh38
3976163 c.1247G>A p.*416* Chr.1: 15065796-15065796 on Build GRCh38
1334430 c.1239G>A p.A413A Chr.1: 15065788-15065788 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756106158 Chr.1: 15065868-15065881 on Build GRCh38
rs757329504 Chr.1: 15065799-15065799 on Build GRCh38
rs536345798 Chr.1: 15065830-15065830 on Build GRCh38
rs944227662 Chr.1: 15065878-15065878 on Build GRCh38
rs1042041469 Chr.1: 15065881-15065881 on Build GRCh38
rs747792165 Chr.1: 15065809-15065809 on Build GRCh38
rs376090885 Chr.1: 15065804-15065804 on Build GRCh38
rs1013755308 Chr.1: 15066163-15066163 on Build GRCh38
rs891302744 Chr.1: 15065860-15065860 on Build GRCh38
rs959686152 Chr.1: 15066134-15066134 on Build GRCh38
rs1038800023 Chr.1: 15065990-15065990 on Build GRCh38
rs1032051233 Chr.1: 15066260-15066260 on Build GRCh38
rs778602508 Chr.1: 15065808-15065808 on Build GRCh38
rs936382512 Chr.1: 15065885-15065885 on Build GRCh38
rs866363732 Chr.1: 15066108-15066108 on Build GRCh38
rs962730808 Chr.1: 15065813-15065813 on Build GRCh38
rs367563649 Chr.1: 15065801-15065801 on Build GRCh38
rs754627148 Chr.1: 15065806-15065806 on Build GRCh38
rs1019950086 Chr.1: 15065919-15065919 on Build GRCh38
rs562595747 Chr.1: 15066036-15066036 on Build GRCh38
rs906986947 Chr.1: 15066137-15066137 on Build GRCh38
rs997882582 Chr.1: 15065903-15065903 on Build GRCh38
rs182127618 Chr.1: 15066242-15066242 on Build GRCh38
rs1038898756 Chr.1: 15065886-15065886 on Build GRCh38
rs72639863 Chr.1: 15066122-15066122 on Build GRCh38
rs572251640 Chr.1: 15065940-15065940 on Build GRCh38
rs755223495 Chr.1: 15066022-15066022 on Build GRCh38
rs192749707 Chr.1: 15065979-15065979 on Build GRCh38
rs892775441 Chr.1: 15065908-15065908 on Build GRCh38
rs962783367 Chr.1: 15065942-15065942 on Build GRCh38
rs774629788 Chr.1: 15065838-15065838 on Build GRCh38
rs375134053 Chr.1: 15065848-15065848 on Build GRCh38
rs1025539436 Chr.1: 15066237-15066237 on Build GRCh38
rs766645238 Chr.1: 15065902-15065902 on Build GRCh38
rs1040299305 Chr.1: 15066065-15066065 on Build GRCh38
rs550173796 Chr.1: 15065871-15065871 on Build GRCh38
rs959562553 Chr.1: 15065963-15065963 on Build GRCh38
rs558715713 Chr.1: 15065895-15065895 on Build GRCh38
rs570254415 Chr.1: 15065874-15065874 on Build GRCh38
rs866064449 Chr.1: 15065822-15065822 on Build GRCh38
rs538813923 Chr.1: 15065894-15065894 on Build GRCh38
rs1041356384 Chr.1: 15066135-15066135 on Build GRCh38
rs758448514 Chr.1: 15066023-15066023 on Build GRCh38
rs115852972 Chr.1: 15066102-15066102 on Build GRCh38
rs936932126 Chr.1: 15066099-15066099 on Build GRCh38
rs547465441 Chr.1: 15065812-15065812 on Build GRCh38
rs867183071 Chr.1: 15065909-15065909 on Build GRCh38
rs775786630 Chr.1: 15065829-15065829 on Build GRCh38
rs761794044 Chr.1: 15065800-15065800 on Build GRCh38
rs959917242 Chr.1: 15065847-15065847 on Build GRCh38
rs369525121 Chr.1: 15066107-15066107 on Build GRCh38
rs1033830309 Chr.1: 15065961-15065961 on Build GRCh38
rs1037024903 Chr.1: 15066062-15066062 on Build GRCh38
rs936434861 Chr.1: 15065986-15065986 on Build GRCh38
rs542928296 Chr.1: 15065989-15065989 on Build GRCh38
rs764482717 Chr.1: 15065832-15065832 on Build GRCh38
rs919930109 Chr.1: 15066016-15066016 on Build GRCh38
rs560883514 Chr.1: 15066149-15066149 on Build GRCh38
rs371846632 Chr.1: 15065802-15065802 on Build GRCh38
rs886378662 Chr.1: 15066040-15066040 on Build GRCh38
rs974232798 Chr.1: 15065818-15065818 on Build GRCh38
rs774068297 Chr.1: 15065798-15065799 on Build GRCh38
rs866948701 Chr.1: 15065985-15065985 on Build GRCh38
rs544186971 Chr.1: 15065864-15065864 on Build GRCh38
rs992852516 Chr.1: 15065966-15065966 on Build GRCh38
rs754179637 Chr.1: 15065824-15065824 on Build GRCh38
rs1002604422 Chr.1: 15066156-15066156 on Build GRCh38
rs544917749 Chr.1: 15066067-15066067 on Build GRCh38
rs79773103 Chr.1: 15066055-15066055 on Build GRCh38
rs779925271 Chr.1: 15065803-15065803 on Build GRCh38
rs772652550 Chr.1: 15065823-15065823 on Build GRCh38
rs918182054 Chr.1: 15065969-15065969 on Build GRCh38
rs886322232 Chr.1: 15065937-15065937 on Build GRCh38
rs12057431 Chr.1: 15065954-15065954 on Build GRCh38
rs187192536 Chr.1: 15066254-15066254 on Build GRCh38
rs762045780 Chr.1: 15065882-15065882 on Build GRCh38
rs7416987 Chr.1: 15065795-15065795 on Build GRCh38
rs921794951 Chr.1: 15066015-15066015 on Build GRCh38
rs967324403 Chr.1: 15065974-15065974 on Build GRCh38
rs372890715 Chr.1: 15065840-15065840 on Build GRCh38
rs899974963 Chr.1: 15066083-15066083 on Build GRCh38
rs767575064 Chr.1: 15065844-15065844 on Build GRCh38
rs917611636 Chr.1: 15065863-15065863 on Build GRCh38

View Full Product Details