Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25247911-25248416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
-
UniGene
Hs.643702
Gene Chromosome Location
Chr.13: 25246201-25287566 on Build GRCh38
Transcripts
NM_004685.3 XM_011535306.1 XM_011535307.1 XM_017020845.1 XM_017020846.1

SNP Detail

SNP ID Chromosome Location
rs1056668554 Chr.13: 25248184-25248184 on Build GRCh38
rs975251931 Chr.13: 25248101-25248102 on Build GRCh38
rs951078803 Chr.13: 25248265-25248265 on Build GRCh38
rs905308774 Chr.13: 25248173-25248173 on Build GRCh38
rs11547268 Chr.13: 25248234-25248234 on Build GRCh38
rs576512781 Chr.13: 25248381-25248381 on Build GRCh38
rs566800466 Chr.13: 25248110-25248110 on Build GRCh38
rs1000802420 Chr.13: 25248235-25248235 on Build GRCh38
rs1029257923 Chr.13: 25248157-25248157 on Build GRCh38
rs1052435625 Chr.13: 25248131-25248131 on Build GRCh38
rs557956035 Chr.13: 25248279-25248279 on Build GRCh38
rs59936632 Chr.13: 25248001-25248001 on Build GRCh38
rs773001748 Chr.13: 25248296-25248296 on Build GRCh38
rs370303133 Chr.13: 25248102-25248102 on Build GRCh38
rs984252830 Chr.13: 25248034-25248034 on Build GRCh38
rs963813698 Chr.13: 25248094-25248094 on Build GRCh38
rs371102067 Chr.13: 25247942-25247942 on Build GRCh38
rs547007912 Chr.13: 25248079-25248079 on Build GRCh38
rs927251391 Chr.13: 25248340-25248340 on Build GRCh38
rs1036526035 Chr.13: 25248049-25248049 on Build GRCh38
rs1025760212 Chr.13: 25247983-25247983 on Build GRCh38
rs956779293 Chr.13: 25248270-25248270 on Build GRCh38
rs533683328 Chr.13: 25248053-25248053 on Build GRCh38
rs992474050 Chr.13: 25247966-25247966 on Build GRCh38
rs760296259 Chr.13: 25248033-25248033 on Build GRCh38
rs1002368424 Chr.13: 25248174-25248174 on Build GRCh38
rs975199405 Chr.13: 25248369-25248369 on Build GRCh38
rs1035262322 Chr.13: 25247956-25247956 on Build GRCh38
rs11841133 Chr.13: 25248148-25248148 on Build GRCh38
rs527770158 Chr.13: 25248245-25248245 on Build GRCh38
rs569551754 Chr.13: 25248249-25248249 on Build GRCh38
rs373520663 Chr.13: 25247999-25247999 on Build GRCh38
rs887507738 Chr.13: 25247994-25247994 on Build GRCh38
rs896725451 Chr.13: 25248199-25248199 on Build GRCh38
rs981737059 Chr.13: 25248307-25248307 on Build GRCh38
rs141319100 Chr.13: 25248271-25248271 on Build GRCh38
rs960984697 Chr.13: 25247961-25247961 on Build GRCh38
rs772615642 Chr.13: 25247940-25247941 on Build GRCh38
rs369734265 Chr.13: 25248342-25248342 on Build GRCh38
rs1044163389 Chr.13: 25248168-25248168 on Build GRCh38

View Full Product Details