Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36865459-36865961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD9
Entrez Gene ID
4093
Gene Name
SMAD family member 9
Gene Aliases
MADH6, MADH9, PPH2, SMAD8, SMAD8/9, SMAD8A, SMAD8B
UniGene
Hs.123119
Gene Chromosome Location
Chr.13: 36844831-36920721 on Build GRCh38
Transcripts
NM_001127217.2 NM_005905.5 XM_005266401.3 XM_005266403.3 XM_005266404.3 XM_006719827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725975 c.850C>A p.R284R Chr.13: 36865690-36865690 on Build GRCh38
6487416 c.791T>C p.F264S Chr.13: 36865638-36865638 on Build GRCh38
6487415 c.902T>C p.F301S Chr.13: 36865638-36865638 on Build GRCh38
1587076 c.937C>A p.L313I Chr.13: 36865603-36865603 on Build GRCh38
4386205 c.676C>T p.R226* Chr.13: 36865753-36865753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773335732 Chr.13: 36865763-36865771 on Build GRCh38
rs530113527 Chr.13: 36865702-36865702 on Build GRCh38
rs553369182 Chr.13: 36865690-36865690 on Build GRCh38
rs761760821 Chr.13: 36865507-36865507 on Build GRCh38
rs781563938 Chr.13: 36865590-36865590 on Build GRCh38
rs867598464 Chr.13: 36865800-36865800 on Build GRCh38
rs78249575 Chr.13: 36865752-36865752 on Build GRCh38
rs772939512 Chr.13: 36865807-36865807 on Build GRCh38
rs1041769139 Chr.13: 36865925-36865925 on Build GRCh38
rs561479397 Chr.13: 36865568-36865568 on Build GRCh38
rs748347542 Chr.13: 36865510-36865510 on Build GRCh38
rs1048305319 Chr.13: 36865656-36865656 on Build GRCh38
rs369783485 Chr.13: 36865633-36865633 on Build GRCh38
rs1571321 Chr.13: 36865801-36865801 on Build GRCh38
rs368389693 Chr.13: 36865609-36865609 on Build GRCh38
rs865835991 Chr.13: 36865543-36865543 on Build GRCh38
rs397514716 Chr.13: 36865660-36865660 on Build GRCh38
rs770243441 Chr.13: 36865585-36865599 on Build GRCh38
rs778547978 Chr.13: 36865791-36865791 on Build GRCh38
rs997923220 Chr.13: 36865692-36865692 on Build GRCh38
rs183967134 Chr.13: 36865862-36865862 on Build GRCh38
rs767433709 Chr.13: 36865545-36865545 on Build GRCh38
rs765054879 Chr.13: 36865888-36865888 on Build GRCh38
rs556124511 Chr.13: 36865892-36865892 on Build GRCh38
rs756445047 Chr.13: 36865784-36865784 on Build GRCh38
rs750319797 Chr.13: 36865549-36865549 on Build GRCh38
rs143817493 Chr.13: 36865620-36865620 on Build GRCh38
rs771214148 Chr.13: 36865586-36865586 on Build GRCh38
rs375838150 Chr.13: 36865487-36865487 on Build GRCh38
rs780288048 Chr.13: 36865786-36865786 on Build GRCh38
rs758521561 Chr.13: 36865570-36865570 on Build GRCh38
rs543294024 Chr.13: 36865482-36865482 on Build GRCh38
rs953071539 Chr.13: 36865836-36865836 on Build GRCh38
rs773886870 Chr.13: 36865898-36865898 on Build GRCh38
rs200467374 Chr.13: 36865648-36865648 on Build GRCh38
rs958332711 Chr.13: 36865813-36865813 on Build GRCh38
rs368154262 Chr.13: 36865716-36865716 on Build GRCh38
rs147382793 Chr.13: 36865730-36865730 on Build GRCh38
rs537538829 Chr.13: 36865883-36865883 on Build GRCh38
rs760510133 Chr.13: 36865647-36865647 on Build GRCh38
rs769776934 Chr.13: 36865738-36865738 on Build GRCh38
rs535002085 Chr.13: 36865635-36865635 on Build GRCh38
rs541164249 Chr.13: 36865838-36865838 on Build GRCh38
rs904426312 Chr.13: 36865652-36865652 on Build GRCh38
rs188107175 Chr.13: 36865918-36865918 on Build GRCh38
rs768181413 Chr.13: 36865631-36865631 on Build GRCh38
rs533765434 Chr.13: 36865843-36865843 on Build GRCh38
rs1007109223 Chr.13: 36865682-36865682 on Build GRCh38
rs142675092 Chr.13: 36865712-36865712 on Build GRCh38
rs367602663 Chr.13: 36865678-36865678 on Build GRCh38
rs756156138 Chr.13: 36865553-36865553 on Build GRCh38
rs747703014 Chr.13: 36865793-36865793 on Build GRCh38
rs3748305 Chr.13: 36865827-36865827 on Build GRCh38
rs570566122 Chr.13: 36865876-36865876 on Build GRCh38
rs750738406 Chr.13: 36865761-36865761 on Build GRCh38
rs888380017 Chr.13: 36865676-36865676 on Build GRCh38
rs771849232 Chr.13: 36865796-36865796 on Build GRCh38
rs771904344 Chr.13: 36865710-36865710 on Build GRCh38
rs485033 Chr.13: 36865924-36865924 on Build GRCh38
rs937482600 Chr.13: 36865881-36865881 on Build GRCh38
rs374907507 Chr.13: 36865715-36865715 on Build GRCh38
rs752874017 Chr.13: 36865569-36865569 on Build GRCh38
rs936800200 Chr.13: 36865509-36865509 on Build GRCh38
rs942660065 Chr.13: 36865934-36865934 on Build GRCh38
rs191749029 Chr.13: 36865739-36865739 on Build GRCh38
rs371433324 Chr.13: 36865689-36865689 on Build GRCh38
rs749383984 Chr.13: 36865778-36865778 on Build GRCh38
rs570886521 Chr.13: 36865506-36865506 on Build GRCh38
rs573558375 Chr.13: 36865933-36865933 on Build GRCh38
rs749774669 Chr.13: 36865687-36865687 on Build GRCh38
rs374668463 Chr.13: 36865664-36865664 on Build GRCh38
rs767384586 Chr.13: 36865636-36865636 on Build GRCh38
rs375675149 Chr.13: 36865870-36865870 on Build GRCh38
rs762489997 Chr.13: 36865499-36865499 on Build GRCh38
rs747243790 Chr.13: 36865574-36865574 on Build GRCh38
rs763846770 Chr.13: 36865500-36865500 on Build GRCh38
rs771053634 Chr.13: 36865780-36865781 on Build GRCh38
rs678037 Chr.13: 36865932-36865932 on Build GRCh38
rs781514663 Chr.13: 36865677-36865677 on Build GRCh38
rs529942038 Chr.13: 36865787-36865787 on Build GRCh38
rs138665617 Chr.13: 36865672-36865672 on Build GRCh38
rs777979202 Chr.13: 36865711-36865711 on Build GRCh38
rs761582055 Chr.13: 36865915-36865915 on Build GRCh38
rs775087497 Chr.13: 36865495-36865495 on Build GRCh38
rs750270551 Chr.13: 36865640-36865640 on Build GRCh38
rs991015364 Chr.13: 36865519-36865519 on Build GRCh38
rs565407365 Chr.13: 36865665-36865665 on Build GRCh38
rs770196600 Chr.13: 36865683-36865683 on Build GRCh38
rs758444458 Chr.13: 36865655-36865655 on Build GRCh38
rs371370412 Chr.13: 36865634-36865634 on Build GRCh38
rs780670552 Chr.13: 36865686-36865686 on Build GRCh38
rs746324666 Chr.13: 36865600-36865600 on Build GRCh38

View Full Product Details