Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 73688331-73688834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF12
Entrez Gene ID
11278
Gene Name
Kruppel like factor 12
Gene Aliases
AP-2rep, AP2REP, HSPC122
UniGene
Hs.373857
Gene Chromosome Location
Chr.13: 73686012-74168295 on Build GRCh38
Transcripts
NM_007249.4 XM_005266251.3 XM_011534907.2 XM_011534909.2 XM_011534910.2 XM_011534911.2 XM_011534912.2 XM_017020384.1 XM_017020385.1

SNP Detail

SNP ID Chromosome Location
rs570162825 Chr.13: 73688564-73688564 on Build GRCh38
rs917567347 Chr.13: 73688691-73688691 on Build GRCh38
rs1048281521 Chr.13: 73688395-73688395 on Build GRCh38
rs1006021613 Chr.13: 73688404-73688404 on Build GRCh38
rs983550255 Chr.13: 73688792-73688792 on Build GRCh38
rs976415955 Chr.13: 73688751-73688751 on Build GRCh38
rs895427268 Chr.13: 73688479-73688479 on Build GRCh38
rs551916117 Chr.13: 73688535-73688535 on Build GRCh38
rs1048110614 Chr.13: 73688745-73688745 on Build GRCh38
rs1025112075 Chr.13: 73688513-73688513 on Build GRCh38
rs886323339 Chr.13: 73688780-73688780 on Build GRCh38
rs978863405 Chr.13: 73688593-73688593 on Build GRCh38
rs879272113 Chr.13: 73688617-73688617 on Build GRCh38
rs575605276 Chr.13: 73688771-73688771 on Build GRCh38
rs886272861 Chr.13: 73688401-73688401 on Build GRCh38
rs545183836 Chr.13: 73688504-73688504 on Build GRCh38
rs17090339 Chr.13: 73688537-73688537 on Build GRCh38
rs942019564 Chr.13: 73688813-73688813 on Build GRCh38
rs192884380 Chr.13: 73688803-73688803 on Build GRCh38
rs900559872 Chr.13: 73688449-73688449 on Build GRCh38
rs923335156 Chr.13: 73688364-73688364 on Build GRCh38
rs937737574 Chr.13: 73688648-73688651 on Build GRCh38
rs957018167 Chr.13: 73688627-73688627 on Build GRCh38
rs545657584 Chr.13: 73688456-73688456 on Build GRCh38
rs575542918 Chr.13: 73688402-73688402 on Build GRCh38
rs923104007 Chr.13: 73688755-73688755 on Build GRCh38
rs372805902 Chr.13: 73688661-73688661 on Build GRCh38
rs1016365851 Chr.13: 73688424-73688424 on Build GRCh38
rs1017512790 Chr.13: 73688740-73688740 on Build GRCh38
rs528825708 Chr.13: 73688685-73688685 on Build GRCh38
rs112366475 Chr.13: 73688431-73688431 on Build GRCh38
rs949315731 Chr.13: 73688394-73688394 on Build GRCh38
rs942072201 Chr.13: 73688458-73688458 on Build GRCh38
rs543939917 Chr.13: 73688779-73688779 on Build GRCh38
rs140714886 Chr.13: 73688388-73688388 on Build GRCh38
rs561553584 Chr.13: 73688684-73688684 on Build GRCh38
rs949370757 Chr.13: 73688724-73688724 on Build GRCh38
rs964650381 Chr.13: 73688750-73688750 on Build GRCh38
rs573063675 Chr.13: 73688522-73688522 on Build GRCh38
rs187117987 Chr.13: 73688486-73688486 on Build GRCh38
rs1014158916 Chr.13: 73688503-73688503 on Build GRCh38
rs766616916 Chr.13: 73688465-73688465 on Build GRCh38

View Full Product Details