Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 78615101-78615610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF219
Entrez Gene ID
79596
Gene Name
ring finger protein 219
Gene Aliases
C13orf7
UniGene
Hs.567576
Gene Chromosome Location
Chr.13: 78614286-78659179 on Build GRCh38
Transcripts
NM_024546.3 XM_011535225.1 XM_017020743.1 NR_047001.1

Gene Symbol
RNF219-AS1
Entrez Gene ID
100874222
Gene Name
RNF219 antisense RNA 1
Gene Aliases
POU4F1-AS1
UniGene
Hs.493062
Gene Chromosome Location
Chr.13: 78054855-78617325 on Build GRCh38
Transcripts
NM_024546.3 XM_011535225.1 XM_017020743.1 NR_047001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1950682 c.2162C>T p.S721L Chr.13: 78615599-78615599 on Build GRCh38
3700425 c.2171C>T p.T724I Chr.13: 78615590-78615590 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901436698 Chr.13: 78615445-78615445 on Build GRCh38
rs981130401 Chr.13: 78615284-78615285 on Build GRCh38
rs1032263577 Chr.13: 78615350-78615350 on Build GRCh38
rs969320539 Chr.13: 78615274-78615274 on Build GRCh38
rs183771922 Chr.13: 78615352-78615352 on Build GRCh38
rs760558047 Chr.13: 78615550-78615551 on Build GRCh38
rs998566741 Chr.13: 78615461-78615461 on Build GRCh38
rs983779329 Chr.13: 78615149-78615149 on Build GRCh38
rs559562369 Chr.13: 78615474-78615474 on Build GRCh38
rs562117237 Chr.13: 78615490-78615490 on Build GRCh38
rs185185938 Chr.13: 78615537-78615537 on Build GRCh38
rs35948422 Chr.13: 78615450-78615451 on Build GRCh38
rs188058834 Chr.13: 78615484-78615484 on Build GRCh38
rs930410966 Chr.13: 78615347-78615347 on Build GRCh38
rs971325720 Chr.13: 78615389-78615389 on Build GRCh38
rs528402708 Chr.13: 78615478-78615478 on Build GRCh38
rs888673274 Chr.13: 78615196-78615196 on Build GRCh38
rs752542492 Chr.13: 78615576-78615576 on Build GRCh38
rs746034088 Chr.13: 78615344-78615346 on Build GRCh38
rs1044383 Chr.13: 78615194-78615194 on Build GRCh38
rs892495130 Chr.13: 78615588-78615588 on Build GRCh38
rs575523959 Chr.13: 78615193-78615193 on Build GRCh38
rs369442665 Chr.13: 78615577-78615577 on Build GRCh38
rs1036559773 Chr.13: 78615135-78615135 on Build GRCh38
rs908462988 Chr.13: 78615240-78615240 on Build GRCh38
rs758653436 Chr.13: 78615403-78615403 on Build GRCh38
rs888642036 Chr.13: 78615361-78615361 on Build GRCh38
rs778344899 Chr.13: 78615419-78615419 on Build GRCh38
rs1035994205 Chr.13: 78615327-78615327 on Build GRCh38
rs1009745247 Chr.13: 78615355-78615355 on Build GRCh38
rs141495359 Chr.13: 78615130-78615130 on Build GRCh38
rs749715036 Chr.13: 78615463-78615463 on Build GRCh38
rs574302208 Chr.13: 78615293-78615293 on Build GRCh38
rs1007575481 Chr.13: 78615198-78615198 on Build GRCh38
rs932485247 Chr.13: 78615255-78615255 on Build GRCh38
rs939655333 Chr.13: 78615499-78615499 on Build GRCh38
rs375276729 Chr.13: 78615566-78615566 on Build GRCh38
rs1033880366 Chr.13: 78615426-78615426 on Build GRCh38
rs998202494 Chr.13: 78615221-78615221 on Build GRCh38
rs200767238 Chr.13: 78615243-78615244 on Build GRCh38
rs543550989 Chr.13: 78615320-78615320 on Build GRCh38
rs983806970 Chr.13: 78615483-78615483 on Build GRCh38
rs561787993 Chr.13: 78615503-78615503 on Build GRCh38
rs570598838 Chr.13: 78615509-78615509 on Build GRCh38
rs1022130729 Chr.13: 78615507-78615507 on Build GRCh38
rs41288264 Chr.13: 78615508-78615508 on Build GRCh38
rs533377892 Chr.13: 78615530-78615530 on Build GRCh38
rs113984764 Chr.13: 78615462-78615462 on Build GRCh38
rs915102440 Chr.13: 78615271-78615271 on Build GRCh38
rs1018270032 Chr.13: 78615201-78615201 on Build GRCh38
rs994261782 Chr.13: 78615192-78615192 on Build GRCh38
rs1053725637 Chr.13: 78615583-78615583 on Build GRCh38
rs932513555 Chr.13: 78615527-78615527 on Build GRCh38
rs557083280 Chr.13: 78615330-78615330 on Build GRCh38
rs535737952 Chr.13: 78615241-78615241 on Build GRCh38
rs1044382 Chr.13: 78615579-78615579 on Build GRCh38
rs191543998 Chr.13: 78615243-78615243 on Build GRCh38
rs766525401 Chr.13: 78615533-78615533 on Build GRCh38
rs765271663 Chr.13: 78615567-78615567 on Build GRCh38
rs989289004 Chr.13: 78615270-78615270 on Build GRCh38
rs568673831 Chr.13: 78615366-78615366 on Build GRCh38
rs965799729 Chr.13: 78615205-78615205 on Build GRCh38
rs901031568 Chr.13: 78615251-78615251 on Build GRCh38
rs542510365 Chr.13: 78615248-78615248 on Build GRCh38
rs918975754 Chr.13: 78615328-78615328 on Build GRCh38

View Full Product Details