Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 97275871-97276408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBNL2
Entrez Gene ID
10150
Gene Name
muscleblind like splicing regulator 2
Gene Aliases
MBLL, MBLL39, PRO2032
UniGene
Hs.657347
Gene Chromosome Location
Chr.13: 97141780-97394120 on Build GRCh38
Transcripts
NM_001306070.1 NM_144778.3 NM_207304.2 XM_005254018.2 XM_005254019.2 XM_005254021.2 XM_005254022.2 XM_006719912.2 XM_011521045.2 XM_017020308.1 XM_017020309.1 XM_017020310.1 XM_017020311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306720 c.136G>T p.E46* Chr.13: 97276371-97276371 on Build GRCh38
432640 c.95A>C p.E32A Chr.13: 97276330-97276330 on Build GRCh38
6306719 c.136G>T p.E46* Chr.13: 97276371-97276371 on Build GRCh38
6709149 c.86G>A p.R29H Chr.13: 97276321-97276321 on Build GRCh38
4701002 c.87C>T p.R29R Chr.13: 97276322-97276322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs36010428 Chr.13: 97276361-97276361 on Build GRCh38
rs775745316 Chr.13: 97276232-97276232 on Build GRCh38
rs778441022 Chr.13: 97276333-97276333 on Build GRCh38
rs955746548 Chr.13: 97276320-97276320 on Build GRCh38
rs925657885 Chr.13: 97275936-97275936 on Build GRCh38
rs201084754 Chr.13: 97276368-97276368 on Build GRCh38
rs1000906044 Chr.13: 97275973-97275973 on Build GRCh38
rs768820298 Chr.13: 97276248-97276248 on Build GRCh38
rs969023672 Chr.13: 97275957-97275957 on Build GRCh38
rs926955948 Chr.13: 97276060-97276060 on Build GRCh38
rs1018165191 Chr.13: 97276353-97276353 on Build GRCh38
rs746232646 Chr.13: 97276225-97276225 on Build GRCh38
rs373812610 Chr.13: 97276385-97276385 on Build GRCh38
rs377260726 Chr.13: 97276185-97276185 on Build GRCh38
rs923320829 Chr.13: 97275997-97275997 on Build GRCh38
rs533230832 Chr.13: 97276049-97276049 on Build GRCh38
rs141173117 Chr.13: 97276292-97276292 on Build GRCh38
rs774132167 Chr.13: 97276253-97276253 on Build GRCh38
rs1032764118 Chr.13: 97275974-97275974 on Build GRCh38
rs999824577 Chr.13: 97276249-97276249 on Build GRCh38
rs908157528 Chr.13: 97276017-97276017 on Build GRCh38
rs926666874 Chr.13: 97275980-97275980 on Build GRCh38
rs766126526 Chr.13: 97276280-97276280 on Build GRCh38
rs780215815 Chr.13: 97276366-97276366 on Build GRCh38
rs139684062 Chr.13: 97276257-97276257 on Build GRCh38
rs936985198 Chr.13: 97276082-97276082 on Build GRCh38
rs763367930 Chr.13: 97276054-97276054 on Build GRCh38
rs773382768 Chr.13: 97276039-97276039 on Build GRCh38
rs769975742 Chr.13: 97276025-97276025 on Build GRCh38
rs373511683 Chr.13: 97276187-97276187 on Build GRCh38
rs763012634 Chr.13: 97276247-97276247 on Build GRCh38
rs115576896 Chr.13: 97275977-97275977 on Build GRCh38
rs768580262 Chr.13: 97275947-97275947 on Build GRCh38
rs781670889 Chr.13: 97276354-97276354 on Build GRCh38
rs768781962 Chr.13: 97276383-97276383 on Build GRCh38
rs770208186 Chr.13: 97276364-97276364 on Build GRCh38
rs757720444 Chr.13: 97276346-97276346 on Build GRCh38
rs958196180 Chr.13: 97275953-97275953 on Build GRCh38
rs954885873 Chr.13: 97276006-97276006 on Build GRCh38
rs549833491 Chr.13: 97276175-97276175 on Build GRCh38
rs766968214 Chr.13: 97276139-97276139 on Build GRCh38
rs767385632 Chr.13: 97276256-97276256 on Build GRCh38
rs752021459 Chr.13: 97276157-97276157 on Build GRCh38
rs1048464159 Chr.13: 97276016-97276016 on Build GRCh38
rs377056466 Chr.13: 97276197-97276197 on Build GRCh38
rs41279106 Chr.13: 97276011-97276011 on Build GRCh38
rs777018340 Chr.13: 97276214-97276214 on Build GRCh38
rs769941940 Chr.13: 97276229-97276229 on Build GRCh38
rs754426281 Chr.13: 97276298-97276298 on Build GRCh38
rs761760187 Chr.13: 97276254-97276254 on Build GRCh38
rs188817032 Chr.13: 97276209-97276209 on Build GRCh38
rs1028234687 Chr.13: 97276108-97276108 on Build GRCh38
rs11544959 Chr.13: 97275948-97275948 on Build GRCh38
rs752183016 Chr.13: 97276339-97276339 on Build GRCh38
rs565743334 Chr.13: 97275978-97275978 on Build GRCh38

View Full Product Details