Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 101721142-101721691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
UniGene
Hs.508616
Gene Chromosome Location
Chr.13: 101720855-102402428 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2

SNP Detail

SNP ID Chromosome Location
rs910600089 Chr.13: 101721404-101721404 on Build GRCh38
rs955955746 Chr.13: 101721246-101721246 on Build GRCh38
rs15608 Chr.13: 101721213-101721213 on Build GRCh38
rs776494941 Chr.13: 101721466-101721466 on Build GRCh38
rs569870029 Chr.13: 101721353-101721354 on Build GRCh38
rs566562589 Chr.13: 101721643-101721644 on Build GRCh38
rs993220059 Chr.13: 101721531-101721531 on Build GRCh38
rs565918763 Chr.13: 101721339-101721340 on Build GRCh38
rs1029460709 Chr.13: 101721590-101721591 on Build GRCh38
rs540292614 Chr.13: 101721535-101721535 on Build GRCh38
rs200743928 Chr.13: 101721438-101721438 on Build GRCh38
rs955894446 Chr.13: 101721552-101721552 on Build GRCh38
rs781681565 Chr.13: 101721340-101721343 on Build GRCh38
rs769098923 Chr.13: 101721343-101721344 on Build GRCh38
rs546246953 Chr.13: 101721607-101721608 on Build GRCh38
rs986662467 Chr.13: 101721385-101721386 on Build GRCh38
rs10626847 Chr.13: 101721440-101721441 on Build GRCh38
rs752386428 Chr.13: 101721653-101721658 on Build GRCh38
rs756177413 Chr.13: 101721467-101721468 on Build GRCh38
rs989113921 Chr.13: 101721273-101721273 on Build GRCh38
rs35551918 Chr.13: 101721441-101721442 on Build GRCh38
rs1037983463 Chr.13: 101721567-101721567 on Build GRCh38
rs997040609 Chr.13: 101721634-101721649 on Build GRCh38
rs1022662054 Chr.13: 101721169-101721169 on Build GRCh38
rs372768269 Chr.13: 101721632-101721632 on Build GRCh38
rs1002509589 Chr.13: 101721526-101721526 on Build GRCh38
rs943368863 Chr.13: 101721419-101721419 on Build GRCh38
rs550137543 Chr.13: 101721353-101721353 on Build GRCh38
rs947077421 Chr.13: 101721436-101721436 on Build GRCh38
rs386380488 Chr.13: 101721444-101721445 on Build GRCh38
rs1022675090 Chr.13: 101721504-101721504 on Build GRCh38
rs762640115 Chr.13: 101721174-101721175 on Build GRCh38
rs573219749 Chr.13: 101721310-101721310 on Build GRCh38
rs542077649 Chr.13: 101721428-101721429 on Build GRCh38
rs887986830 Chr.13: 101721493-101721493 on Build GRCh38
rs1006313806 Chr.13: 101721497-101721497 on Build GRCh38
rs886696133 Chr.13: 101721223-101721223 on Build GRCh38
rs749492811 Chr.13: 101721575-101721575 on Build GRCh38
rs531142211 Chr.13: 101721345-101721345 on Build GRCh38
rs914449213 Chr.13: 101721302-101721302 on Build GRCh38
rs142480530 Chr.13: 101721205-101721207 on Build GRCh38
rs771196126 Chr.13: 101721201-101721202 on Build GRCh38
rs1057319033 Chr.13: 101721546-101721546 on Build GRCh38
rs1019908865 Chr.13: 101721393-101721393 on Build GRCh38
rs761796539 Chr.13: 101721520-101721520 on Build GRCh38
rs766695039 Chr.13: 101721195-101721196 on Build GRCh38
rs141506954 Chr.13: 101721384-101721385 on Build GRCh38
rs1029120398 Chr.13: 101721350-101721351 on Build GRCh38
rs1035383926 Chr.13: 101721537-101721537 on Build GRCh38
rs181961434 Chr.13: 101721578-101721578 on Build GRCh38
rs146196463 Chr.13: 101721659-101721662 on Build GRCh38
rs940910701 Chr.13: 101721553-101721553 on Build GRCh38
rs535930764 Chr.13: 101721572-101721572 on Build GRCh38
rs1021535006 Chr.13: 101721640-101721640 on Build GRCh38
rs985267509 Chr.13: 101721608-101721608 on Build GRCh38
rs979799753 Chr.13: 101721658-101721659 on Build GRCh38
rs1043839520 Chr.13: 101721384-101721384 on Build GRCh38
rs1046197 Chr.13: 101721219-101721219 on Build GRCh38
rs371959735 Chr.13: 101721429-101721429 on Build GRCh38
rs947198015 Chr.13: 101721305-101721305 on Build GRCh38
rs1040368163 Chr.13: 101721423-101721423 on Build GRCh38
rs75757057 Chr.13: 101721440-101721440 on Build GRCh38
rs570879019 Chr.13: 101721282-101721283 on Build GRCh38
rs1019454348 Chr.13: 101721599-101721599 on Build GRCh38
rs938815005 Chr.13: 101721503-101721503 on Build GRCh38
rs906007712 Chr.13: 101721216-101721216 on Build GRCh38
rs910652307 Chr.13: 101721631-101721631 on Build GRCh38
rs386773958 Chr.13: 101721213-101721219 on Build GRCh38
rs775124846 Chr.13: 101721177-101721177 on Build GRCh38
rs751381998 Chr.13: 101721631-101721633 on Build GRCh38
rs988776034 Chr.13: 101721557-101721557 on Build GRCh38
rs993788846 Chr.13: 101721449-101721449 on Build GRCh38
rs996245960 Chr.13: 101721344-101721345 on Build GRCh38
rs1008451714 Chr.13: 101721598-101721598 on Build GRCh38
rs914298159 Chr.13: 101721181-101721181 on Build GRCh38
rs890545273 Chr.13: 101721591-101721592 on Build GRCh38
rs547146646 Chr.13: 101721495-101721496 on Build GRCh38
rs759620996 Chr.13: 101721219-101721222 on Build GRCh38
rs747506157 Chr.13: 101721222-101721223 on Build GRCh38

View Full Product Details