Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 101721575-101722086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
UniGene
Hs.508616
Gene Chromosome Location
Chr.13: 101720855-102402428 on Build GRCh38
Transcripts
NM_001321931.1 NM_001321932.1 NM_001321933.1 NM_001321934.1 NM_001321935.1 NM_001321936.1 NM_001321938.1 NM_001321939.1 NM_001321940.1 NM_001321941.1 NM_001321942.1 NM_001321943.1 NM_001321944.1 NM_001321945.1 NM_001321946.1 NM_001321947.1 NM_001321948.1 NM_001321949.1 NM_004115.3 NM_175929.2

SNP Detail

SNP ID Chromosome Location
rs886049935 Chr.13: 101722048-101722048 on Build GRCh38
rs557319654 Chr.13: 101721858-101721859 on Build GRCh38
rs968609249 Chr.13: 101721814-101721814 on Build GRCh38
rs370179321 Chr.13: 101721829-101721829 on Build GRCh38
rs1001440479 Chr.13: 101721989-101721989 on Build GRCh38
rs566562589 Chr.13: 101721643-101721644 on Build GRCh38
rs1010276909 Chr.13: 101721935-101721935 on Build GRCh38
rs911880656 Chr.13: 101721895-101721896 on Build GRCh38
rs992586327 Chr.13: 101721850-101721850 on Build GRCh38
rs546246953 Chr.13: 101721607-101721608 on Build GRCh38
rs920825970 Chr.13: 101721881-101721881 on Build GRCh38
rs1009986559 Chr.13: 101721963-101721963 on Build GRCh38
rs767732567 Chr.13: 101721826-101721826 on Build GRCh38
rs544295020 Chr.13: 101721695-101721695 on Build GRCh38
rs869305706 Chr.13: 101721869-101721870 on Build GRCh38
rs556285749 Chr.13: 101721969-101721969 on Build GRCh38
rs752386428 Chr.13: 101721653-101721658 on Build GRCh38
rs1014343422 Chr.13: 101722021-101722021 on Build GRCh38
rs375885231 Chr.13: 101721817-101721817 on Build GRCh38
rs756431012 Chr.13: 101721975-101721975 on Build GRCh38
rs1034717897 Chr.13: 101722012-101722013 on Build GRCh38
rs138068963 Chr.13: 101721714-101721714 on Build GRCh38
rs997040609 Chr.13: 101721634-101721649 on Build GRCh38
rs551683892 Chr.13: 101721737-101721737 on Build GRCh38
rs973540867 Chr.13: 101722041-101722042 on Build GRCh38
rs372768269 Chr.13: 101721632-101721632 on Build GRCh38
rs752896423 Chr.13: 101721838-101721838 on Build GRCh38
rs548711835 Chr.13: 101721769-101721769 on Build GRCh38
rs891628806 Chr.13: 101721770-101721770 on Build GRCh38
rs890509660 Chr.13: 101721758-101721758 on Build GRCh38
rs76977018 Chr.13: 101721859-101721859 on Build GRCh38
rs1010060116 Chr.13: 101721771-101721772 on Build GRCh38
rs764377802 Chr.13: 101722023-101722023 on Build GRCh38
rs1050470728 Chr.13: 101721756-101721756 on Build GRCh38
rs777720618 Chr.13: 101721891-101721892 on Build GRCh38
rs1014641347 Chr.13: 101722025-101722026 on Build GRCh38
rs190937243 Chr.13: 101721974-101721974 on Build GRCh38
rs955738175 Chr.13: 101721929-101721929 on Build GRCh38
rs867925528 Chr.13: 101722036-101722036 on Build GRCh38
rs886049932 Chr.13: 101721759-101721759 on Build GRCh38
rs34628929 Chr.13: 101721859-101721860 on Build GRCh38
rs185008055 Chr.13: 101721846-101721846 on Build GRCh38
rs58555898 Chr.13: 101721768-101721768 on Build GRCh38
rs886049933 Chr.13: 101721760-101721760 on Build GRCh38
rs959724606 Chr.13: 101722019-101722020 on Build GRCh38
rs906028900 Chr.13: 101721796-101721796 on Build GRCh38
rs959829493 Chr.13: 101721677-101721677 on Build GRCh38
rs932236298 Chr.13: 101721888-101721889 on Build GRCh38
rs749730478 Chr.13: 101721669-101721670 on Build GRCh38
rs577253536 Chr.13: 101721768-101721769 on Build GRCh38
rs146196463 Chr.13: 101721659-101721662 on Build GRCh38
rs920852238 Chr.13: 101721734-101721734 on Build GRCh38
rs892559025 Chr.13: 101721933-101721934 on Build GRCh38
rs1021535006 Chr.13: 101721640-101721640 on Build GRCh38
rs985267509 Chr.13: 101721608-101721608 on Build GRCh38
rs979799753 Chr.13: 101721658-101721659 on Build GRCh38
rs768342824 Chr.13: 101721908-101721909 on Build GRCh38
rs761838007 Chr.13: 101722028-101722029 on Build GRCh38
rs973568534 Chr.13: 101721874-101721875 on Build GRCh38
rs1035838081 Chr.13: 101721906-101721906 on Build GRCh38
rs907657431 Chr.13: 101721852-101721852 on Build GRCh38
rs567206934 Chr.13: 101721797-101721797 on Build GRCh38
rs573249049 Chr.13: 101722023-101722024 on Build GRCh38
rs886049934 Chr.13: 101721857-101721857 on Build GRCh38
rs754379658 Chr.13: 101722058-101722058 on Build GRCh38
rs910652307 Chr.13: 101721631-101721631 on Build GRCh38
rs886049936 Chr.13: 101722054-101722054 on Build GRCh38
rs751381998 Chr.13: 101721631-101721633 on Build GRCh38
rs887989135 Chr.13: 101721748-101721748 on Build GRCh38
rs371464074 Chr.13: 101722026-101722029 on Build GRCh38
rs1040464283 Chr.13: 101721709-101721709 on Build GRCh38
rs943461946 Chr.13: 101721678-101721678 on Build GRCh38
rs746765508 Chr.13: 101721886-101721887 on Build GRCh38
rs567073335 Chr.13: 101721747-101721747 on Build GRCh38
rs962168798 Chr.13: 101721857-101721857 on Build GRCh38
rs147404825 Chr.13: 101721713-101721713 on Build GRCh38

View Full Product Details