Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102787698-102788199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDELC1
Entrez Gene ID
79070
Gene Name
KDEL motif containing 1
Gene Aliases
EP58, ERp58, KDEL1
UniGene
Hs.408629
Gene Chromosome Location
Chr.13: 102784281-102799054 on Build GRCh38
Transcripts
NM_001318732.1 NM_024089.2 XM_005254075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692152 c.1375C>A p.L459I Chr.13: 102787842-102787842 on Build GRCh38
4694832 c.1337A>C p.N446T Chr.13: 102787880-102787880 on Build GRCh38
4402859 c.1359C>T p.F453F Chr.13: 102787858-102787858 on Build GRCh38
1211698 c.1346G>A p.G449D Chr.13: 102787871-102787871 on Build GRCh38
1365312 c.1308A>T p.A436A Chr.13: 102787909-102787909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530287692 Chr.13: 102787939-102787939 on Build GRCh38
rs758631015 Chr.13: 102787815-102787815 on Build GRCh38
rs760010033 Chr.13: 102787929-102787929 on Build GRCh38
rs528891829 Chr.13: 102788140-102788140 on Build GRCh38
rs1047876671 Chr.13: 102787980-102787980 on Build GRCh38
rs144543760 Chr.13: 102787942-102787943 on Build GRCh38
rs747119957 Chr.13: 102787843-102787843 on Build GRCh38
rs183974374 Chr.13: 102788074-102788074 on Build GRCh38
rs565936313 Chr.13: 102787928-102787928 on Build GRCh38
rs773815713 Chr.13: 102787890-102787890 on Build GRCh38
rs772533851 Chr.13: 102788141-102788141 on Build GRCh38
rs768975095 Chr.13: 102787869-102787869 on Build GRCh38
rs748953821 Chr.13: 102787886-102787886 on Build GRCh38
rs777008728 Chr.13: 102787926-102787926 on Build GRCh38
rs563139299 Chr.13: 102788042-102788042 on Build GRCh38
rs544052067 Chr.13: 102787994-102787994 on Build GRCh38
rs377423123 Chr.13: 102787870-102787870 on Build GRCh38
rs777785246 Chr.13: 102787830-102787830 on Build GRCh38
rs1025214758 Chr.13: 102788160-102788160 on Build GRCh38
rs945608726 Chr.13: 102788041-102788041 on Build GRCh38
rs528187568 Chr.13: 102787803-102787803 on Build GRCh38
rs996493025 Chr.13: 102788017-102788017 on Build GRCh38
rs1054803122 Chr.13: 102788086-102788087 on Build GRCh38
rs930735065 Chr.13: 102787977-102787977 on Build GRCh38
rs920620487 Chr.13: 102788173-102788173 on Build GRCh38
rs1024909648 Chr.13: 102787785-102787785 on Build GRCh38
rs761334297 Chr.13: 102787900-102787900 on Build GRCh38
rs570893101 Chr.13: 102788073-102788073 on Build GRCh38
rs1013797310 Chr.13: 102787754-102787754 on Build GRCh38
rs1033504605 Chr.13: 102787728-102787728 on Build GRCh38
rs1028422215 Chr.13: 102788027-102788027 on Build GRCh38
rs989659459 Chr.13: 102788043-102788043 on Build GRCh38
rs558624131 Chr.13: 102787876-102787876 on Build GRCh38
rs756349658 Chr.13: 102787794-102787794 on Build GRCh38
rs768358353 Chr.13: 102787889-102787889 on Build GRCh38
rs564193017 Chr.13: 102787725-102787725 on Build GRCh38
rs578200131 Chr.13: 102787737-102787737 on Build GRCh38
rs752908430 Chr.13: 102787807-102787807 on Build GRCh38
rs1040920967 Chr.13: 102788002-102788002 on Build GRCh38
rs1009147991 Chr.13: 102788004-102788004 on Build GRCh38
rs766885139 Chr.13: 102787788-102787788 on Build GRCh38
rs548461846 Chr.13: 102787956-102787956 on Build GRCh38
rs919242585 Chr.13: 102787864-102787864 on Build GRCh38
rs191728466 Chr.13: 102787957-102787957 on Build GRCh38
rs1019271251 Chr.13: 102787735-102787735 on Build GRCh38
rs777993673 Chr.13: 102787798-102787798 on Build GRCh38
rs113796537 Chr.13: 102788012-102788012 on Build GRCh38
rs754294198 Chr.13: 102787789-102787789 on Build GRCh38
rs7321481 Chr.13: 102788147-102788147 on Build GRCh38
rs752368640 Chr.13: 102788133-102788133 on Build GRCh38
rs111390008 Chr.13: 102788136-102788136 on Build GRCh38
rs758625756 Chr.13: 102787940-102787940 on Build GRCh38
rs900761047 Chr.13: 102788013-102788013 on Build GRCh38
rs367935251 Chr.13: 102787973-102787973 on Build GRCh38
rs762524017 Chr.13: 102788158-102788158 on Build GRCh38
rs765602550 Chr.13: 102787806-102787806 on Build GRCh38
rs751367606 Chr.13: 102787793-102787793 on Build GRCh38
rs757359037 Chr.13: 102787868-102787868 on Build GRCh38
rs764393728 Chr.13: 102787953-102787953 on Build GRCh38
rs975149780 Chr.13: 102787749-102787749 on Build GRCh38

View Full Product Details