Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 106492789-106493291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB2
Entrez Gene ID
1948
Gene Name
ephrin B2
Gene Aliases
EPLG5, HTKL, Htk-L, LERK5
UniGene
Hs.149239
Gene Chromosome Location
Chr.13: 106489731-106535040 on Build GRCh38
Transcripts
NM_004093.3 XM_017020406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5039283 c.943G>T p.V315L Chr.13: 106493099-106493099 on Build GRCh38
7149160 c.779C>T p.S260L Chr.13: 106493263-106493263 on Build GRCh38
5052040 c.924C>T p.S308S Chr.13: 106493118-106493118 on Build GRCh38
330168 c.923G>T p.S308I Chr.13: 106493119-106493119 on Build GRCh38
7182345 c.848G>T p.G283V Chr.13: 106493194-106493194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551530122 Chr.13: 106492859-106492859 on Build GRCh38
rs759571591 Chr.13: 106493078-106493078 on Build GRCh38
rs894300596 Chr.13: 106493142-106493142 on Build GRCh38
rs188174925 Chr.13: 106493193-106493193 on Build GRCh38
rs750852379 Chr.13: 106493190-106493190 on Build GRCh38
rs1026640558 Chr.13: 106492907-106492907 on Build GRCh38
rs765521052 Chr.13: 106493251-106493251 on Build GRCh38
rs771862640 Chr.13: 106493236-106493236 on Build GRCh38
rs768156053 Chr.13: 106492908-106492908 on Build GRCh38
rs200969432 Chr.13: 106493061-106493061 on Build GRCh38
rs890963211 Chr.13: 106492861-106492861 on Build GRCh38
rs750286889 Chr.13: 106492993-106492993 on Build GRCh38
rs141519659 Chr.13: 106493259-106493259 on Build GRCh38
rs778078295 Chr.13: 106493032-106493032 on Build GRCh38
rs767538286 Chr.13: 106493156-106493156 on Build GRCh38
rs544759056 Chr.13: 106492944-106492944 on Build GRCh38
rs977007352 Chr.13: 106492809-106492809 on Build GRCh38
rs773001048 Chr.13: 106493133-106493133 on Build GRCh38
rs954164031 Chr.13: 106493239-106493239 on Build GRCh38
rs927359074 Chr.13: 106492949-106492949 on Build GRCh38
rs770833245 Chr.13: 106493147-106493147 on Build GRCh38
rs377711689 Chr.13: 106493213-106493213 on Build GRCh38
rs771536266 Chr.13: 106492991-106492991 on Build GRCh38
rs778447773 Chr.13: 106492930-106492930 on Build GRCh38
rs368022759 Chr.13: 106493130-106493130 on Build GRCh38
rs752849130 Chr.13: 106493026-106493026 on Build GRCh38
rs377256166 Chr.13: 106493265-106493265 on Build GRCh38
rs74444474 Chr.13: 106492947-106492947 on Build GRCh38
rs571385795 Chr.13: 106492892-106492892 on Build GRCh38
rs780547614 Chr.13: 106493127-106493127 on Build GRCh38
rs764771514 Chr.13: 106493196-106493198 on Build GRCh38
rs779399309 Chr.13: 106493041-106493041 on Build GRCh38
rs752669368 Chr.13: 106493166-106493166 on Build GRCh38
rs575805487 Chr.13: 106492893-106492893 on Build GRCh38
rs761345466 Chr.13: 106493065-106493065 on Build GRCh38
rs766350222 Chr.13: 106493075-106493075 on Build GRCh38
rs377602354 Chr.13: 106493022-106493022 on Build GRCh38
rs374006513 Chr.13: 106493253-106493253 on Build GRCh38
rs191693363 Chr.13: 106492962-106492962 on Build GRCh38
rs143343883 Chr.13: 106493100-106493100 on Build GRCh38
rs762774468 Chr.13: 106493145-106493145 on Build GRCh38
rs766438398 Chr.13: 106493000-106493000 on Build GRCh38
rs931101990 Chr.13: 106493103-106493103 on Build GRCh38
rs190855238 Chr.13: 106493245-106493245 on Build GRCh38
rs772248894 Chr.13: 106493109-106493109 on Build GRCh38
rs41275060 Chr.13: 106493262-106493262 on Build GRCh38
rs1039877438 Chr.13: 106492889-106492889 on Build GRCh38
rs537317777 Chr.13: 106492937-106492937 on Build GRCh38
rs114841344 Chr.13: 106492810-106492810 on Build GRCh38
rs370518437 Chr.13: 106493250-106493250 on Build GRCh38
rs775512916 Chr.13: 106493238-106493238 on Build GRCh38
rs958739933 Chr.13: 106493220-106493220 on Build GRCh38
rs7995379 Chr.13: 106493115-106493115 on Build GRCh38
rs755013813 Chr.13: 106493010-106493010 on Build GRCh38
rs758390429 Chr.13: 106492998-106492998 on Build GRCh38
rs903033183 Chr.13: 106492897-106492897 on Build GRCh38
rs751647102 Chr.13: 106493007-106493007 on Build GRCh38
rs115514643 Chr.13: 106492821-106492821 on Build GRCh38
rs776275451 Chr.13: 106493062-106493062 on Build GRCh38
rs888114930 Chr.13: 106493189-106493189 on Build GRCh38
rs770747304 Chr.13: 106493211-106493211 on Build GRCh38
rs751552616 Chr.13: 106493076-106493076 on Build GRCh38
rs759930636 Chr.13: 106493108-106493108 on Build GRCh38
rs764803364 Chr.13: 106493206-106493206 on Build GRCh38
rs553579869 Chr.13: 106493073-106493073 on Build GRCh38
rs374559924 Chr.13: 106493212-106493212 on Build GRCh38
rs150898512 Chr.13: 106493172-106493172 on Build GRCh38
rs71640250 Chr.13: 106493244-106493244 on Build GRCh38
rs370876035 Chr.13: 106493191-106493191 on Build GRCh38
rs771389582 Chr.13: 106493039-106493039 on Build GRCh38
rs113878095 Chr.13: 106493196-106493196 on Build GRCh38
rs558118969 Chr.13: 106492961-106492961 on Build GRCh38
rs952292858 Chr.13: 106492908-106492910 on Build GRCh38
rs945987057 Chr.13: 106492828-106492828 on Build GRCh38
rs530509657 Chr.13: 106493117-106493117 on Build GRCh38
rs756424821 Chr.13: 106493031-106493031 on Build GRCh38
rs374833315 Chr.13: 106492842-106492842 on Build GRCh38
rs757543125 Chr.13: 106493183-106493183 on Build GRCh38
rs755606363 Chr.13: 106493264-106493264 on Build GRCh38
rs1012769087 Chr.13: 106492921-106492921 on Build GRCh38
rs535855285 Chr.13: 106493129-106493129 on Build GRCh38
rs747541545 Chr.13: 106493194-106493194 on Build GRCh38
rs777267114 Chr.13: 106492913-106492913 on Build GRCh38
rs573369301 Chr.13: 106492960-106492960 on Build GRCh38
rs759361194 Chr.13: 106493154-106493154 on Build GRCh38
rs995121032 Chr.13: 106492895-106492895 on Build GRCh38
rs566875957 Chr.13: 106493091-106493091 on Build GRCh38
rs536066094 Chr.13: 106492849-106492849 on Build GRCh38
rs142390625 Chr.13: 106493057-106493057 on Build GRCh38
rs765559133 Chr.13: 106493184-106493184 on Build GRCh38
rs894437943 Chr.13: 106492911-106492911 on Build GRCh38
rs748940240 Chr.13: 106493208-106493208 on Build GRCh38
rs767489766 Chr.13: 106493084-106493084 on Build GRCh38
rs958854366 Chr.13: 106492951-106492951 on Build GRCh38
rs141594061 Chr.13: 106493118-106493118 on Build GRCh38
rs774268362 Chr.13: 106493148-106493148 on Build GRCh38
rs893787101 Chr.13: 106492862-106492862 on Build GRCh38
rs546052825 Chr.13: 106492987-106492987 on Build GRCh38
rs1002622544 Chr.13: 106492898-106492898 on Build GRCh38
rs200504952 Chr.13: 106493064-106493064 on Build GRCh38
rs752089244 Chr.13: 106493263-106493263 on Build GRCh38
rs183251835 Chr.13: 106493074-106493074 on Build GRCh38
rs945913681 Chr.13: 106492990-106492990 on Build GRCh38

View Full Product Details