Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110713504-110714008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
XM_006719953.2 NM_198217.2 NM_198218.2 NM_198219.2 XM_011521137.2

Gene Symbol
ING1
Entrez Gene ID
3621
Gene Name
inhibitor of growth family member 1
Gene Aliases
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a
UniGene
Hs.46700
Gene Chromosome Location
Chr.13: 110712623-110721074 on Build GRCh38
Transcripts
XM_006719953.2 NM_198217.2 NM_198218.2 NM_198219.2 XM_011521137.2

Gene Symbol
LOC105376684
Entrez Gene ID
105376684
Gene Name
translation initiation factor IF-2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.13: 110710644-110714090 on Build GRCh38
Transcripts
XM_006719953.2 NM_198217.2 NM_198218.2 NM_198219.2 XM_011521137.2

SNP Detail

SNP ID Chromosome Location
rs970459002 Chr.13: 110713641-110713641 on Build GRCh38
rs941955712 Chr.13: 110713572-110713572 on Build GRCh38
rs887092934 Chr.13: 110713725-110713725 on Build GRCh38
rs991003635 Chr.13: 110713637-110713637 on Build GRCh38
rs946219841 Chr.13: 110713600-110713600 on Build GRCh38
rs1037559797 Chr.13: 110713585-110713585 on Build GRCh38
rs1047380981 Chr.13: 110713559-110713559 on Build GRCh38
rs1051328568 Chr.13: 110713964-110713964 on Build GRCh38
rs995043319 Chr.13: 110713776-110713776 on Build GRCh38
rs934259352 Chr.13: 110713723-110713723 on Build GRCh38
rs1020967538 Chr.13: 110713610-110713610 on Build GRCh38
rs1001128742 Chr.13: 110713629-110713629 on Build GRCh38
rs1052239288 Chr.13: 110713613-110713613 on Build GRCh38
rs976026946 Chr.13: 110713907-110713907 on Build GRCh38
rs1000478664 Chr.13: 110713607-110713607 on Build GRCh38
rs963358890 Chr.13: 110713574-110713574 on Build GRCh38
rs907515258 Chr.13: 110713564-110713564 on Build GRCh38
rs549380869 Chr.13: 110713617-110713617 on Build GRCh38
rs973715666 Chr.13: 110713581-110713581 on Build GRCh38
rs996004323 Chr.13: 110713597-110713597 on Build GRCh38
rs1051129136 Chr.13: 110713724-110713724 on Build GRCh38
rs958404411 Chr.13: 110713669-110713669 on Build GRCh38
rs886325459 Chr.13: 110713613-110713637 on Build GRCh38
rs71127970 Chr.13: 110713860-110713861 on Build GRCh38
rs907566385 Chr.13: 110713660-110713660 on Build GRCh38
rs1004323908 Chr.13: 110713727-110713727 on Build GRCh38
rs868287918 Chr.13: 110713751-110713751 on Build GRCh38
rs959831975 Chr.13: 110713636-110713636 on Build GRCh38
rs927001190 Chr.13: 110713592-110713592 on Build GRCh38
rs1037604105 Chr.13: 110713677-110713677 on Build GRCh38
rs973361753 Chr.13: 110713664-110713664 on Build GRCh38
rs983123774 Chr.13: 110713652-110713652 on Build GRCh38
rs538030078 Chr.13: 110713912-110713912 on Build GRCh38
rs556821678 Chr.13: 110713913-110713913 on Build GRCh38
rs1008279250 Chr.13: 110713609-110713609 on Build GRCh38
rs1032583906 Chr.13: 110713635-110713635 on Build GRCh38
rs958911021 Chr.13: 110713858-110713858 on Build GRCh38
rs900045326 Chr.13: 110713711-110713711 on Build GRCh38
rs1039964388 Chr.13: 110713700-110713700 on Build GRCh38
rs71127969 Chr.13: 110713853-110713853 on Build GRCh38
rs1023950130 Chr.13: 110713874-110713874 on Build GRCh38
rs1008569238 Chr.13: 110713671-110713691 on Build GRCh38
rs192537175 Chr.13: 110713879-110713879 on Build GRCh38
rs891203550 Chr.13: 110713608-110713608 on Build GRCh38
rs899844655 Chr.13: 110713625-110713625 on Build GRCh38
rs977156273 Chr.13: 110713883-110713883 on Build GRCh38
rs917695154 Chr.13: 110713640-110713640 on Build GRCh38
rs534070711 Chr.13: 110713766-110713766 on Build GRCh38
rs992692660 Chr.13: 110713866-110713866 on Build GRCh38
rs182296162 Chr.13: 110713962-110713962 on Build GRCh38
rs936140335 Chr.13: 110713692-110713692 on Build GRCh38
rs945830787 Chr.13: 110713678-110713678 on Build GRCh38
rs941672411 Chr.13: 110713668-110713668 on Build GRCh38
rs558770251 Chr.13: 110713768-110713768 on Build GRCh38
rs71445078 Chr.13: 110713605-110713605 on Build GRCh38
rs900361524 Chr.13: 110713594-110713594 on Build GRCh38
rs548544774 Chr.13: 110713898-110713916 on Build GRCh38
rs1041904511 Chr.13: 110713603-110713603 on Build GRCh38
rs983244349 Chr.13: 110713560-110713560 on Build GRCh38
rs926163744 Chr.13: 110713679-110713679 on Build GRCh38
rs921436355 Chr.13: 110713954-110713954 on Build GRCh38
rs2289461 Chr.13: 110713558-110713558 on Build GRCh38
rs966675951 Chr.13: 110713616-110713616 on Build GRCh38
rs977125016 Chr.13: 110713679-110713680 on Build GRCh38
rs936186585 Chr.13: 110713906-110713906 on Build GRCh38
rs536248412 Chr.13: 110713543-110713543 on Build GRCh38
rs1034270852 Chr.13: 110713810-110713810 on Build GRCh38
rs925660272 Chr.13: 110713896-110713896 on Build GRCh38

View Full Product Details