Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110719517-110720023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ING1
Entrez Gene ID
3621
Gene Name
inhibitor of growth family member 1
Gene Aliases
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a
UniGene
Hs.46700
Gene Chromosome Location
Chr.13: 110712623-110721074 on Build GRCh38
Transcripts
NM_001267728.1 NM_005537.5 NM_198217.2 NM_198218.2 NM_198219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2263330 c.923A>C p.N308T Chr.13: 110719586-110719586 on Build GRCh38
1586841 c.573G>A p.W191* Chr.13: 110719797-110719797 on Build GRCh38
4829300 c.886C>T p.Q296* Chr.13: 110719549-110719549 on Build GRCh38
4147583 c.355T>C p.S119P Chr.13: 110719579-110719579 on Build GRCh38
6218688 c.513C>T p.C171C Chr.13: 110719737-110719737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963641178 Chr.13: 110719590-110719590 on Build GRCh38
rs144877032 Chr.13: 110719821-110719821 on Build GRCh38
rs370012603 Chr.13: 110719560-110719560 on Build GRCh38
rs121909252 Chr.13: 110719667-110719667 on Build GRCh38
rs758157180 Chr.13: 110719823-110719823 on Build GRCh38
rs1028537467 Chr.13: 110719808-110719808 on Build GRCh38
rs769708556 Chr.13: 110719892-110719892 on Build GRCh38
rs773244634 Chr.13: 110719694-110719694 on Build GRCh38
rs770053893 Chr.13: 110719689-110719689 on Build GRCh38
rs748124745 Chr.13: 110719872-110719872 on Build GRCh38
rs752202339 Chr.13: 110719546-110719546 on Build GRCh38
rs772040695 Chr.13: 110719573-110719573 on Build GRCh38
rs749996318 Chr.13: 110719818-110719818 on Build GRCh38
rs762672395 Chr.13: 110719951-110719951 on Build GRCh38
rs755560607 Chr.13: 110719554-110719554 on Build GRCh38
rs767282482 Chr.13: 110719991-110719991 on Build GRCh38
rs773617626 Chr.13: 110719605-110719605 on Build GRCh38
rs761159633 Chr.13: 110719781-110719781 on Build GRCh38
rs376129156 Chr.13: 110719918-110719929 on Build GRCh38
rs140897205 Chr.13: 110719619-110719619 on Build GRCh38
rs745708355 Chr.13: 110719569-110719569 on Build GRCh38
rs142623633 Chr.13: 110719599-110719599 on Build GRCh38
rs948223028 Chr.13: 110719779-110719779 on Build GRCh38
rs749479059 Chr.13: 110719908-110719908 on Build GRCh38
rs751450371 Chr.13: 110719833-110719833 on Build GRCh38
rs753333733 Chr.13: 110719644-110719644 on Build GRCh38
rs372476660 Chr.13: 110719764-110719764 on Build GRCh38
rs987306261 Chr.13: 110719977-110719977 on Build GRCh38
rs936939145 Chr.13: 110719725-110719725 on Build GRCh38
rs778495983 Chr.13: 110719654-110719654 on Build GRCh38
rs760010029 Chr.13: 110719755-110719755 on Build GRCh38
rs121909251 Chr.13: 110719739-110719739 on Build GRCh38
rs753383406 Chr.13: 110719770-110719770 on Build GRCh38
rs765598475 Chr.13: 110719537-110719537 on Build GRCh38
rs761392197 Chr.13: 110719983-110719983 on Build GRCh38
rs930579374 Chr.13: 110719601-110719601 on Build GRCh38
rs750797494 Chr.13: 110719538-110719538 on Build GRCh38
rs563266503 Chr.13: 110719685-110719685 on Build GRCh38
rs754956901 Chr.13: 110719856-110719856 on Build GRCh38
rs1033198415 Chr.13: 110719686-110719686 on Build GRCh38
rs1018746719 Chr.13: 110719988-110719988 on Build GRCh38
rs953171378 Chr.13: 110719610-110719610 on Build GRCh38
rs756920289 Chr.13: 110719653-110719653 on Build GRCh38
rs540953691 Chr.13: 110719602-110719602 on Build GRCh38
rs771242725 Chr.13: 110719746-110719746 on Build GRCh38
rs774395584 Chr.13: 110719554-110719556 on Build GRCh38
rs908523705 Chr.13: 110719914-110719914 on Build GRCh38
rs121909250 Chr.13: 110719736-110719736 on Build GRCh38
rs376816952 Chr.13: 110719673-110719673 on Build GRCh38
rs76490034 Chr.13: 110719611-110719611 on Build GRCh38
rs758921732 Chr.13: 110719541-110719541 on Build GRCh38
rs762064751 Chr.13: 110719600-110719600 on Build GRCh38
rs780018985 Chr.13: 110719672-110719672 on Build GRCh38
rs138833737 Chr.13: 110719869-110719869 on Build GRCh38
rs768754146 Chr.13: 110719674-110719674 on Build GRCh38
rs372262720 Chr.13: 110719594-110719594 on Build GRCh38
rs368230046 Chr.13: 110719955-110719955 on Build GRCh38
rs113769970 Chr.13: 110719550-110719550 on Build GRCh38
rs148838936 Chr.13: 110719545-110719545 on Build GRCh38
rs775801641 Chr.13: 110719943-110719943 on Build GRCh38
rs191659851 Chr.13: 110719939-110719939 on Build GRCh38
rs761265448 Chr.13: 110719945-110719945 on Build GRCh38
rs372407664 Chr.13: 110719973-110719973 on Build GRCh38
rs764815430 Chr.13: 110719794-110719794 on Build GRCh38
rs1046167942 Chr.13: 110719788-110719788 on Build GRCh38
rs763256127 Chr.13: 110719731-110719731 on Build GRCh38
rs777936457 Chr.13: 110719904-110719904 on Build GRCh38
rs758208164 Chr.13: 110719664-110719664 on Build GRCh38
rs367836624 Chr.13: 110719583-110719583 on Build GRCh38
rs878889369 Chr.13: 110719578-110719578 on Build GRCh38
rs754755764 Chr.13: 110719978-110719978 on Build GRCh38
rs750093259 Chr.13: 110719656-110719656 on Build GRCh38
rs370587007 Chr.13: 110719919-110719919 on Build GRCh38
rs778745842 Chr.13: 110719568-110719568 on Build GRCh38
rs767411112 Chr.13: 110719671-110719671 on Build GRCh38
rs953233104 Chr.13: 110719974-110719974 on Build GRCh38
rs368016398 Chr.13: 110719916-110719916 on Build GRCh38
rs760064895 Chr.13: 110719618-110719618 on Build GRCh38
rs368239053 Chr.13: 110719678-110719678 on Build GRCh38
rs768809382 Chr.13: 110719596-110719596 on Build GRCh38
rs772821020 Chr.13: 110719948-110719948 on Build GRCh38
rs748926410 Chr.13: 110719563-110719563 on Build GRCh38
rs373692673 Chr.13: 110719567-110719567 on Build GRCh38
rs1013960719 Chr.13: 110719616-110719616 on Build GRCh38
rs944102743 Chr.13: 110719652-110719652 on Build GRCh38
rs759960582 Chr.13: 110719922-110719922 on Build GRCh38
rs781310053 Chr.13: 110719677-110719677 on Build GRCh38
rs779964058 Chr.13: 110719825-110719825 on Build GRCh38

View Full Product Details