Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22971556-22972065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJUBA
Entrez Gene ID
84962
Gene Name
ajuba LIM protein
Gene Aliases
JUB
UniGene
Hs.655832
Gene Chromosome Location
Chr.14: 22971174-22982642 on Build GRCh38
Transcripts
NM_032876.5 NM_198086.2

SNP Detail

SNP ID Chromosome Location
rs1034787615 Chr.14: 22971658-22971658 on Build GRCh38
rs1013176388 Chr.14: 22971783-22971783 on Build GRCh38
rs1039068073 Chr.14: 22971893-22971893 on Build GRCh38
rs1042754715 Chr.14: 22971620-22971620 on Build GRCh38
rs138976325 Chr.14: 22971809-22971809 on Build GRCh38
rs112248555 Chr.14: 22971698-22971698 on Build GRCh38
rs142877927 Chr.14: 22971832-22971832 on Build GRCh38
rs920593461 Chr.14: 22971931-22971931 on Build GRCh38
rs753112817 Chr.14: 22971641-22971641 on Build GRCh38
rs556414097 Chr.14: 22971858-22971858 on Build GRCh38
rs972372335 Chr.14: 22971968-22971970 on Build GRCh38
rs534702311 Chr.14: 22971731-22971731 on Build GRCh38
rs1047592773 Chr.14: 22971621-22971621 on Build GRCh38
rs988022358 Chr.14: 22971831-22971831 on Build GRCh38
rs879318108 Chr.14: 22971884-22971884 on Build GRCh38
rs998813321 Chr.14: 22971666-22971669 on Build GRCh38
rs754333910 Chr.14: 22972026-22972026 on Build GRCh38
rs1022372561 Chr.14: 22971705-22971705 on Build GRCh38
rs1923 Chr.14: 22971970-22971970 on Build GRCh38
rs374972649 Chr.14: 22971988-22971988 on Build GRCh38
rs886252382 Chr.14: 22971637-22971637 on Build GRCh38
rs978991004 Chr.14: 22971781-22971781 on Build GRCh38
rs181021342 Chr.14: 22972030-22972030 on Build GRCh38
rs886327510 Chr.14: 22972036-22972036 on Build GRCh38
rs1011924218 Chr.14: 22971700-22971700 on Build GRCh38
rs368443909 Chr.14: 22971800-22971800 on Build GRCh38
rs903095761 Chr.14: 22971648-22971648 on Build GRCh38
rs969000438 Chr.14: 22971744-22971744 on Build GRCh38

View Full Product Details