Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24172792-24173288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
UniGene
Hs.419259
Gene Chromosome Location
Chr.14: 24171853-24180408 on Build GRCh38
Transcripts
NM_001048205.1 NM_005132.2 XM_011537421.1 XM_017021841.1

SNP Detail

SNP ID Chromosome Location
rs766969462 Chr.14: 24173008-24173008 on Build GRCh38
rs117843614 Chr.14: 24173186-24173186 on Build GRCh38
rs373659526 Chr.14: 24172884-24172884 on Build GRCh38
rs370925288 Chr.14: 24172984-24172984 on Build GRCh38
rs774002488 Chr.14: 24173122-24173122 on Build GRCh38
rs377239745 Chr.14: 24172890-24172890 on Build GRCh38
rs755993029 Chr.14: 24173212-24173212 on Build GRCh38
rs768500858 Chr.14: 24172971-24172971 on Build GRCh38
rs769505314 Chr.14: 24172999-24172999 on Build GRCh38
rs746788540 Chr.14: 24172964-24172964 on Build GRCh38
rs759974100 Chr.14: 24173015-24173015 on Build GRCh38
rs531673518 Chr.14: 24173047-24173047 on Build GRCh38
rs566992765 Chr.14: 24173238-24173238 on Build GRCh38
rs374527186 Chr.14: 24172827-24172827 on Build GRCh38
rs774340062 Chr.14: 24172895-24172895 on Build GRCh38
rs757328194 Chr.14: 24173164-24173164 on Build GRCh38
rs759362693 Chr.14: 24172821-24172821 on Build GRCh38
rs775269272 Chr.14: 24172825-24172825 on Build GRCh38
rs776318769 Chr.14: 24172914-24172914 on Build GRCh38
rs200486769 Chr.14: 24172932-24172932 on Build GRCh38
rs756243247 Chr.14: 24173051-24173051 on Build GRCh38
rs754866991 Chr.14: 24173190-24173190 on Build GRCh38
rs535859181 Chr.14: 24173105-24173105 on Build GRCh38
rs185284630 Chr.14: 24173046-24173046 on Build GRCh38
rs530107293 Chr.14: 24173150-24173150 on Build GRCh38
rs200268439 Chr.14: 24173177-24173177 on Build GRCh38
rs377625914 Chr.14: 24172862-24172862 on Build GRCh38
rs748903793 Chr.14: 24173226-24173226 on Build GRCh38
rs772554361 Chr.14: 24173001-24173001 on Build GRCh38
rs770707012 Chr.14: 24172815-24172816 on Build GRCh38
rs776800870 Chr.14: 24172815-24172815 on Build GRCh38
rs780010233 Chr.14: 24172956-24172956 on Build GRCh38
rs779957226 Chr.14: 24173073-24173073 on Build GRCh38
rs770631076 Chr.14: 24173118-24173118 on Build GRCh38
rs747019445 Chr.14: 24173085-24173085 on Build GRCh38
rs367867254 Chr.14: 24173152-24173152 on Build GRCh38
rs775219506 Chr.14: 24172908-24172908 on Build GRCh38
rs753834153 Chr.14: 24173065-24173065 on Build GRCh38
rs532908197 Chr.14: 24172818-24172818 on Build GRCh38
rs759494531 Chr.14: 24173251-24173252 on Build GRCh38
rs772061360 Chr.14: 24172903-24172903 on Build GRCh38
rs764449043 Chr.14: 24173254-24173254 on Build GRCh38
rs749410088 Chr.14: 24172952-24172952 on Build GRCh38
rs574388918 Chr.14: 24172852-24172852 on Build GRCh38
rs747871841 Chr.14: 24173199-24173199 on Build GRCh38
rs770720883 Chr.14: 24173228-24173228 on Build GRCh38
rs776265767 Chr.14: 24172980-24172980 on Build GRCh38
rs370197598 Chr.14: 24172900-24172900 on Build GRCh38
rs773083264 Chr.14: 24172913-24172913 on Build GRCh38
rs977055303 Chr.14: 24173070-24173070 on Build GRCh38
rs777285252 Chr.14: 24173222-24173222 on Build GRCh38
rs769297310 Chr.14: 24173099-24173099 on Build GRCh38
rs374389257 Chr.14: 24173251-24173251 on Build GRCh38
rs374576793 Chr.14: 24173086-24173086 on Build GRCh38
rs1014681635 Chr.14: 24172861-24172861 on Build GRCh38
rs11547058 Chr.14: 24173139-24173139 on Build GRCh38
rs774075683 Chr.14: 24172869-24172870 on Build GRCh38
rs987911856 Chr.14: 24172998-24172998 on Build GRCh38
rs371053208 Chr.14: 24172873-24172873 on Build GRCh38
rs757409570 Chr.14: 24173071-24173071 on Build GRCh38
rs745339797 Chr.14: 24173241-24173241 on Build GRCh38
rs955946243 Chr.14: 24173033-24173034 on Build GRCh38
rs940841948 Chr.14: 24172963-24172963 on Build GRCh38
rs368888997 Chr.14: 24173242-24173242 on Build GRCh38
rs761377685 Chr.14: 24172924-24172924 on Build GRCh38
rs1037171367 Chr.14: 24173208-24173208 on Build GRCh38
rs371730396 Chr.14: 24173184-24173184 on Build GRCh38
rs35282724 Chr.14: 24173132-24173133 on Build GRCh38
rs761142959 Chr.14: 24173149-24173149 on Build GRCh38
rs753034831 Chr.14: 24172942-24172942 on Build GRCh38
rs776013931 Chr.14: 24173245-24173245 on Build GRCh38
rs764580362 Chr.14: 24172931-24172931 on Build GRCh38
rs200093612 Chr.14: 24172933-24172933 on Build GRCh38
rs745726857 Chr.14: 24172899-24172899 on Build GRCh38
rs552447292 Chr.14: 24173006-24173006 on Build GRCh38
rs1037287634 Chr.14: 24172819-24172819 on Build GRCh38
rs748004158 Chr.14: 24173097-24173097 on Build GRCh38
rs776851779 Chr.14: 24173262-24173262 on Build GRCh38
rs1033024541 Chr.14: 24172912-24172912 on Build GRCh38
rs781145214 Chr.14: 24173196-24173196 on Build GRCh38
rs767283219 Chr.14: 24172822-24172822 on Build GRCh38
rs771866577 Chr.14: 24172953-24172953 on Build GRCh38
rs778089515 Chr.14: 24173058-24173058 on Build GRCh38
rs368303020 Chr.14: 24173109-24173109 on Build GRCh38
rs960665046 Chr.14: 24172829-24172835 on Build GRCh38
rs868041668 Chr.14: 24173137-24173138 on Build GRCh38
rs752168235 Chr.14: 24173011-24173011 on Build GRCh38
rs748267625 Chr.14: 24172816-24172816 on Build GRCh38
rs763760543 Chr.14: 24172831-24172831 on Build GRCh38
rs756363583 Chr.14: 24172947-24172947 on Build GRCh38
rs905859018 Chr.14: 24172866-24172866 on Build GRCh38
rs751939300 Chr.14: 24172937-24172937 on Build GRCh38
rs758943574 Chr.14: 24172876-24172876 on Build GRCh38
rs930774819 Chr.14: 24172896-24172896 on Build GRCh38
rs1040617224 Chr.14: 24172978-24172978 on Build GRCh38
rs372055976 Chr.14: 24173127-24173127 on Build GRCh38
rs1020841174 Chr.14: 24173037-24173037 on Build GRCh38
rs774203794 Chr.14: 24172820-24172820 on Build GRCh38
rs780222210 Chr.14: 24172881-24172881 on Build GRCh38
rs367739958 Chr.14: 24172941-24172941 on Build GRCh38
rs780912384 Chr.14: 24173096-24173096 on Build GRCh38
rs754149940 Chr.14: 24173159-24173159 on Build GRCh38

View Full Product Details