Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24373615-24374119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFATC4
Entrez Gene ID
4776
Gene Name
nuclear factor of activated T-cells 4
Gene Aliases
NF-AT3, NF-ATC4, NFAT3
UniGene
Hs.77810
Gene Chromosome Location
Chr.14: 24366911-24379604 on Build GRCh38
Transcripts
NM_001136022.2 NM_001198965.1 NM_001198966.2 NM_001198967.2 NM_001288802.1 NM_001320043.1 NM_004554.4 XM_011536797.2 XM_011536798.1 XM_011536799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955954 c.1904C>T p.S635L Chr.14: 24373850-24373850 on Build GRCh38
4245017 c.1546A>T p.K516* Chr.14: 24373717-24373717 on Build GRCh38
4917712 c.1884C>A p.V628V Chr.14: 24373830-24373830 on Build GRCh38
3955956 c.1679C>T p.S560L Chr.14: 24373850-24373850 on Build GRCh38
6356222 c.1533C>T p.C511C Chr.14: 24373704-24373704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766872126 Chr.14: 24374065-24374065 on Build GRCh38
rs376056202 Chr.14: 24373956-24373956 on Build GRCh38
rs750925454 Chr.14: 24373910-24373910 on Build GRCh38
rs996396793 Chr.14: 24373640-24373640 on Build GRCh38
rs202057536 Chr.14: 24373677-24373677 on Build GRCh38
rs772343312 Chr.14: 24373744-24373744 on Build GRCh38
rs778408359 Chr.14: 24373706-24373706 on Build GRCh38
rs113966193 Chr.14: 24374061-24374061 on Build GRCh38
rs374808156 Chr.14: 24373807-24373807 on Build GRCh38
rs775517670 Chr.14: 24373745-24373745 on Build GRCh38
rs370448673 Chr.14: 24373963-24373963 on Build GRCh38
rs376632262 Chr.14: 24373977-24373977 on Build GRCh38
rs377261658 Chr.14: 24373834-24373834 on Build GRCh38
rs775177810 Chr.14: 24373976-24373976 on Build GRCh38
rs1006249919 Chr.14: 24374082-24374082 on Build GRCh38
rs762964789 Chr.14: 24373764-24373764 on Build GRCh38
rs757790727 Chr.14: 24373819-24373819 on Build GRCh38
rs926989284 Chr.14: 24373837-24373837 on Build GRCh38
rs748255553 Chr.14: 24373955-24373955 on Build GRCh38
rs552610225 Chr.14: 24373704-24373704 on Build GRCh38
rs375252788 Chr.14: 24373919-24373920 on Build GRCh38
rs779325479 Chr.14: 24373820-24373820 on Build GRCh38
rs914916058 Chr.14: 24373940-24373940 on Build GRCh38
rs763486854 Chr.14: 24373812-24373812 on Build GRCh38
rs754704463 Chr.14: 24373687-24373687 on Build GRCh38
rs771994022 Chr.14: 24373975-24373975 on Build GRCh38
rs368810099 Chr.14: 24373685-24373685 on Build GRCh38
rs764696742 Chr.14: 24374046-24374046 on Build GRCh38
rs760449995 Chr.14: 24373987-24373987 on Build GRCh38
rs762909955 Chr.14: 24373657-24373657 on Build GRCh38
rs1004932777 Chr.14: 24373781-24373781 on Build GRCh38
rs773243252 Chr.14: 24373762-24373762 on Build GRCh38
rs372251932 Chr.14: 24373703-24373703 on Build GRCh38
rs56375538 Chr.14: 24373831-24373831 on Build GRCh38
rs781501608 Chr.14: 24373950-24373950 on Build GRCh38
rs773759241 Chr.14: 24374058-24374058 on Build GRCh38
rs781446521 Chr.14: 24373836-24373836 on Build GRCh38
rs757991603 Chr.14: 24373942-24373942 on Build GRCh38
rs140124494 Chr.14: 24373723-24373723 on Build GRCh38
rs755562231 Chr.14: 24373705-24373705 on Build GRCh38
rs750800356 Chr.14: 24373821-24373821 on Build GRCh38
rs780261317 Chr.14: 24373920-24373920 on Build GRCh38
rs902102797 Chr.14: 24373988-24373988 on Build GRCh38
rs886458276 Chr.14: 24374070-24374070 on Build GRCh38
rs780390718 Chr.14: 24373830-24373830 on Build GRCh38
rs767254119 Chr.14: 24373806-24373806 on Build GRCh38
rs369520588 Chr.14: 24373818-24373818 on Build GRCh38
rs147385533 Chr.14: 24373815-24373815 on Build GRCh38
rs761379514 Chr.14: 24374022-24374022 on Build GRCh38
rs371593262 Chr.14: 24373673-24373673 on Build GRCh38
rs887836181 Chr.14: 24374071-24374071 on Build GRCh38
rs200178397 Chr.14: 24373851-24373851 on Build GRCh38
rs761254745 Chr.14: 24373887-24373887 on Build GRCh38
rs759341999 Chr.14: 24373804-24373804 on Build GRCh38
rs373067097 Chr.14: 24373903-24373903 on Build GRCh38
rs768310407 Chr.14: 24374012-24374012 on Build GRCh38
rs779030311 Chr.14: 24373960-24373960 on Build GRCh38
rs966630689 Chr.14: 24373828-24373828 on Build GRCh38
rs34396832 Chr.14: 24373693-24373694 on Build GRCh38
rs751878261 Chr.14: 24374072-24374072 on Build GRCh38
rs534231892 Chr.14: 24373860-24373860 on Build GRCh38
rs372002281 Chr.14: 24373769-24373769 on Build GRCh38
rs763760316 Chr.14: 24373870-24373870 on Build GRCh38
rs142749204 Chr.14: 24373757-24373757 on Build GRCh38
rs142988462 Chr.14: 24373755-24373755 on Build GRCh38
rs755189601 Chr.14: 24373933-24373933 on Build GRCh38
rs749761329 Chr.14: 24373713-24373713 on Build GRCh38
rs1029459365 Chr.14: 24373643-24373643 on Build GRCh38
rs370444869 Chr.14: 24373867-24373867 on Build GRCh38
rs774413775 Chr.14: 24373855-24373855 on Build GRCh38
rs750315466 Chr.14: 24373918-24373919 on Build GRCh38
rs774322080 Chr.14: 24373793-24373793 on Build GRCh38
rs200783328 Chr.14: 24373833-24373833 on Build GRCh38
rs746189978 Chr.14: 24373743-24373743 on Build GRCh38
rs757320547 Chr.14: 24373912-24373913 on Build GRCh38
rs748202233 Chr.14: 24373843-24373843 on Build GRCh38
rs944994295 Chr.14: 24373942-24373942 on Build GRCh38
rs1054959977 Chr.14: 24373980-24373980 on Build GRCh38
rs930580275 Chr.14: 24374029-24374029 on Build GRCh38
rs776251655 Chr.14: 24374021-24374021 on Build GRCh38
rs147469288 Chr.14: 24373924-24373924 on Build GRCh38
rs372741212 Chr.14: 24373822-24373822 on Build GRCh38
rs753349529 Chr.14: 24373875-24373875 on Build GRCh38
rs573902036 Chr.14: 24374009-24374009 on Build GRCh38
rs995281931 Chr.14: 24373734-24373734 on Build GRCh38
rs757665853 Chr.14: 24373719-24373719 on Build GRCh38
rs758837842 Chr.14: 24373915-24373915 on Build GRCh38

View Full Product Details