Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31673355-31673853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUBPL
Entrez Gene ID
80224
Gene Name
nucleotide binding protein like
Gene Aliases
C14orf127, IND1, huInd1
UniGene
Hs.288981
Gene Chromosome Location
Chr.14: 31561385-31861293 on Build GRCh38
Transcripts
NM_001201573.1 NM_025152.2 NR_120408.1 XM_011537181.2 XM_011537182.2 XM_011537183.2 XM_017021664.1 XM_017021665.1 XM_017021666.1 XM_017021667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554293 c.448G>T p.E150* Chr.14: 31673509-31673509 on Build GRCh38
4429589 c.451A>G p.S151G Chr.14: 31673512-31673512 on Build GRCh38
4148079 c.400C>A p.L134I Chr.14: 31673372-31673372 on Build GRCh38
6463469 c.422+7A>G p.? Chr.14: 31673401-31673401 on Build GRCh38
6554294 c.358G>T p.E120* Chr.14: 31673509-31673509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201412882 Chr.14: 31673385-31673385 on Build GRCh38
rs893611788 Chr.14: 31673781-31673781 on Build GRCh38
rs749347684 Chr.14: 31673550-31673550 on Build GRCh38
rs759920548 Chr.14: 31673451-31673451 on Build GRCh38
rs368916142 Chr.14: 31673737-31673737 on Build GRCh38
rs761032613 Chr.14: 31673471-31673471 on Build GRCh38
rs768640478 Chr.14: 31673397-31673397 on Build GRCh38
rs999382263 Chr.14: 31673730-31673730 on Build GRCh38
rs372872090 Chr.14: 31673723-31673723 on Build GRCh38
rs991925697 Chr.14: 31673696-31673696 on Build GRCh38
rs749998110 Chr.14: 31673518-31673518 on Build GRCh38
rs8013530 Chr.14: 31673724-31673724 on Build GRCh38
rs1049540131 Chr.14: 31673810-31673810 on Build GRCh38
rs901863850 Chr.14: 31673544-31673544 on Build GRCh38
rs35568340 Chr.14: 31673652-31673653 on Build GRCh38
rs775705854 Chr.14: 31673583-31673583 on Build GRCh38
rs868061310 Chr.14: 31673402-31673402 on Build GRCh38
rs761169075 Chr.14: 31673598-31673598 on Build GRCh38
rs754322347 Chr.14: 31673793-31673793 on Build GRCh38
rs917544476 Chr.14: 31673708-31673708 on Build GRCh38
rs777174450 Chr.14: 31673609-31673609 on Build GRCh38
rs863224123 Chr.14: 31673483-31673483 on Build GRCh38
rs374317410 Chr.14: 31673599-31673599 on Build GRCh38
rs751380293 Chr.14: 31673528-31673528 on Build GRCh38
rs754303878 Chr.14: 31673733-31673733 on Build GRCh38
rs148053784 Chr.14: 31673649-31673649 on Build GRCh38
rs188151242 Chr.14: 31673526-31673526 on Build GRCh38
rs1032194712 Chr.14: 31673750-31673750 on Build GRCh38
rs761356840 Chr.14: 31673491-31673491 on Build GRCh38
rs779923548 Chr.14: 31673379-31673379 on Build GRCh38
rs1042319437 Chr.14: 31673436-31673436 on Build GRCh38
rs754143300 Chr.14: 31673507-31673510 on Build GRCh38
rs879199957 Chr.14: 31673772-31673772 on Build GRCh38
rs774755018 Chr.14: 31673448-31673448 on Build GRCh38
rs77139143 Chr.14: 31673695-31673695 on Build GRCh38
rs117470156 Chr.14: 31673663-31673663 on Build GRCh38
rs775929865 Chr.14: 31673469-31673469 on Build GRCh38
rs539880891 Chr.14: 31673710-31673710 on Build GRCh38
rs758237955 Chr.14: 31673637-31673638 on Build GRCh38
rs779154807 Chr.14: 31673565-31673565 on Build GRCh38
rs112193287 Chr.14: 31673776-31673776 on Build GRCh38
rs752671452 Chr.14: 31673403-31673403 on Build GRCh38
rs749433462 Chr.14: 31673401-31673401 on Build GRCh38
rs777939696 Chr.14: 31673399-31673399 on Build GRCh38
rs756704678 Chr.14: 31673386-31673387 on Build GRCh38
rs754853511 Chr.14: 31673535-31673535 on Build GRCh38
rs771183085 Chr.14: 31673552-31673552 on Build GRCh38
rs781341998 Chr.14: 31673549-31673549 on Build GRCh38
rs764852401 Chr.14: 31673504-31673504 on Build GRCh38
rs370326751 Chr.14: 31673435-31673435 on Build GRCh38
rs758104608 Chr.14: 31673519-31673519 on Build GRCh38
rs863224124 Chr.14: 31673487-31673487 on Build GRCh38
rs772353623 Chr.14: 31673581-31673581 on Build GRCh38
rs762334348 Chr.14: 31673622-31673622 on Build GRCh38
rs932074986 Chr.14: 31673690-31673691 on Build GRCh38
rs944685589 Chr.14: 31673719-31673719 on Build GRCh38
rs746035231 Chr.14: 31673568-31673568 on Build GRCh38
rs557538167 Chr.14: 31673453-31673453 on Build GRCh38
rs764643986 Chr.14: 31673477-31673477 on Build GRCh38
rs764688784 Chr.14: 31673623-31673623 on Build GRCh38

View Full Product Details