Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52634696-52635241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR137C
Entrez Gene ID
283554
Gene Name
G protein-coupled receptor 137C
Gene Aliases
TM7SF1L2
UniGene
Hs.416214
Gene Chromosome Location
Chr.14: 52553148-52637713 on Build GRCh38
Transcripts
NM_001099652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369426 c.1219A>T p.S407C Chr.14: 52634996-52634996 on Build GRCh38
6015523 c.1235C>T p.P412L Chr.14: 52635012-52635012 on Build GRCh38
4051282 c.1175A>G p.Q392R Chr.14: 52634952-52634952 on Build GRCh38
7123810 c.1300_1303delAACA p.N434fs*7 Chr.14: 52635077-52635080 on Build GRCh38
2199584 c.1262C>A p.T421N Chr.14: 52635039-52635039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746716737 Chr.14: 52635036-52635036 on Build GRCh38
rs773699890 Chr.14: 52634880-52634880 on Build GRCh38
rs761948230 Chr.14: 52634922-52634923 on Build GRCh38
rs776261186 Chr.14: 52635064-52635064 on Build GRCh38
rs939307317 Chr.14: 52635209-52635209 on Build GRCh38
rs775971492 Chr.14: 52634945-52634945 on Build GRCh38
rs769362925 Chr.14: 52635072-52635072 on Build GRCh38
rs376670463 Chr.14: 52634926-52634926 on Build GRCh38
rs774252808 Chr.14: 52635181-52635181 on Build GRCh38
rs898168185 Chr.14: 52634859-52634860 on Build GRCh38
rs780712927 Chr.14: 52635165-52635165 on Build GRCh38
rs752791772 Chr.14: 52634891-52634891 on Build GRCh38
rs939191243 Chr.14: 52634814-52634814 on Build GRCh38
rs754540403 Chr.14: 52634726-52634726 on Build GRCh38
rs926539337 Chr.14: 52635169-52635169 on Build GRCh38
rs199957068 Chr.14: 52634908-52634908 on Build GRCh38
rs754029226 Chr.14: 52635108-52635108 on Build GRCh38
rs772449876 Chr.14: 52635023-52635023 on Build GRCh38
rs556023679 Chr.14: 52635171-52635171 on Build GRCh38
rs773997522 Chr.14: 52635084-52635084 on Build GRCh38
rs367668896 Chr.14: 52634894-52634894 on Build GRCh38
rs192084281 Chr.14: 52634797-52634797 on Build GRCh38
rs761018237 Chr.14: 52635099-52635099 on Build GRCh38
rs765753279 Chr.14: 52634841-52634841 on Build GRCh38
rs148689145 Chr.14: 52634743-52634743 on Build GRCh38
rs1000412519 Chr.14: 52634995-52634995 on Build GRCh38
rs747320687 Chr.14: 52635158-52635158 on Build GRCh38
rs762932223 Chr.14: 52635074-52635074 on Build GRCh38
rs759576879 Chr.14: 52635101-52635102 on Build GRCh38
rs17803993 Chr.14: 52634863-52634863 on Build GRCh38
rs556636863 Chr.14: 52634954-52634954 on Build GRCh38
rs760199200 Chr.14: 52634936-52634936 on Build GRCh38
rs776759364 Chr.14: 52634923-52634923 on Build GRCh38
rs964064942 Chr.14: 52634742-52634742 on Build GRCh38
rs754725690 Chr.14: 52635160-52635160 on Build GRCh38
rs766215331 Chr.14: 52634957-52634957 on Build GRCh38
rs777960857 Chr.14: 52634971-52634971 on Build GRCh38
rs907761233 Chr.14: 52634836-52634836 on Build GRCh38
rs142832293 Chr.14: 52635138-52635138 on Build GRCh38
rs756180629 Chr.14: 52635162-52635162 on Build GRCh38
rs1037334844 Chr.14: 52634848-52634848 on Build GRCh38
rs761857890 Chr.14: 52634949-52634949 on Build GRCh38
rs779311512 Chr.14: 52634909-52634909 on Build GRCh38
rs956128799 Chr.14: 52635002-52635002 on Build GRCh38
rs990151543 Chr.14: 52634728-52634728 on Build GRCh38
rs571327668 Chr.14: 52634904-52634904 on Build GRCh38
rs752984384 Chr.14: 52634958-52634958 on Build GRCh38
rs768246062 Chr.14: 52634942-52634942 on Build GRCh38
rs563726406 Chr.14: 52634748-52634748 on Build GRCh38
rs368565116 Chr.14: 52634950-52634950 on Build GRCh38
rs751475120 Chr.14: 52634932-52634933 on Build GRCh38
rs376672640 Chr.14: 52635156-52635156 on Build GRCh38
rs765420821 Chr.14: 52634929-52634931 on Build GRCh38
rs951564674 Chr.14: 52635216-52635216 on Build GRCh38
rs746230706 Chr.14: 52634913-52634913 on Build GRCh38
rs746316568 Chr.14: 52634994-52634994 on Build GRCh38
rs372409862 Chr.14: 52634889-52634889 on Build GRCh38
rs551634506 Chr.14: 52634864-52634864 on Build GRCh38
rs757831670 Chr.14: 52634991-52634991 on Build GRCh38
rs761402036 Chr.14: 52635068-52635068 on Build GRCh38
rs185338147 Chr.14: 52634834-52634834 on Build GRCh38
rs369204749 Chr.14: 52635145-52635145 on Build GRCh38
rs766305588 Chr.14: 52635083-52635083 on Build GRCh38
rs549270499 Chr.14: 52634822-52634822 on Build GRCh38
rs779401282 Chr.14: 52634992-52634992 on Build GRCh38
rs1020892704 Chr.14: 52634981-52634981 on Build GRCh38
rs764222363 Chr.14: 52635105-52635105 on Build GRCh38
rs200551681 Chr.14: 52635063-52635063 on Build GRCh38
rs373870825 Chr.14: 52634893-52634893 on Build GRCh38
rs202161909 Chr.14: 52635133-52635133 on Build GRCh38
rs753852584 Chr.14: 52634990-52634990 on Build GRCh38
rs933196184 Chr.14: 52634823-52634823 on Build GRCh38
rs780242846 Chr.14: 52635031-52635031 on Build GRCh38
rs775137834 Chr.14: 52634932-52634932 on Build GRCh38
rs771408127 Chr.14: 52634907-52634907 on Build GRCh38
rs529130938 Chr.14: 52634758-52634758 on Build GRCh38
rs373734865 Chr.14: 52635152-52635152 on Build GRCh38

View Full Product Details