Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57205691-57206235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_006544.3 XM_005267272.3

SNP Detail

SNP ID Chromosome Location
rs994786965 Chr.14: 57206005-57206005 on Build GRCh38
rs923599954 Chr.14: 57206149-57206149 on Build GRCh38
rs550322594 Chr.14: 57206115-57206115 on Build GRCh38
rs935237227 Chr.14: 57205787-57205787 on Build GRCh38
rs762055848 Chr.14: 57205734-57205734 on Build GRCh38
rs201768301 Chr.14: 57205907-57205908 on Build GRCh38
rs1048635428 Chr.14: 57205883-57205883 on Build GRCh38
rs576731092 Chr.14: 57205890-57205890 on Build GRCh38
rs1041053033 Chr.14: 57206210-57206210 on Build GRCh38
rs949009668 Chr.14: 57205777-57205777 on Build GRCh38
rs775376793 Chr.14: 57205762-57205762 on Build GRCh38
rs929642787 Chr.14: 57206196-57206196 on Build GRCh38
rs997760491 Chr.14: 57205996-57205996 on Build GRCh38
rs555509675 Chr.14: 57206175-57206175 on Build GRCh38
rs920084022 Chr.14: 57205730-57205730 on Build GRCh38
rs201413464 Chr.14: 57205742-57205743 on Build GRCh38
rs530797271 Chr.14: 57205775-57205775 on Build GRCh38
rs552282223 Chr.14: 57205779-57205779 on Build GRCh38
rs1008208448 Chr.14: 57205928-57205928 on Build GRCh38
rs377164569 Chr.14: 57206174-57206174 on Build GRCh38
rs568666186 Chr.14: 57206010-57206010 on Build GRCh38
rs3742577 Chr.14: 57205822-57205822 on Build GRCh38
rs990634080 Chr.14: 57205812-57205812 on Build GRCh38
rs749021863 Chr.14: 57206184-57206185 on Build GRCh38
rs893591834 Chr.14: 57205820-57205820 on Build GRCh38
rs767964731 Chr.14: 57206037-57206037 on Build GRCh38
rs113683876 Chr.14: 57205794-57205794 on Build GRCh38
rs889805213 Chr.14: 57205863-57205863 on Build GRCh38
rs138867006 Chr.14: 57206193-57206193 on Build GRCh38
rs1023413009 Chr.14: 57206106-57206106 on Build GRCh38
rs1039184617 Chr.14: 57205973-57205973 on Build GRCh38
rs568908051 Chr.14: 57206138-57206138 on Build GRCh38
rs1046079822 Chr.14: 57205859-57205859 on Build GRCh38
rs1006810116 Chr.14: 57205917-57205917 on Build GRCh38
rs1049773355 Chr.14: 57205844-57205844 on Build GRCh38
rs944120561 Chr.14: 57206205-57206205 on Build GRCh38
rs952962186 Chr.14: 57206093-57206093 on Build GRCh38
rs923910109 Chr.14: 57205786-57205786 on Build GRCh38
rs572931730 Chr.14: 57206164-57206164 on Build GRCh38
rs536122131 Chr.14: 57206097-57206097 on Build GRCh38
rs528701179 Chr.14: 57205881-57205881 on Build GRCh38
rs573866469 Chr.14: 57206182-57206182 on Build GRCh38
rs3742578 Chr.14: 57205997-57205997 on Build GRCh38
rs1053665989 Chr.14: 57205816-57205816 on Build GRCh38
rs929763529 Chr.14: 57205868-57205868 on Build GRCh38
rs764508650 Chr.14: 57205939-57205939 on Build GRCh38
rs7141911 Chr.14: 57206153-57206153 on Build GRCh38
rs760032230 Chr.14: 57206036-57206036 on Build GRCh38
rs972957214 Chr.14: 57205727-57205727 on Build GRCh38
rs958122791 Chr.14: 57205811-57205811 on Build GRCh38
rs961913376 Chr.14: 57205723-57205723 on Build GRCh38
rs117870712 Chr.14: 57206183-57206183 on Build GRCh38
rs997523457 Chr.14: 57205744-57205745 on Build GRCh38
rs1023416652 Chr.14: 57206161-57206161 on Build GRCh38
rs900797012 Chr.14: 57205991-57205991 on Build GRCh38
rs528382169 Chr.14: 57206150-57206150 on Build GRCh38
rs931195561 Chr.14: 57205906-57205907 on Build GRCh38

View Full Product Details