Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60282963-60283460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPM1A
Entrez Gene ID
5494
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases
PP2C-ALPHA, PP2CA, PP2Calpha
UniGene
Hs.130036
Gene Chromosome Location
Chr.14: 60245752-60299087 on Build GRCh38
Transcripts
NM_021003.4 NM_177951.2 NM_177952.2 XM_005267779.1 XM_005267781.1 XM_011536872.1 XM_011536876.1 XM_011536877.1 XM_011536878.1 XM_011536879.2 XM_011536880.2 XM_011536881.2 XM_011536882.2 XM_011536883.1 XM_011536884.1 XM_017021381.1 XM_017021382.1 XM_017021383.1 XM_017021384.1 XM_017021385.1 XM_017021386.1 XM_017021387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607801 c.740A>G p.N247S Chr.14: 60283443-60283443 on Build GRCh38
6458600 c.846_847insA p.P283fs*3 Chr.14: 60283330-60283331 on Build GRCh38
3956125 c.483A>G p.T161T Chr.14: 60283186-60283186 on Build GRCh38
1707403 c.883C>T p.H295Y Chr.14: 60283367-60283367 on Build GRCh38
6337879 c.534T>C p.F178F Chr.14: 60283018-60283018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757003031 Chr.14: 60283391-60283391 on Build GRCh38
rs760984276 Chr.14: 60283291-60283291 on Build GRCh38
rs966406542 Chr.14: 60283134-60283134 on Build GRCh38
rs143557303 Chr.14: 60283039-60283039 on Build GRCh38
rs766728463 Chr.14: 60282994-60282994 on Build GRCh38
rs758285377 Chr.14: 60283414-60283414 on Build GRCh38
rs746688567 Chr.14: 60283144-60283144 on Build GRCh38
rs1005007522 Chr.14: 60283084-60283084 on Build GRCh38
rs771540938 Chr.14: 60283120-60283120 on Build GRCh38
rs757235666 Chr.14: 60283114-60283114 on Build GRCh38
rs79077687 Chr.14: 60283210-60283210 on Build GRCh38
rs141310821 Chr.14: 60283141-60283141 on Build GRCh38
rs752773220 Chr.14: 60283282-60283282 on Build GRCh38
rs752932710 Chr.14: 60283044-60283044 on Build GRCh38
rs753724691 Chr.14: 60283390-60283390 on Build GRCh38
rs768255466 Chr.14: 60283151-60283151 on Build GRCh38
rs767681089 Chr.14: 60283270-60283270 on Build GRCh38
rs777084659 Chr.14: 60283048-60283048 on Build GRCh38
rs745451715 Chr.14: 60283116-60283116 on Build GRCh38
rs376869115 Chr.14: 60283024-60283024 on Build GRCh38
rs771521785 Chr.14: 60283216-60283216 on Build GRCh38
rs200599256 Chr.14: 60283155-60283155 on Build GRCh38
rs758821937 Chr.14: 60283064-60283064 on Build GRCh38
rs756020328 Chr.14: 60283050-60283050 on Build GRCh38
rs990260326 Chr.14: 60283249-60283249 on Build GRCh38
rs199542062 Chr.14: 60283311-60283311 on Build GRCh38
rs945792260 Chr.14: 60283324-60283324 on Build GRCh38
rs751803506 Chr.14: 60283428-60283428 on Build GRCh38
rs76181826 Chr.14: 60283280-60283280 on Build GRCh38
rs183906525 Chr.14: 60283336-60283336 on Build GRCh38
rs748897766 Chr.14: 60283051-60283051 on Build GRCh38
rs773629894 Chr.14: 60283168-60283168 on Build GRCh38
rs750341699 Chr.14: 60283403-60283403 on Build GRCh38
rs759729228 Chr.14: 60283240-60283240 on Build GRCh38
rs774715909 Chr.14: 60283219-60283219 on Build GRCh38
rs767914442 Chr.14: 60283036-60283036 on Build GRCh38
rs201779982 Chr.14: 60283094-60283094 on Build GRCh38
rs976111360 Chr.14: 60283209-60283209 on Build GRCh38
rs751672907 Chr.14: 60283021-60283021 on Build GRCh38
rs748972263 Chr.14: 60283053-60283053 on Build GRCh38

View Full Product Details