Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 61766755-61767279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAPC1
Entrez Gene ID
6617
Gene Name
small nuclear RNA activating complex polypeptide 1
Gene Aliases
PTFgamma, SNAP43
UniGene
Hs.179312
Gene Chromosome Location
Chr.14: 61762357-61796428 on Build GRCh38
Transcripts
NM_003082.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1226942 c.140G>T p.R47I Chr.14: 61766887-61766887 on Build GRCh38
5690210 c.286A>T p.K96* Chr.14: 61767033-61767033 on Build GRCh38
956816 c.284A>C p.Q95P Chr.14: 61767031-61767031 on Build GRCh38
956817 c.354G>T p.Q118H Chr.14: 61767277-61767277 on Build GRCh38
5470267 c.219G>A p.Q73Q Chr.14: 61766966-61766966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896614896 Chr.14: 61767229-61767229 on Build GRCh38
rs201269619 Chr.14: 61766939-61766939 on Build GRCh38
rs771687434 Chr.14: 61766983-61766983 on Build GRCh38
rs769365619 Chr.14: 61767255-61767255 on Build GRCh38
rs767986570 Chr.14: 61767069-61767069 on Build GRCh38
rs757292475 Chr.14: 61766973-61766973 on Build GRCh38
rs747665400 Chr.14: 61767239-61767239 on Build GRCh38
rs564400454 Chr.14: 61766793-61766793 on Build GRCh38
rs535458066 Chr.14: 61767138-61767138 on Build GRCh38
rs555011582 Chr.14: 61767146-61767146 on Build GRCh38
rs763658142 Chr.14: 61766886-61766886 on Build GRCh38
rs143102386 Chr.14: 61766792-61766792 on Build GRCh38
rs1036442487 Chr.14: 61767023-61767023 on Build GRCh38
rs377002053 Chr.14: 61766801-61766801 on Build GRCh38
rs766695182 Chr.14: 61766914-61766914 on Build GRCh38
rs759818826 Chr.14: 61767041-61767041 on Build GRCh38
rs763495489 Chr.14: 61766897-61766897 on Build GRCh38
rs371774333 Chr.14: 61767150-61767154 on Build GRCh38
rs953286704 Chr.14: 61766787-61766787 on Build GRCh38
rs768128467 Chr.14: 61767006-61767006 on Build GRCh38
rs375857200 Chr.14: 61766996-61766996 on Build GRCh38
rs372497186 Chr.14: 61766946-61766946 on Build GRCh38
rs753993133 Chr.14: 61767179-61767179 on Build GRCh38
rs200038137 Chr.14: 61766850-61766850 on Build GRCh38
rs1029530737 Chr.14: 61766995-61766995 on Build GRCh38
rs572732686 Chr.14: 61767106-61767106 on Build GRCh38
rs751754263 Chr.14: 61766827-61766827 on Build GRCh38
rs764231541 Chr.14: 61767177-61767177 on Build GRCh38
rs200916347 Chr.14: 61766937-61766937 on Build GRCh38
rs144535324 Chr.14: 61766936-61766936 on Build GRCh38
rs897530497 Chr.14: 61767080-61767080 on Build GRCh38
rs749298195 Chr.14: 61766849-61766849 on Build GRCh38
rs773693765 Chr.14: 61766894-61766894 on Build GRCh38
rs537207506 Chr.14: 61766951-61766951 on Build GRCh38
rs200019888 Chr.14: 61766993-61766993 on Build GRCh38
rs149332356 Chr.14: 61766917-61766917 on Build GRCh38
rs760105590 Chr.14: 61766871-61766871 on Build GRCh38
rs756446737 Chr.14: 61767085-61767085 on Build GRCh38
rs148216766 Chr.14: 61766847-61766847 on Build GRCh38
rs780626166 Chr.14: 61767224-61767224 on Build GRCh38
rs766887525 Chr.14: 61767022-61767022 on Build GRCh38
rs779944010 Chr.14: 61767211-61767211 on Build GRCh38
rs1254677 Chr.14: 61767088-61767088 on Build GRCh38
rs756654567 Chr.14: 61767189-61767190 on Build GRCh38
rs1000832247 Chr.14: 61766953-61766953 on Build GRCh38
rs778375845 Chr.14: 61767201-61767201 on Build GRCh38
rs550803347 Chr.14: 61766964-61766964 on Build GRCh38
rs780740457 Chr.14: 61766860-61766863 on Build GRCh38
rs201965687 Chr.14: 61767010-61767010 on Build GRCh38
rs148447482 Chr.14: 61766974-61766974 on Build GRCh38
rs73324981 Chr.14: 61766851-61766851 on Build GRCh38
rs145250202 Chr.14: 61767015-61767015 on Build GRCh38
rs746609227 Chr.14: 61767221-61767221 on Build GRCh38
rs758112318 Chr.14: 61767201-61767201 on Build GRCh38
rs756250420 Chr.14: 61766948-61766948 on Build GRCh38
rs774713306 Chr.14: 61767026-61767026 on Build GRCh38
rs758881012 Chr.14: 61766862-61766862 on Build GRCh38
rs368351922 Chr.14: 61767190-61767190 on Build GRCh38
rs941644903 Chr.14: 61767002-61767002 on Build GRCh38
rs746729135 Chr.14: 61767004-61767004 on Build GRCh38
rs753183177 Chr.14: 61766941-61766942 on Build GRCh38
rs377122128 Chr.14: 61766860-61766860 on Build GRCh38
rs374459392 Chr.14: 61767183-61767183 on Build GRCh38
rs547318581 Chr.14: 61766848-61766848 on Build GRCh38
rs768325066 Chr.14: 61767223-61767223 on Build GRCh38
rs372912670 Chr.14: 61766954-61766954 on Build GRCh38
rs745884632 Chr.14: 61767195-61767195 on Build GRCh38
rs529322466 Chr.14: 61766854-61766854 on Build GRCh38
rs752841044 Chr.14: 61767077-61767077 on Build GRCh38

View Full Product Details